Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 19737894-19738385 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALDH3A1
Entrez Gene ID
218
Gene Name
aldehyde dehydrogenase 3 family member A1
Gene Aliases
ALDH3, ALDHIII
UniGene
Hs.531682
Gene Chromosome Location
Chr.17: 19737984-19748433 on Build GRCh38
Transcripts
NM_000691.4 NM_001135167.1 NM_001135168.1 XM_005256522.1 XM_005256523.2 XM_005256524.3 XM_011523730.1 XM_011523731.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5726081 c.1352C>T p.T451I Chr.17: 19738231-19738231 on Build GRCh38
344488 c.1330C>T p.P444S Chr.17: 19738340-19738340 on Build GRCh38
3969881 c.1347+1G>A p.? Chr.17: 19738322-19738322 on Build GRCh38
6598259 c.1303A>G p.N435D Chr.17: 19738367-19738367 on Build GRCh38
5116215 c.1334_1335insC p.S446fs*>9 Chr.17: 19738335-19738336 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs899263904 Chr.17: 19738075-19738075 on Build GRCh38
rs759823213 Chr.17: 19738349-19738349 on Build GRCh38
rs765020206 Chr.17: 19738133-19738133 on Build GRCh38
rs760336161 Chr.17: 19738084-19738091 on Build GRCh38
rs763924725 Chr.17: 19738214-19738214 on Build GRCh38
rs996340543 Chr.17: 19738357-19738357 on Build GRCh38
rs143691519 Chr.17: 19738206-19738206 on Build GRCh38
rs760303323 Chr.17: 19738122-19738122 on Build GRCh38
rs748688486 Chr.17: 19738178-19738178 on Build GRCh38
rs755519068 Chr.17: 19738329-19738329 on Build GRCh38
rs548172023 Chr.17: 19738238-19738238 on Build GRCh38
rs745577678 Chr.17: 19738040-19738040 on Build GRCh38
rs146094687 Chr.17: 19737987-19737987 on Build GRCh38
rs375771662 Chr.17: 19738308-19738308 on Build GRCh38
rs774198203 Chr.17: 19738288-19738288 on Build GRCh38
rs778623472 Chr.17: 19738336-19738336 on Build GRCh38
rs746107983 Chr.17: 19738060-19738060 on Build GRCh38
rs768369860 Chr.17: 19738286-19738286 on Build GRCh38
rs968774241 Chr.17: 19737933-19737933 on Build GRCh38
rs1022055505 Chr.17: 19737944-19737944 on Build GRCh38
rs755250908 Chr.17: 19738260-19738260 on Build GRCh38
rs757438230 Chr.17: 19738055-19738055 on Build GRCh38
rs759044533 Chr.17: 19738274-19738274 on Build GRCh38
rs754122948 Chr.17: 19738251-19738251 on Build GRCh38
rs947911131 Chr.17: 19738036-19738036 on Build GRCh38
rs1042183 Chr.17: 19738008-19738008 on Build GRCh38
rs781349167 Chr.17: 19738057-19738057 on Build GRCh38
rs112263426 Chr.17: 19738009-19738009 on Build GRCh38
rs774212642 Chr.17: 19738216-19738216 on Build GRCh38
rs1021576399 Chr.17: 19737926-19737926 on Build GRCh38
rs200532620 Chr.17: 19738306-19738306 on Build GRCh38
rs774954930 Chr.17: 19738283-19738283 on Build GRCh38
rs532568627 Chr.17: 19738179-19738179 on Build GRCh38
rs200012428 Chr.17: 19738204-19738204 on Build GRCh38
rs779433897 Chr.17: 19738263-19738263 on Build GRCh38
rs753756012 Chr.17: 19738250-19738251 on Build GRCh38
rs555443127 Chr.17: 19738156-19738156 on Build GRCh38
rs554106630 Chr.17: 19738193-19738193 on Build GRCh38
rs759276441 Chr.17: 19738132-19738132 on Build GRCh38
rs564024276 Chr.17: 19738172-19738172 on Build GRCh38
rs1044422041 Chr.17: 19738063-19738063 on Build GRCh38
rs930638393 Chr.17: 19738082-19738082 on Build GRCh38
rs780285968 Chr.17: 19738079-19738079 on Build GRCh38
rs767580208 Chr.17: 19738223-19738223 on Build GRCh38
rs756357538 Chr.17: 19738165-19738165 on Build GRCh38
rs770598190 Chr.17: 19738128-19738128 on Build GRCh38
rs987185807 Chr.17: 19738043-19738043 on Build GRCh38
rs756334935 Chr.17: 19738228-19738228 on Build GRCh38
rs772824617 Chr.17: 19738108-19738108 on Build GRCh38
rs745422678 Chr.17: 19738276-19738276 on Build GRCh38
rs369647404 Chr.17: 19738293-19738293 on Build GRCh38
rs923876356 Chr.17: 19738007-19738007 on Build GRCh38
rs758954521 Chr.17: 19738319-19738319 on Build GRCh38
rs146499259 Chr.17: 19738207-19738207 on Build GRCh38
rs757253344 Chr.17: 19738317-19738317 on Build GRCh38
rs781389564 Chr.17: 19738316-19738316 on Build GRCh38
rs745941149 Chr.17: 19738320-19738320 on Build GRCh38
rs550569625 Chr.17: 19738314-19738314 on Build GRCh38
rs967669929 Chr.17: 19737946-19737946 on Build GRCh38
rs753175531 Chr.17: 19738317-19738317 on Build GRCh38
rs780451866 Chr.17: 19738167-19738167 on Build GRCh38
rs748884426 Chr.17: 19738336-19738336 on Build GRCh38
rs375889932 Chr.17: 19738176-19738176 on Build GRCh38
rs1010102300 Chr.17: 19738233-19738233 on Build GRCh38
rs974457489 Chr.17: 19737975-19737975 on Build GRCh38
rs570592149 Chr.17: 19737956-19737956 on Build GRCh38
rs370379937 Chr.17: 19738322-19738322 on Build GRCh38
rs761724950 Chr.17: 19738292-19738292 on Build GRCh38
rs776289387 Chr.17: 19738181-19738181 on Build GRCh38
rs369145420 Chr.17: 19738273-19738273 on Build GRCh38
rs772975076 Chr.17: 19738341-19738341 on Build GRCh38
rs116087608 Chr.17: 19738301-19738301 on Build GRCh38
rs762937288 Chr.17: 19738138-19738138 on Build GRCh38
rs749894728 Chr.17: 19738358-19738359 on Build GRCh38
rs372494751 Chr.17: 19738307-19738307 on Build GRCh38
rs377018930 Chr.17: 19738282-19738282 on Build GRCh38
rs917520 Chr.17: 19738134-19738134 on Build GRCh38
rs749898582 Chr.17: 19738087-19738087 on Build GRCh38
rs751526560 Chr.17: 19738147-19738147 on Build GRCh38
rs771884353 Chr.17: 19738340-19738340 on Build GRCh38
rs763895608 Chr.17: 19738214-19738214 on Build GRCh38
rs370815018 Chr.17: 19738338-19738338 on Build GRCh38
rs111501205 Chr.17: 19738135-19738135 on Build GRCh38
rs781258287 Chr.17: 19738158-19738158 on Build GRCh38
rs115563042 Chr.17: 19738164-19738164 on Build GRCh38
rs763976487 Chr.17: 19738145-19738145 on Build GRCh38
rs770133924 Chr.17: 19738073-19738073 on Build GRCh38
rs916372967 Chr.17: 19738030-19738030 on Build GRCh38
rs750559783 Chr.17: 19738224-19738224 on Build GRCh38
rs374060290 Chr.17: 19738330-19738330 on Build GRCh38
rs149086635 Chr.17: 19738325-19738325 on Build GRCh38
rs372148589 Chr.17: 19738275-19738275 on Build GRCh38
rs765608757 Chr.17: 19738350-19738350 on Build GRCh38
rs769100782 Chr.17: 19738086-19738086 on Build GRCh38
rs545990205 Chr.17: 19738188-19738188 on Build GRCh38
rs978193412 Chr.17: 19737978-19737978 on Build GRCh38
rs747570938 Chr.17: 19738327-19738327 on Build GRCh38
rs776838494 Chr.17: 19738343-19738343 on Build GRCh38
rs776374305 Chr.17: 19738130-19738130 on Build GRCh38
rs543788431 Chr.17: 19738168-19738168 on Build GRCh38
rs916502665 Chr.17: 19738259-19738259 on Build GRCh38

View Full Product Details