Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 19771815-19772326 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ULK2
Entrez Gene ID
9706
Gene Name
unc-51 like autophagy activating kinase 2
Gene Aliases
ATG1B, Unc51.2
UniGene
Hs.168762
Gene Chromosome Location
Chr.17: 19770828-19867926 on Build GRCh38
Transcripts
NM_001142610.1 NM_014683.3 XM_017025425.1

SNP Detail

SNP ID Chromosome Location
rs942726046 Chr.17: 19771870-19771870 on Build GRCh38
rs111417623 Chr.17: 19772118-19772118 on Build GRCh38
rs530962533 Chr.17: 19772025-19772025 on Build GRCh38
rs553086100 Chr.17: 19772173-19772173 on Build GRCh38
rs9895806 Chr.17: 19772234-19772234 on Build GRCh38
rs548413070 Chr.17: 19771941-19771941 on Build GRCh38
rs1010807238 Chr.17: 19772177-19772177 on Build GRCh38
rs775186443 Chr.17: 19772258-19772258 on Build GRCh38
rs909151458 Chr.17: 19772044-19772044 on Build GRCh38
rs970903663 Chr.17: 19771981-19771981 on Build GRCh38
rs368796948 Chr.17: 19771992-19771992 on Build GRCh38
rs543928725 Chr.17: 19772169-19772169 on Build GRCh38
rs934116805 Chr.17: 19772134-19772134 on Build GRCh38
rs981094015 Chr.17: 19772008-19772008 on Build GRCh38
rs958361095 Chr.17: 19772037-19772037 on Build GRCh38
rs962023316 Chr.17: 19772279-19772279 on Build GRCh38
rs771301559 Chr.17: 19772231-19772231 on Build GRCh38
rs1012614847 Chr.17: 19771967-19771967 on Build GRCh38
rs546416290 Chr.17: 19771850-19771850 on Build GRCh38
rs889751289 Chr.17: 19772140-19772140 on Build GRCh38
rs372721950 Chr.17: 19772058-19772058 on Build GRCh38
rs982718611 Chr.17: 19771994-19771994 on Build GRCh38
rs918665987 Chr.17: 19772109-19772109 on Build GRCh38
rs886997936 Chr.17: 19771928-19771928 on Build GRCh38
rs915246576 Chr.17: 19771862-19771862 on Build GRCh38
rs1016516648 Chr.17: 19771959-19771959 on Build GRCh38
rs1037502163 Chr.17: 19771845-19771845 on Build GRCh38
rs1047313018 Chr.17: 19772130-19772130 on Build GRCh38
rs894145206 Chr.17: 19772183-19772183 on Build GRCh38
rs205110 Chr.17: 19772275-19772275 on Build GRCh38
rs1033275287 Chr.17: 19771945-19771945 on Build GRCh38
rs777995997 Chr.17: 19771983-19771983 on Build GRCh38
rs566573877 Chr.17: 19772186-19772186 on Build GRCh38
rs1039845555 Chr.17: 19771890-19771890 on Build GRCh38
rs1007057920 Chr.17: 19771921-19771921 on Build GRCh38
rs971130981 Chr.17: 19772210-19772210 on Build GRCh38
rs1051082273 Chr.17: 19771916-19771916 on Build GRCh38
rs1024050198 Chr.17: 19771980-19771981 on Build GRCh38
rs144288830 Chr.17: 19772139-19772139 on Build GRCh38
rs934105626 Chr.17: 19772120-19772120 on Build GRCh38
rs971165181 Chr.17: 19772000-19772000 on Build GRCh38
rs897270551 Chr.17: 19771881-19771881 on Build GRCh38
rs972307705 Chr.17: 19772286-19772286 on Build GRCh38
rs957741415 Chr.17: 19771946-19771946 on Build GRCh38
rs998616472 Chr.17: 19772241-19772241 on Build GRCh38
rs1002582474 Chr.17: 19772220-19772220 on Build GRCh38
rs559703610 Chr.17: 19771919-19771919 on Build GRCh38
rs1034105982 Chr.17: 19772229-19772229 on Build GRCh38
rs550890895 Chr.17: 19772038-19772038 on Build GRCh38
rs574870129 Chr.17: 19772300-19772300 on Build GRCh38
rs958372994 Chr.17: 19771962-19771962 on Build GRCh38
rs1006906183 Chr.17: 19772154-19772154 on Build GRCh38
rs998130631 Chr.17: 19771906-19771906 on Build GRCh38
rs940541407 Chr.17: 19771839-19771839 on Build GRCh38
rs940502898 Chr.17: 19772048-19772048 on Build GRCh38
rs568429765 Chr.17: 19771979-19771979 on Build GRCh38
rs1036230058 Chr.17: 19772070-19772070 on Build GRCh38
rs747196906 Chr.17: 19772010-19772010 on Build GRCh38
rs528538855 Chr.17: 19771939-19771939 on Build GRCh38
rs942716501 Chr.17: 19772046-19772046 on Build GRCh38

View Full Product Details