Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 21531316-21531854 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C17orf51
Entrez Gene ID
339263
Gene Name
chromosome 17 open reading frame 51
Gene Aliases
-
UniGene
Hs.514016
Gene Chromosome Location
Chr.17: 21524606-21551678 on Build GRCh38
Transcripts
NM_001113434.4 XM_005256621.3 XM_006721514.2 XM_011523822.2

SNP Detail

SNP ID Chromosome Location
rs931545774 Chr.17: 21531700-21531700 on Build GRCh38
rs58042140 Chr.17: 21531403-21531403 on Build GRCh38
rs782223567 Chr.17: 21531621-21531621 on Build GRCh38
rs940337934 Chr.17: 21531742-21531742 on Build GRCh38
rs576546807 Chr.17: 21531759-21531759 on Build GRCh38
rs992152639 Chr.17: 21531596-21531596 on Build GRCh38
rs1024997514 Chr.17: 21531598-21531598 on Build GRCh38
rs983448769 Chr.17: 21531715-21531715 on Build GRCh38
rs782616455 Chr.17: 21531578-21531578 on Build GRCh38
rs1048699872 Chr.17: 21531416-21531416 on Build GRCh38
rs548642107 Chr.17: 21531814-21531814 on Build GRCh38
rs115464229 Chr.17: 21531498-21531498 on Build GRCh38
rs782451795 Chr.17: 21531488-21531488 on Build GRCh38
rs558328841 Chr.17: 21531752-21531752 on Build GRCh38
rs566589863 Chr.17: 21531420-21531420 on Build GRCh38
rs188635680 Chr.17: 21531360-21531360 on Build GRCh38
rs538205685 Chr.17: 21531531-21531531 on Build GRCh38
rs782554672 Chr.17: 21531453-21531453 on Build GRCh38
rs782678606 Chr.17: 21531479-21531479 on Build GRCh38
rs781856171 Chr.17: 21531452-21531452 on Build GRCh38
rs535808228 Chr.17: 21531622-21531622 on Build GRCh38
rs768216468 Chr.17: 21531419-21531419 on Build GRCh38
rs568752046 Chr.17: 21531595-21531595 on Build GRCh38
rs780008502 Chr.17: 21531362-21531362 on Build GRCh38
rs992205370 Chr.17: 21531381-21531381 on Build GRCh38
rs931730651 Chr.17: 21531821-21531821 on Build GRCh38
rs548349518 Chr.17: 21531408-21531408 on Build GRCh38
rs980930161 Chr.17: 21531405-21531405 on Build GRCh38
rs948214844 Chr.17: 21531404-21531404 on Build GRCh38
rs903737924 Chr.17: 21531516-21531516 on Build GRCh38
rs141467582 Chr.17: 21531820-21531820 on Build GRCh38
rs1040084298 Chr.17: 21531798-21531798 on Build GRCh38
rs34858935 Chr.17: 21531501-21531502 on Build GRCh38
rs546383485 Chr.17: 21531713-21531713 on Build GRCh38
rs931570994 Chr.17: 21531413-21531413 on Build GRCh38
rs556619837 Chr.17: 21531539-21531539 on Build GRCh38
rs908829608 Chr.17: 21531727-21531727 on Build GRCh38
rs570965075 Chr.17: 21531511-21531511 on Build GRCh38
rs1030961959 Chr.17: 21531576-21531576 on Build GRCh38
rs920177014 Chr.17: 21531407-21531407 on Build GRCh38
rs995990409 Chr.17: 21531510-21531510 on Build GRCh38
rs980877714 Chr.17: 21531645-21531645 on Build GRCh38
rs76942678 Chr.17: 21531629-21531629 on Build GRCh38
rs901634359 Chr.17: 21531500-21531500 on Build GRCh38
rs782192698 Chr.17: 21531481-21531481 on Build GRCh38
rs146602421 Chr.17: 21531699-21531699 on Build GRCh38

View Full Product Details