Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 28388091-28388595 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SARM1
Entrez Gene ID
23098
Gene Name
sterile alpha and TIR motif containing 1
Gene Aliases
MyD88-5, SAMD2, SARM
UniGene
Hs.743510
Gene Chromosome Location
Chr.17: 28371662-28401045 on Build GRCh38
Transcripts
NM_015077.3

SNP Detail

SNP ID Chromosome Location
rs948740126 Chr.17: 28388519-28388519 on Build GRCh38
rs924706333 Chr.17: 28388197-28388197 on Build GRCh38
rs898412619 Chr.17: 28388566-28388566 on Build GRCh38
rs977609076 Chr.17: 28388417-28388417 on Build GRCh38
rs782321764 Chr.17: 28388450-28388450 on Build GRCh38
rs368957811 Chr.17: 28388289-28388289 on Build GRCh38
rs782083292 Chr.17: 28388478-28388478 on Build GRCh38
rs139403866 Chr.17: 28388190-28388190 on Build GRCh38
rs975670103 Chr.17: 28388156-28388156 on Build GRCh38
rs782159743 Chr.17: 28388459-28388459 on Build GRCh38
rs782047820 Chr.17: 28388429-28388429 on Build GRCh38
rs782719843 Chr.17: 28388133-28388133 on Build GRCh38
rs942891342 Chr.17: 28388144-28388144 on Build GRCh38
rs782008489 Chr.17: 28388451-28388451 on Build GRCh38
rs529921858 Chr.17: 28388565-28388565 on Build GRCh38
rs782559954 Chr.17: 28388389-28388389 on Build GRCh38
rs782225790 Chr.17: 28388534-28388534 on Build GRCh38
rs782581724 Chr.17: 28388548-28388548 on Build GRCh38
rs368394015 Chr.17: 28388493-28388493 on Build GRCh38
rs938798379 Chr.17: 28388217-28388217 on Build GRCh38
rs781893510 Chr.17: 28388308-28388309 on Build GRCh38
rs782407924 Chr.17: 28388398-28388398 on Build GRCh38
rs782753946 Chr.17: 28388460-28388460 on Build GRCh38
rs370037595 Chr.17: 28388308-28388308 on Build GRCh38
rs782749087 Chr.17: 28388482-28388482 on Build GRCh38
rs947556072 Chr.17: 28388181-28388181 on Build GRCh38
rs782456355 Chr.17: 28388487-28388487 on Build GRCh38
rs782500070 Chr.17: 28388368-28388368 on Build GRCh38
rs914716164 Chr.17: 28388178-28388178 on Build GRCh38
rs147478392 Chr.17: 28388562-28388562 on Build GRCh38
rs562369897 Chr.17: 28388563-28388563 on Build GRCh38
rs781906310 Chr.17: 28388504-28388504 on Build GRCh38
rs995289515 Chr.17: 28388292-28388292 on Build GRCh38
rs782381095 Chr.17: 28388303-28388303 on Build GRCh38
rs782246298 Chr.17: 28388425-28388425 on Build GRCh38
rs782491086 Chr.17: 28388385-28388385 on Build GRCh38
rs782605765 Chr.17: 28388301-28388301 on Build GRCh38
rs782648225 Chr.17: 28388532-28388532 on Build GRCh38
rs141324431 Chr.17: 28388486-28388486 on Build GRCh38
rs782648839 Chr.17: 28388369-28388369 on Build GRCh38
rs539229444 Chr.17: 28388252-28388252 on Build GRCh38
rs781993643 Chr.17: 28388313-28388313 on Build GRCh38
rs782172400 Chr.17: 28388235-28388235 on Build GRCh38
rs199517604 Chr.17: 28388521-28388521 on Build GRCh38
rs781829395 Chr.17: 28388379-28388379 on Build GRCh38
rs782549886 Chr.17: 28388420-28388421 on Build GRCh38
rs914807846 Chr.17: 28388322-28388322 on Build GRCh38
rs1037697118 Chr.17: 28388531-28388531 on Build GRCh38
rs571724138 Chr.17: 28388248-28388248 on Build GRCh38
rs35201495 Chr.17: 28388422-28388422 on Build GRCh38
rs782196205 Chr.17: 28388394-28388394 on Build GRCh38
rs781783820 Chr.17: 28388333-28388333 on Build GRCh38
rs782432477 Chr.17: 28388541-28388541 on Build GRCh38
rs782534777 Chr.17: 28388288-28388288 on Build GRCh38
rs373002076 Chr.17: 28388300-28388300 on Build GRCh38
rs1044973636 Chr.17: 28388276-28388276 on Build GRCh38
rs781797528 Chr.17: 28388257-28388257 on Build GRCh38
rs781817572 Chr.17: 28388249-28388249 on Build GRCh38
rs782001345 Chr.17: 28388474-28388474 on Build GRCh38
rs200378586 Chr.17: 28388564-28388564 on Build GRCh38
rs144683600 Chr.17: 28388293-28388293 on Build GRCh38
rs782740249 Chr.17: 28388318-28388318 on Build GRCh38
rs111581438 Chr.17: 28388355-28388355 on Build GRCh38
rs782340970 Chr.17: 28388573-28388573 on Build GRCh38
rs557953224 Chr.17: 28388304-28388304 on Build GRCh38
rs782806850 Chr.17: 28388348-28388348 on Build GRCh38
rs782078420 Chr.17: 28388314-28388314 on Build GRCh38
rs150052156 Chr.17: 28388256-28388256 on Build GRCh38
rs564784908 Chr.17: 28388365-28388365 on Build GRCh38

View Full Product Details