Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 28737848-28738341 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NEK8
Entrez Gene ID
284086
Gene Name
NIMA related kinase 8
Gene Aliases
JCK, NEK12A, NPHP9, RHPD2
UniGene
Hs.448468
Gene Chromosome Location
Chr.17: 28727899-28743457 on Build GRCh38
Transcripts
NM_178170.2 XM_011524638.2 XM_011524640.2 XM_017024499.1 XM_017024500.1 XM_017024501.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5798220 c.969C>A p.T323T Chr.17: 28737898-28737898 on Build GRCh38
6724541 c.1012G>T p.A338S Chr.17: 28737941-28737941 on Build GRCh38
5810873 c.1112G>T p.G371V Chr.17: 28738135-28738135 on Build GRCh38
4065053 c.948C>A p.A316A Chr.17: 28737877-28737877 on Build GRCh38
5499294 c.1026G>T p.T342T Chr.17: 28737955-28737955 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374930363 Chr.17: 28738204-28738204 on Build GRCh38
rs375613242 Chr.17: 28738256-28738256 on Build GRCh38
rs368061384 Chr.17: 28738255-28738255 on Build GRCh38
rs770032175 Chr.17: 28738091-28738091 on Build GRCh38
rs749011475 Chr.17: 28737970-28737970 on Build GRCh38
rs776765761 Chr.17: 28737946-28737946 on Build GRCh38
rs1027208322 Chr.17: 28737985-28737985 on Build GRCh38
rs747712613 Chr.17: 28738234-28738235 on Build GRCh38
rs758754646 Chr.17: 28737894-28737894 on Build GRCh38
rs146989750 Chr.17: 28738168-28738168 on Build GRCh38
rs760766665 Chr.17: 28737942-28737942 on Build GRCh38
rs368688884 Chr.17: 28738101-28738101 on Build GRCh38
rs779393817 Chr.17: 28737901-28737901 on Build GRCh38
rs889694432 Chr.17: 28737974-28737974 on Build GRCh38
rs775731270 Chr.17: 28738100-28738100 on Build GRCh38
rs371295223 Chr.17: 28737897-28737897 on Build GRCh38
rs776828602 Chr.17: 28737991-28737991 on Build GRCh38
rs751987503 Chr.17: 28737964-28737964 on Build GRCh38
rs56286645 Chr.17: 28737905-28737905 on Build GRCh38
rs959589808 Chr.17: 28738104-28738104 on Build GRCh38
rs3809797 Chr.17: 28738193-28738193 on Build GRCh38
rs773847705 Chr.17: 28738269-28738269 on Build GRCh38
rs757916936 Chr.17: 28738137-28738137 on Build GRCh38
rs367654888 Chr.17: 28738136-28738136 on Build GRCh38
rs755900226 Chr.17: 28738221-28738221 on Build GRCh38
rs763411744 Chr.17: 28738017-28738017 on Build GRCh38
rs754385014 Chr.17: 28737925-28737926 on Build GRCh38
rs773902843 Chr.17: 28738190-28738190 on Build GRCh38
rs371406871 Chr.17: 28738200-28738200 on Build GRCh38
rs1052596453 Chr.17: 28738127-28738127 on Build GRCh38
rs199933041 Chr.17: 28737984-28737984 on Build GRCh38
rs777152445 Chr.17: 28738141-28738141 on Build GRCh38
rs569568989 Chr.17: 28738008-28738008 on Build GRCh38
rs760874541 Chr.17: 28738003-28738003 on Build GRCh38
rs199962228 Chr.17: 28737917-28737917 on Build GRCh38
rs752452156 Chr.17: 28738125-28738125 on Build GRCh38
rs767947984 Chr.17: 28737950-28737950 on Build GRCh38
rs145862009 Chr.17: 28738209-28738209 on Build GRCh38
rs767840061 Chr.17: 28738053-28738053 on Build GRCh38
rs374298548 Chr.17: 28738070-28738070 on Build GRCh38
rs200489118 Chr.17: 28737963-28737963 on Build GRCh38
rs191806152 Chr.17: 28738276-28738276 on Build GRCh38
rs762225317 Chr.17: 28738021-28738021 on Build GRCh38
rs536256211 Chr.17: 28738019-28738019 on Build GRCh38
rs983735559 Chr.17: 28737890-28737890 on Build GRCh38
rs750419550 Chr.17: 28738220-28738220 on Build GRCh38
rs757806007 Chr.17: 28738270-28738270 on Build GRCh38
rs778519415 Chr.17: 28738059-28738059 on Build GRCh38
rs777749160 Chr.17: 28738068-28738068 on Build GRCh38
rs769454263 Chr.17: 28738251-28738251 on Build GRCh38
rs757603639 Chr.17: 28737967-28737967 on Build GRCh38
rs200428176 Chr.17: 28738119-28738119 on Build GRCh38
rs767422789 Chr.17: 28738216-28738216 on Build GRCh38
rs775567721 Chr.17: 28738013-28738013 on Build GRCh38
rs772333473 Chr.17: 28738262-28738262 on Build GRCh38
rs151217692 Chr.17: 28737973-28737973 on Build GRCh38
rs773075496 Chr.17: 28737940-28737940 on Build GRCh38
rs759758319 Chr.17: 28737994-28737994 on Build GRCh38
rs771928671 Chr.17: 28737929-28737929 on Build GRCh38
rs763610690 Chr.17: 28738206-28738206 on Build GRCh38
rs199853008 Chr.17: 28738189-28738189 on Build GRCh38
rs771364513 Chr.17: 28738280-28738280 on Build GRCh38
rs142234912 Chr.17: 28737914-28737914 on Build GRCh38
rs758918296 Chr.17: 28737954-28737954 on Build GRCh38
rs752221099 Chr.17: 28738065-28738065 on Build GRCh38
rs368639779 Chr.17: 28737955-28737955 on Build GRCh38
rs751089803 Chr.17: 28737893-28737898 on Build GRCh38
rs749473321 Chr.17: 28738243-28738243 on Build GRCh38
rs775138270 Chr.17: 28738202-28738202 on Build GRCh38
rs372172665 Chr.17: 28737968-28737968 on Build GRCh38
rs780241960 Chr.17: 28738170-28738170 on Build GRCh38
rs376531637 Chr.17: 28737972-28737972 on Build GRCh38
rs1004689857 Chr.17: 28738052-28738052 on Build GRCh38
rs375211540 Chr.17: 28738058-28738058 on Build GRCh38
rs374175041 Chr.17: 28738123-28738123 on Build GRCh38
rs370940925 Chr.17: 28738060-28738060 on Build GRCh38
rs138066977 Chr.17: 28738171-28738171 on Build GRCh38
rs140363905 Chr.17: 28737975-28737975 on Build GRCh38
rs982291329 Chr.17: 28738079-28738079 on Build GRCh38
rs574347342 Chr.17: 28738208-28738208 on Build GRCh38
rs753256955 Chr.17: 28737882-28737882 on Build GRCh38
rs778201727 Chr.17: 28737895-28737895 on Build GRCh38
rs780916071 Chr.17: 28738171-28738172 on Build GRCh38
rs186958324 Chr.17: 28738223-28738223 on Build GRCh38
rs954240143 Chr.17: 28738111-28738111 on Build GRCh38
rs748643938 Chr.17: 28738183-28738183 on Build GRCh38
rs906602387 Chr.17: 28738257-28738257 on Build GRCh38
rs757891805 Chr.17: 28738066-28738066 on Build GRCh38
rs780977790 Chr.17: 28738083-28738083 on Build GRCh38
rs779830257 Chr.17: 28738239-28738239 on Build GRCh38
rs183679495 Chr.17: 28738069-28738069 on Build GRCh38
rs753134520 Chr.17: 28737952-28737952 on Build GRCh38
rs768677742 Chr.17: 28738102-28738102 on Build GRCh38
rs554563297 Chr.17: 28738055-28738055 on Build GRCh38
rs751455589 Chr.17: 28738143-28738143 on Build GRCh38
rs765364445 Chr.17: 28737951-28737951 on Build GRCh38
rs755816415 Chr.17: 28737900-28737900 on Build GRCh38
rs202091769 Chr.17: 28738109-28738109 on Build GRCh38
rs780922648 Chr.17: 28738169-28738169 on Build GRCh38
rs746794190 Chr.17: 28737978-28737978 on Build GRCh38
rs201321032 Chr.17: 28738115-28738115 on Build GRCh38
rs202102961 Chr.17: 28738116-28738116 on Build GRCh38
rs200972000 Chr.17: 28737906-28737906 on Build GRCh38

View Full Product Details