Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 28858488-28858987 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ERAL1
Entrez Gene ID
26284
Gene Name
Era like 12S mitochondrial rRNA chaperone 1
Gene Aliases
CEGA, ERA, ERA-W, ERAL1A, H-ERA, HERA-A, HERA-B
UniGene
Hs.3426
Gene Chromosome Location
Chr.17: 28854957-28861067 on Build GRCh38
Transcripts
NM_001317985.1 NM_001317986.1 NM_005702.3 NR_134328.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
269114 c.931C>A p.L311I Chr.17: 28858795-28858795 on Build GRCh38
5004347 c.712-6G>T p.? Chr.17: 28858570-28858570 on Build GRCh38
6031551 c.870_871delTG p.V291fs*50 Chr.17: 28858734-28858735 on Build GRCh38
5988773 c.909G>T p.K303N Chr.17: 28858773-28858773 on Build GRCh38
7152541 c.970C>G p.L324V Chr.17: 28858973-28858973 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746762561 Chr.17: 28858576-28858576 on Build GRCh38
rs1045419991 Chr.17: 28858579-28858579 on Build GRCh38
rs778417271 Chr.17: 28858734-28858735 on Build GRCh38
rs368966727 Chr.17: 28858589-28858589 on Build GRCh38
rs750796585 Chr.17: 28858632-28858632 on Build GRCh38
rs924595592 Chr.17: 28858806-28858806 on Build GRCh38
rs35863803 Chr.17: 28858523-28858524 on Build GRCh38
rs147448803 Chr.17: 28858767-28858767 on Build GRCh38
rs201357781 Chr.17: 28858754-28858754 on Build GRCh38
rs912893351 Chr.17: 28858567-28858567 on Build GRCh38
rs144133158 Chr.17: 28858866-28858866 on Build GRCh38
rs763661240 Chr.17: 28858915-28858915 on Build GRCh38
rs114192384 Chr.17: 28858587-28858587 on Build GRCh38
rs757166533 Chr.17: 28858670-28858670 on Build GRCh38
rs774547396 Chr.17: 28858517-28858517 on Build GRCh38
rs775706245 Chr.17: 28858604-28858604 on Build GRCh38
rs753485526 Chr.17: 28858536-28858536 on Build GRCh38
rs374587408 Chr.17: 28858835-28858835 on Build GRCh38
rs754646187 Chr.17: 28858549-28858549 on Build GRCh38
rs761277916 Chr.17: 28858930-28858930 on Build GRCh38
rs990472696 Chr.17: 28858845-28858845 on Build GRCh38
rs764479923 Chr.17: 28858727-28858727 on Build GRCh38
rs756740739 Chr.17: 28858545-28858546 on Build GRCh38
rs2242345 Chr.17: 28858809-28858809 on Build GRCh38
rs775166713 Chr.17: 28858766-28858766 on Build GRCh38
rs866099383 Chr.17: 28858792-28858792 on Build GRCh38
rs558388003 Chr.17: 28858696-28858696 on Build GRCh38
rs142986474 Chr.17: 28858684-28858684 on Build GRCh38
rs144472222 Chr.17: 28858815-28858815 on Build GRCh38
rs911115381 Chr.17: 28858530-28858530 on Build GRCh38
rs769021353 Chr.17: 28858608-28858608 on Build GRCh38
rs376842462 Chr.17: 28858569-28858569 on Build GRCh38
rs767410240 Chr.17: 28858803-28858803 on Build GRCh38
rs374024882 Chr.17: 28858660-28858660 on Build GRCh38
rs773761350 Chr.17: 28858837-28858837 on Build GRCh38
rs774312450 Chr.17: 28858720-28858720 on Build GRCh38
rs766215501 Chr.17: 28858961-28858961 on Build GRCh38
rs750346199 Chr.17: 28858948-28858948 on Build GRCh38
rs954424200 Chr.17: 28858637-28858637 on Build GRCh38
rs745723656 Chr.17: 28858688-28858688 on Build GRCh38
rs375611257 Chr.17: 28858510-28858510 on Build GRCh38
rs183991298 Chr.17: 28858619-28858619 on Build GRCh38
rs897788776 Chr.17: 28858707-28858707 on Build GRCh38
rs763611007 Chr.17: 28858770-28858770 on Build GRCh38
rs373363951 Chr.17: 28858552-28858552 on Build GRCh38
rs144284910 Chr.17: 28858642-28858642 on Build GRCh38
rs749468371 Chr.17: 28858819-28858819 on Build GRCh38
rs756389430 Chr.17: 28858694-28858694 on Build GRCh38
rs577080134 Chr.17: 28858832-28858832 on Build GRCh38
rs756138225 Chr.17: 28858950-28858950 on Build GRCh38
rs780707242 Chr.17: 28858938-28858938 on Build GRCh38
rs757153779 Chr.17: 28858785-28858785 on Build GRCh38
rs140743229 Chr.17: 28858654-28858654 on Build GRCh38
rs768738574 Chr.17: 28858716-28858716 on Build GRCh38
rs577237073 Chr.17: 28858811-28858811 on Build GRCh38
rs771246520 Chr.17: 28858841-28858841 on Build GRCh38
rs773897573 Chr.17: 28858924-28858924 on Build GRCh38
rs899379908 Chr.17: 28858843-28858843 on Build GRCh38
rs1057446505 Chr.17: 28858810-28858810 on Build GRCh38
rs373273950 Chr.17: 28858609-28858609 on Build GRCh38
rs761088895 Chr.17: 28858758-28858758 on Build GRCh38
rs867244332 Chr.17: 28858553-28858553 on Build GRCh38
rs1057246408 Chr.17: 28858522-28858522 on Build GRCh38
rs371302748 Chr.17: 28858833-28858833 on Build GRCh38
rs568931390 Chr.17: 28858513-28858513 on Build GRCh38
rs752210661 Chr.17: 28858563-28858563 on Build GRCh38
rs770800247 Chr.17: 28858577-28858577 on Build GRCh38
rs748627987 Chr.17: 28858725-28858725 on Build GRCh38
rs759124850 Chr.17: 28858639-28858639 on Build GRCh38
rs772458858 Chr.17: 28858750-28858750 on Build GRCh38
rs767529868 Chr.17: 28858940-28858940 on Build GRCh38
rs749347644 Chr.17: 28858700-28858700 on Build GRCh38
rs964834535 Chr.17: 28858698-28858698 on Build GRCh38
rs751045543 Chr.17: 28858783-28858783 on Build GRCh38
rs767871960 Chr.17: 28858624-28858624 on Build GRCh38
rs756602406 Chr.17: 28858643-28858643 on Build GRCh38
rs758984925 Chr.17: 28858511-28858511 on Build GRCh38
rs758330184 Chr.17: 28858565-28858565 on Build GRCh38
rs993691817 Chr.17: 28858869-28858869 on Build GRCh38
rs553726243 Chr.17: 28858824-28858824 on Build GRCh38
rs754603789 Chr.17: 28858884-28858884 on Build GRCh38
rs377478694 Chr.17: 28858641-28858641 on Build GRCh38
rs113900197 Chr.17: 28858848-28858848 on Build GRCh38
rs762253323 Chr.17: 28858617-28858617 on Build GRCh38
rs149599529 Chr.17: 28858616-28858616 on Build GRCh38
rs747348206 Chr.17: 28858840-28858840 on Build GRCh38
rs762609894 Chr.17: 28858870-28858870 on Build GRCh38
rs540949990 Chr.17: 28858834-28858834 on Build GRCh38
rs763181209 Chr.17: 28858606-28858606 on Build GRCh38
rs764852414 Chr.17: 28858762-28858762 on Build GRCh38

View Full Product Details