Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 42991443-42991936 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RUNDC1
Entrez Gene ID
146923
Gene Name
RUN domain containing 1
Gene Aliases
-
UniGene
Hs.632255
Gene Chromosome Location
Chr.17: 42980510-42993692 on Build GRCh38
Transcripts
NM_001321381.1 NM_173079.3 XM_005257078.3 XM_005257080.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3795647 c.1646A>G p.Q549R Chr.17: 42991520-42991520 on Build GRCh38
3189905 c.1810C>T p.R604C Chr.17: 42991684-42991684 on Build GRCh38
6300766 c.1776C>G p.S592R Chr.17: 42991650-42991650 on Build GRCh38
5134512 c.1587delT p.F530fs*9 Chr.17: 42991461-42991461 on Build GRCh38
6770601 c.1595G>A p.R532H Chr.17: 42991469-42991469 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766700541 Chr.17: 42991624-42991624 on Build GRCh38
rs754919339 Chr.17: 42991481-42991481 on Build GRCh38
rs751166538 Chr.17: 42991663-42991663 on Build GRCh38
rs747650861 Chr.17: 42991497-42991497 on Build GRCh38
rs542521733 Chr.17: 42991752-42991752 on Build GRCh38
rs779392796 Chr.17: 42991695-42991695 on Build GRCh38
rs544725908 Chr.17: 42991780-42991780 on Build GRCh38
rs758033721 Chr.17: 42991711-42991711 on Build GRCh38
rs776720498 Chr.17: 42991748-42991748 on Build GRCh38
rs1047878956 Chr.17: 42991842-42991842 on Build GRCh38
rs773251964 Chr.17: 42991721-42991721 on Build GRCh38
rs756385044 Chr.17: 42991471-42991471 on Build GRCh38
rs369542823 Chr.17: 42991698-42991698 on Build GRCh38
rs375833434 Chr.17: 42991510-42991510 on Build GRCh38
rs533614478 Chr.17: 42991796-42991796 on Build GRCh38
rs895870657 Chr.17: 42991858-42991858 on Build GRCh38
rs141180839 Chr.17: 42991532-42991532 on Build GRCh38
rs768916891 Chr.17: 42991685-42991685 on Build GRCh38
rs753619133 Chr.17: 42991573-42991573 on Build GRCh38
rs759277627 Chr.17: 42991607-42991607 on Build GRCh38
rs866004705 Chr.17: 42991745-42991745 on Build GRCh38
rs761831894 Chr.17: 42991468-42991468 on Build GRCh38
rs767660129 Chr.17: 42991469-42991469 on Build GRCh38
rs772125890 Chr.17: 42991563-42991563 on Build GRCh38
rs766156837 Chr.17: 42991474-42991474 on Build GRCh38
rs756924818 Chr.17: 42991665-42991665 on Build GRCh38
rs76551330 Chr.17: 42991770-42991770 on Build GRCh38
rs55857539 Chr.17: 42991843-42991843 on Build GRCh38
rs749602274 Chr.17: 42991684-42991684 on Build GRCh38
rs142985946 Chr.17: 42991888-42991888 on Build GRCh38
rs1000877143 Chr.17: 42991892-42991892 on Build GRCh38
rs116930490 Chr.17: 42991758-42991758 on Build GRCh38
rs368354222 Chr.17: 42991753-42991753 on Build GRCh38
rs769168134 Chr.17: 42991536-42991536 on Build GRCh38
rs528009638 Chr.17: 42991877-42991877 on Build GRCh38
rs112582719 Chr.17: 42991557-42991557 on Build GRCh38
rs757983154 Chr.17: 42991506-42991506 on Build GRCh38
rs752585544 Chr.17: 42991633-42991633 on Build GRCh38
rs770751462 Chr.17: 42991522-42991522 on Build GRCh38
rs369376117 Chr.17: 42991493-42991493 on Build GRCh38
rs113828381 Chr.17: 42991648-42991648 on Build GRCh38
rs147605440 Chr.17: 42991571-42991571 on Build GRCh38
rs764924991 Chr.17: 42991755-42991755 on Build GRCh38
rs555965831 Chr.17: 42991482-42991482 on Build GRCh38
rs201943128 Chr.17: 42991727-42991727 on Build GRCh38
rs567119897 Chr.17: 42991470-42991470 on Build GRCh38
rs774726247 Chr.17: 42991537-42991537 on Build GRCh38
rs970676957 Chr.17: 42991615-42991616 on Build GRCh38
rs529433902 Chr.17: 42991908-42991908 on Build GRCh38
rs773346413 Chr.17: 42991567-42991567 on Build GRCh38
rs148971671 Chr.17: 42991792-42991792 on Build GRCh38
rs1014427374 Chr.17: 42991863-42991863 on Build GRCh38
rs746659117 Chr.17: 42991515-42991515 on Build GRCh38
rs375753180 Chr.17: 42991639-42991639 on Build GRCh38
rs560629614 Chr.17: 42991844-42991844 on Build GRCh38
rs960236076 Chr.17: 42991543-42991543 on Build GRCh38
rs771141494 Chr.17: 42991731-42991731 on Build GRCh38
rs200217728 Chr.17: 42991475-42991475 on Build GRCh38
rs549386404 Chr.17: 42991883-42991883 on Build GRCh38
rs774442115 Chr.17: 42991466-42991466 on Build GRCh38
rs369271006 Chr.17: 42991640-42991640 on Build GRCh38
rs3744241 Chr.17: 42991570-42991570 on Build GRCh38
rs147773660 Chr.17: 42991523-42991523 on Build GRCh38
rs933845516 Chr.17: 42991715-42991715 on Build GRCh38
rs35176532 Chr.17: 42991588-42991589 on Build GRCh38
rs764151999 Chr.17: 42991768-42991768 on Build GRCh38
rs780741452 Chr.17: 42991675-42991675 on Build GRCh38
rs537970489 Chr.17: 42991479-42991479 on Build GRCh38

View Full Product Details