Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 43013179-43013673 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IFI35
Entrez Gene ID
3430
Gene Name
interferon induced protein 35
Gene Aliases
IFP35
UniGene
Hs.632258
Gene Chromosome Location
Chr.17: 43006725-43014459 on Build GRCh38
Transcripts
NM_005533.4 XM_005257302.4 XM_017024584.1 XM_017024585.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
436659 c.345C>G p.P115P Chr.17: 43013343-43013343 on Build GRCh38
4168391 c.445A>C p.S149R Chr.17: 43013539-43013539 on Build GRCh38
4066732 c.516C>T p.D172D Chr.17: 43013610-43013610 on Build GRCh38
71127 c.517G>T p.V173F Chr.17: 43013611-43013611 on Build GRCh38
4843897 c.306C>G p.I102M Chr.17: 43013304-43013304 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766537555 Chr.17: 43013375-43013375 on Build GRCh38
rs138975410 Chr.17: 43013625-43013625 on Build GRCh38
rs770379286 Chr.17: 43013621-43013621 on Build GRCh38
rs140488187 Chr.17: 43013611-43013611 on Build GRCh38
rs748086140 Chr.17: 43013224-43013224 on Build GRCh38
rs370544876 Chr.17: 43013604-43013604 on Build GRCh38
rs748921534 Chr.17: 43013558-43013558 on Build GRCh38
rs376147900 Chr.17: 43013629-43013629 on Build GRCh38
rs775576765 Chr.17: 43013542-43013544 on Build GRCh38
rs758201555 Chr.17: 43013407-43013407 on Build GRCh38
rs961121303 Chr.17: 43013584-43013584 on Build GRCh38
rs776729572 Chr.17: 43013479-43013479 on Build GRCh38
rs138159139 Chr.17: 43013610-43013610 on Build GRCh38
rs749011342 Chr.17: 43013460-43013460 on Build GRCh38
rs746942241 Chr.17: 43013417-43013417 on Build GRCh38
rs772660054 Chr.17: 43013233-43013233 on Build GRCh38
rs772563305 Chr.17: 43013468-43013468 on Build GRCh38
rs751379735 Chr.17: 43013505-43013505 on Build GRCh38
rs764333198 Chr.17: 43013580-43013580 on Build GRCh38
rs762859746 Chr.17: 43013633-43013634 on Build GRCh38
rs763115088 Chr.17: 43013576-43013576 on Build GRCh38
rs768083989 Chr.17: 43013365-43013365 on Build GRCh38
rs199510319 Chr.17: 43013640-43013640 on Build GRCh38
rs924751797 Chr.17: 43013569-43013569 on Build GRCh38
rs750468739 Chr.17: 43013303-43013303 on Build GRCh38
rs906327387 Chr.17: 43013394-43013394 on Build GRCh38
rs779686162 Chr.17: 43013555-43013555 on Build GRCh38
rs1021825359 Chr.17: 43013542-43013542 on Build GRCh38
rs201068538 Chr.17: 43013601-43013601 on Build GRCh38
rs745513010 Chr.17: 43013333-43013333 on Build GRCh38
rs746463710 Chr.17: 43013326-43013326 on Build GRCh38
rs768809782 Chr.17: 43013639-43013639 on Build GRCh38
rs778231273 Chr.17: 43013563-43013563 on Build GRCh38
rs759166671 Chr.17: 43013276-43013276 on Build GRCh38
rs369559189 Chr.17: 43013575-43013575 on Build GRCh38
rs747914826 Chr.17: 43013321-43013321 on Build GRCh38
rs369228705 Chr.17: 43013614-43013614 on Build GRCh38
rs769421604 Chr.17: 43013439-43013439 on Build GRCh38
rs752469643 Chr.17: 43013405-43013405 on Build GRCh38
rs201830704 Chr.17: 43013320-43013320 on Build GRCh38
rs753911866 Chr.17: 43013381-43013381 on Build GRCh38
rs369524898 Chr.17: 43013304-43013304 on Build GRCh38
rs776299342 Chr.17: 43013257-43013257 on Build GRCh38
rs147582201 Chr.17: 43013373-43013373 on Build GRCh38
rs762166248 Chr.17: 43013644-43013644 on Build GRCh38
rs767407648 Chr.17: 43013597-43013597 on Build GRCh38
rs189813077 Chr.17: 43013634-43013634 on Build GRCh38
rs148896168 Chr.17: 43013592-43013592 on Build GRCh38
rs769354659 Chr.17: 43013356-43013356 on Build GRCh38
rs760145503 Chr.17: 43013573-43013573 on Build GRCh38
rs968901082 Chr.17: 43013550-43013550 on Build GRCh38
rs773112261 Chr.17: 43013652-43013652 on Build GRCh38
rs1006152161 Chr.17: 43013465-43013465 on Build GRCh38
rs914851029 Chr.17: 43013219-43013219 on Build GRCh38
rs887726895 Chr.17: 43013463-43013463 on Build GRCh38
rs377676655 Chr.17: 43013559-43013559 on Build GRCh38
rs1041538049 Chr.17: 43013366-43013366 on Build GRCh38
rs747405142 Chr.17: 43013568-43013568 on Build GRCh38
rs755570355 Chr.17: 43013536-43013536 on Build GRCh38
rs757746415 Chr.17: 43013200-43013211 on Build GRCh38
rs588703 Chr.17: 43013476-43013476 on Build GRCh38
rs778638664 Chr.17: 43013221-43013221 on Build GRCh38
rs976936685 Chr.17: 43013293-43013293 on Build GRCh38
rs748179486 Chr.17: 43013406-43013406 on Build GRCh38
rs150695787 Chr.17: 43013615-43013615 on Build GRCh38
rs780997177 Chr.17: 43013531-43013531 on Build GRCh38
rs757148523 Chr.17: 43013507-43013507 on Build GRCh38
rs771950606 Chr.17: 43013231-43013231 on Build GRCh38
rs773755551 Chr.17: 43013471-43013471 on Build GRCh38
rs768170062 Chr.17: 43013301-43013301 on Build GRCh38
rs779312766 Chr.17: 43013246-43013247 on Build GRCh38
rs777017425 Chr.17: 43013572-43013572 on Build GRCh38
rs570575342 Chr.17: 43013397-43013397 on Build GRCh38
rs73983785 Chr.17: 43013415-43013415 on Build GRCh38
rs763844158 Chr.17: 43013498-43013498 on Build GRCh38
rs776276033 Chr.17: 43013264-43013264 on Build GRCh38
rs780705521 Chr.17: 43013419-43013419 on Build GRCh38
rs73983784 Chr.17: 43013245-43013245 on Build GRCh38
rs769757979 Chr.17: 43013574-43013574 on Build GRCh38
rs376682517 Chr.17: 43013327-43013327 on Build GRCh38
rs145578882 Chr.17: 43013602-43013602 on Build GRCh38
rs754088504 Chr.17: 43013317-43013317 on Build GRCh38
rs755193476 Chr.17: 43013318-43013318 on Build GRCh38
rs376711701 Chr.17: 43013241-43013241 on Build GRCh38
rs779590467 Chr.17: 43013458-43013458 on Build GRCh38
rs201256493 Chr.17: 43013378-43013378 on Build GRCh38
rs765440953 Chr.17: 43013485-43013485 on Build GRCh38
rs904640323 Chr.17: 43013477-43013477 on Build GRCh38
rs564028702 Chr.17: 43013650-43013650 on Build GRCh38
rs148110949 Chr.17: 43013619-43013619 on Build GRCh38
rs759535040 Chr.17: 43013483-43013483 on Build GRCh38
rs145567403 Chr.17: 43013570-43013570 on Build GRCh38
rs749891046 Chr.17: 43013535-43013535 on Build GRCh38
rs202144105 Chr.17: 43013593-43013593 on Build GRCh38
rs149709197 Chr.17: 43013497-43013497 on Build GRCh38
rs766627219 Chr.17: 43013308-43013308 on Build GRCh38
rs1002597752 Chr.17: 43013494-43013494 on Build GRCh38
rs762089420 Chr.17: 43013359-43013359 on Build GRCh38
rs750326884 Chr.17: 43013599-43013599 on Build GRCh38
rs746241316 Chr.17: 43013335-43013348 on Build GRCh38
rs745402288 Chr.17: 43013434-43013434 on Build GRCh38
rs374815335 Chr.17: 43013300-43013300 on Build GRCh38
rs909534000 Chr.17: 43013578-43013578 on Build GRCh38
rs752849055 Chr.17: 43013487-43013487 on Build GRCh38
rs759202201 Chr.17: 43013501-43013502 on Build GRCh38
rs765405814 Chr.17: 43013398-43013398 on Build GRCh38
rs774738003 Chr.17: 43013357-43013357 on Build GRCh38

View Full Product Details