Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 43966353-43966895 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PYY
Entrez Gene ID
5697
Gene Name
peptide YY
Gene Aliases
PYY-I, PYY1
UniGene
Hs.169249
Gene Chromosome Location
Chr.17: 43952733-44024260 on Build GRCh38
Transcripts
NM_004160.5 XM_011525035.1

SNP Detail

SNP ID Chromosome Location
rs1049263803 Chr.17: 43966825-43966825 on Build GRCh38
rs574019679 Chr.17: 43966771-43966771 on Build GRCh38
rs763330470 Chr.17: 43966627-43966627 on Build GRCh38
rs764034822 Chr.17: 43966737-43966737 on Build GRCh38
rs114251383 Chr.17: 43966645-43966645 on Build GRCh38
rs906560800 Chr.17: 43966702-43966702 on Build GRCh38
rs1025050468 Chr.17: 43966508-43966508 on Build GRCh38
rs531301309 Chr.17: 43966410-43966410 on Build GRCh38
rs977942609 Chr.17: 43966497-43966497 on Build GRCh38
rs920213380 Chr.17: 43966792-43966792 on Build GRCh38
rs905269667 Chr.17: 43966829-43966829 on Build GRCh38
rs1038789556 Chr.17: 43966575-43966575 on Build GRCh38
rs989323567 Chr.17: 43966385-43966385 on Build GRCh38
rs780673131 Chr.17: 43966601-43966601 on Build GRCh38
rs769695717 Chr.17: 43966647-43966647 on Build GRCh38
rs144925858 Chr.17: 43966794-43966794 on Build GRCh38
rs940156991 Chr.17: 43966873-43966873 on Build GRCh38
rs146487436 Chr.17: 43966606-43966606 on Build GRCh38
rs969899530 Chr.17: 43966512-43966512 on Build GRCh38
rs193083320 Chr.17: 43966796-43966796 on Build GRCh38
rs1011372011 Chr.17: 43966750-43966750 on Build GRCh38
rs911799198 Chr.17: 43966791-43966791 on Build GRCh38
rs150418725 Chr.17: 43966597-43966597 on Build GRCh38
rs988314103 Chr.17: 43966605-43966605 on Build GRCh38
rs944349763 Chr.17: 43966552-43966552 on Build GRCh38
rs915175461 Chr.17: 43966408-43966408 on Build GRCh38
rs183343231 Chr.17: 43966506-43966506 on Build GRCh38
rs1014077424 Chr.17: 43966813-43966813 on Build GRCh38
rs966608663 Chr.17: 43966453-43966453 on Build GRCh38
rs959474649 Chr.17: 43966744-43966744 on Build GRCh38
rs943153993 Chr.17: 43966665-43966665 on Build GRCh38
rs762687837 Chr.17: 43966776-43966776 on Build GRCh38
rs1002667134 Chr.17: 43966860-43966860 on Build GRCh38
rs534155978 Chr.17: 43966650-43966650 on Build GRCh38
rs555474353 Chr.17: 43966692-43966692 on Build GRCh38
rs895764162 Chr.17: 43966806-43966806 on Build GRCh38
rs189196638 Chr.17: 43966560-43966560 on Build GRCh38
rs977993506 Chr.17: 43966785-43966785 on Build GRCh38
rs377747781 Chr.17: 43966658-43966658 on Build GRCh38
rs770403065 Chr.17: 43966389-43966389 on Build GRCh38
rs1003525579 Chr.17: 43966723-43966723 on Build GRCh38
rs762092449 Chr.17: 43966861-43966861 on Build GRCh38
rs376191287 Chr.17: 43966437-43966437 on Build GRCh38
rs141007828 Chr.17: 43966710-43966710 on Build GRCh38
rs58895551 Chr.17: 43966816-43966816 on Build GRCh38

View Full Product Details