Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 44376171-44376586 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ITGA2B
Entrez Gene ID
3674
Gene Name
integrin subunit alpha 2b
Gene Aliases
BDPLT16, BDPLT2, CD41, CD41B, GP2B, GPIIb, GT, GTA, HPA3, PPP1R93
UniGene
Hs.411312
Gene Chromosome Location
Chr.17: 44372181-44389601 on Build GRCh38
Transcripts
NM_000419.4 XM_011524749.1 XM_011524750.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3191077 c.2326G>C p.E776Q Chr.17: 44376330-44376330 on Build GRCh38
359101 c.2345G>C p.R782P Chr.17: 44376311-44376311 on Build GRCh38
979949 c.2315C>A p.P772Q Chr.17: 44376341-44376341 on Build GRCh38
4946489 c.2311G>T p.V771L Chr.17: 44376345-44376345 on Build GRCh38
3191078 c.2318T>A p.V773D Chr.17: 44376338-44376338 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs368492216 Chr.17: 44376271-44376271 on Build GRCh38
rs1050232582 Chr.17: 44376503-44376503 on Build GRCh38
rs368953599 Chr.17: 44376345-44376345 on Build GRCh38
rs764819935 Chr.17: 44376320-44376320 on Build GRCh38
rs760530417 Chr.17: 44376359-44376359 on Build GRCh38
rs750733496 Chr.17: 44376417-44376417 on Build GRCh38
rs1027937329 Chr.17: 44376415-44376415 on Build GRCh38
rs117870452 Chr.17: 44376322-44376322 on Build GRCh38
rs961430928 Chr.17: 44376542-44376542 on Build GRCh38
rs754656542 Chr.17: 44376318-44376318 on Build GRCh38
rs372100833 Chr.17: 44376274-44376274 on Build GRCh38
rs764863569 Chr.17: 44376300-44376301 on Build GRCh38
rs972246341 Chr.17: 44376549-44376549 on Build GRCh38
rs759094178 Chr.17: 44376379-44376379 on Build GRCh38
rs772998090 Chr.17: 44376353-44376353 on Build GRCh38
rs760644213 Chr.17: 44376305-44376305 on Build GRCh38
rs1005967499 Chr.17: 44376537-44376537 on Build GRCh38
rs757804234 Chr.17: 44376211-44376211 on Build GRCh38
rs369736608 Chr.17: 44376431-44376431 on Build GRCh38
rs765883011 Chr.17: 44376311-44376311 on Build GRCh38
rs748537647 Chr.17: 44376367-44376368 on Build GRCh38
rs778115155 Chr.17: 44376202-44376202 on Build GRCh38
rs74475415 Chr.17: 44376323-44376323 on Build GRCh38
rs754377689 Chr.17: 44376424-44376424 on Build GRCh38
rs564193897 Chr.17: 44376250-44376250 on Build GRCh38
rs751970484 Chr.17: 44376301-44376302 on Build GRCh38
rs564982122 Chr.17: 44376543-44376543 on Build GRCh38
rs755514607 Chr.17: 44376308-44376338 on Build GRCh38
rs560139155 Chr.17: 44376217-44376217 on Build GRCh38
rs199682503 Chr.17: 44376195-44376195 on Build GRCh38
rs765174345 Chr.17: 44376399-44376399 on Build GRCh38
rs963535818 Chr.17: 44376249-44376249 on Build GRCh38
rs776140064 Chr.17: 44376367-44376367 on Build GRCh38
rs755151321 Chr.17: 44376429-44376429 on Build GRCh38
rs996430198 Chr.17: 44376475-44376478 on Build GRCh38
rs776442328 Chr.17: 44376303-44376303 on Build GRCh38
rs1043739041 Chr.17: 44376346-44376346 on Build GRCh38
rs764694431 Chr.17: 44376381-44376381 on Build GRCh38
rs754562766 Chr.17: 44376196-44376196 on Build GRCh38
rs74563314 Chr.17: 44376388-44376388 on Build GRCh38
rs1028759287 Chr.17: 44376251-44376251 on Build GRCh38
rs547581737 Chr.17: 44376336-44376336 on Build GRCh38
rs146892478 Chr.17: 44376335-44376335 on Build GRCh38
rs781357804 Chr.17: 44376366-44376366 on Build GRCh38
rs368755996 Chr.17: 44376340-44376340 on Build GRCh38
rs753582197 Chr.17: 44376206-44376207 on Build GRCh38
rs1014586407 Chr.17: 44376309-44376309 on Build GRCh38
rs377319442 Chr.17: 44376405-44376405 on Build GRCh38
rs752360716 Chr.17: 44376394-44376394 on Build GRCh38
rs369924762 Chr.17: 44376356-44376356 on Build GRCh38
rs531356248 Chr.17: 44376341-44376341 on Build GRCh38
rs1002511113 Chr.17: 44376554-44376554 on Build GRCh38
rs189881723 Chr.17: 44376432-44376432 on Build GRCh38
rs773087433 Chr.17: 44376294-44376294 on Build GRCh38
rs201613707 Chr.17: 44376213-44376213 on Build GRCh38
rs375220061 Chr.17: 44376270-44376270 on Build GRCh38
rs1016658438 Chr.17: 44376534-44376534 on Build GRCh38
rs200571001 Chr.17: 44376293-44376293 on Build GRCh38
rs770815377 Chr.17: 44376363-44376363 on Build GRCh38
rs542591743 Chr.17: 44376315-44376315 on Build GRCh38
rs781635466 Chr.17: 44376393-44376394 on Build GRCh38
rs1005176662 Chr.17: 44376481-44376481 on Build GRCh38
rs756723560 Chr.17: 44376419-44376419 on Build GRCh38
rs780661624 Chr.17: 44376420-44376420 on Build GRCh38
rs762654815 Chr.17: 44376400-44376400 on Build GRCh38
rs938961548 Chr.17: 44376295-44376295 on Build GRCh38
rs761470035 Chr.17: 44376279-44376279 on Build GRCh38
rs372003753 Chr.17: 44376236-44376236 on Build GRCh38
rs756806627 Chr.17: 44376297-44376298 on Build GRCh38

View Full Product Details