Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 32669481-32669994 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RBBP4
Entrez Gene ID
5928
Gene Name
RB binding protein 4, chromatin remodeling factor
Gene Aliases
NURF55, RBAP48, lin-53
UniGene
Hs.16003
Gene Chromosome Location
Chr.1: 32651148-32686211 on Build GRCh38
Transcripts
NM_001135255.1 NM_001135256.1 NM_005610.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7179840 c.918G>A p.L306L Chr.1: 32669515-32669515 on Build GRCh38
6305050 c.955G>T p.E319* Chr.1: 32669552-32669552 on Build GRCh38
1687344 c.935C>T p.S312F Chr.1: 32669532-32669532 on Build GRCh38
5538053 c.897C>T p.A299A Chr.1: 32669494-32669494 on Build GRCh38
3488184 c.936C>T p.S312S Chr.1: 32669533-32669533 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs192849185 Chr.1: 32669944-32669944 on Build GRCh38
rs539292156 Chr.1: 32669665-32669665 on Build GRCh38
rs760606458 Chr.1: 32669758-32669758 on Build GRCh38
rs919872443 Chr.1: 32669699-32669699 on Build GRCh38
rs911765772 Chr.1: 32669631-32669631 on Build GRCh38
rs359957 Chr.1: 32669922-32669922 on Build GRCh38
rs11459018 Chr.1: 32669899-32669900 on Build GRCh38
rs570402567 Chr.1: 32669747-32669747 on Build GRCh38
rs375953366 Chr.1: 32669593-32669593 on Build GRCh38
rs745583595 Chr.1: 32669671-32669671 on Build GRCh38
rs541022949 Chr.1: 32669661-32669661 on Build GRCh38
rs1023081799 Chr.1: 32669755-32669755 on Build GRCh38
rs775351873 Chr.1: 32669746-32669746 on Build GRCh38
rs6696422 Chr.1: 32669907-32669907 on Build GRCh38
rs1690907 Chr.1: 32669649-32669649 on Build GRCh38
rs935553859 Chr.1: 32669832-32669832 on Build GRCh38
rs969716845 Chr.1: 32669764-32669764 on Build GRCh38
rs192534491 Chr.1: 32669601-32669601 on Build GRCh38
rs138881478 Chr.1: 32669891-32669891 on Build GRCh38
rs1044612180 Chr.1: 32669935-32669935 on Build GRCh38
rs1052626399 Chr.1: 32669840-32669840 on Build GRCh38
rs368283019 Chr.1: 32669676-32669676 on Build GRCh38
rs552085893 Chr.1: 32669727-32669727 on Build GRCh38
rs751460935 Chr.1: 32669586-32669586 on Build GRCh38
rs895977605 Chr.1: 32669913-32669913 on Build GRCh38
rs766045193 Chr.1: 32669660-32669660 on Build GRCh38
rs764823257 Chr.1: 32669533-32669533 on Build GRCh38
rs973949216 Chr.1: 32669506-32669506 on Build GRCh38
rs894105945 Chr.1: 32669616-32669616 on Build GRCh38
rs1032737347 Chr.1: 32669776-32669776 on Build GRCh38
rs974656321 Chr.1: 32669814-32669814 on Build GRCh38
rs201984533 Chr.1: 32669608-32669608 on Build GRCh38
rs185412911 Chr.1: 32669734-32669734 on Build GRCh38
rs372273368 Chr.1: 32669583-32669583 on Build GRCh38
rs974063648 Chr.1: 32669686-32669686 on Build GRCh38
rs1748052 Chr.1: 32669787-32669787 on Build GRCh38
rs1037645389 Chr.1: 32669893-32669900 on Build GRCh38
rs530594725 Chr.1: 32669673-32669673 on Build GRCh38
rs1043987839 Chr.1: 32669620-32669620 on Build GRCh38
rs372380199 Chr.1: 32669596-32669596 on Build GRCh38
rs113576220 Chr.1: 32669662-32669662 on Build GRCh38
rs904099219 Chr.1: 32669626-32669626 on Build GRCh38
rs776094865 Chr.1: 32669783-32669783 on Build GRCh38
rs368502528 Chr.1: 32669759-32669759 on Build GRCh38
rs761186145 Chr.1: 32669524-32669524 on Build GRCh38
rs563357085 Chr.1: 32669666-32669666 on Build GRCh38
rs149121780 Chr.1: 32669513-32669513 on Build GRCh38
rs764753297 Chr.1: 32669936-32669936 on Build GRCh38
rs535594034 Chr.1: 32669858-32669858 on Build GRCh38
rs932632564 Chr.1: 32669868-32669868 on Build GRCh38
rs1018406052 Chr.1: 32669878-32669878 on Build GRCh38
rs557399383 Chr.1: 32669886-32669886 on Build GRCh38
rs576948699 Chr.1: 32669617-32669617 on Build GRCh38
rs6670385 Chr.1: 32669774-32669774 on Build GRCh38
rs773474350 Chr.1: 32669839-32669839 on Build GRCh38
rs772061137 Chr.1: 32669716-32669716 on Build GRCh38
rs866246795 Chr.1: 32669664-32669664 on Build GRCh38
rs68153136 Chr.1: 32669899-32669899 on Build GRCh38
rs983063593 Chr.1: 32669955-32669955 on Build GRCh38
rs966393860 Chr.1: 32669805-32669805 on Build GRCh38
rs988670419 Chr.1: 32669788-32669788 on Build GRCh38
rs546683371 Chr.1: 32669782-32669782 on Build GRCh38
rs145752475 Chr.1: 32669588-32669588 on Build GRCh38
rs535898657 Chr.1: 32669580-32669580 on Build GRCh38
rs188732043 Chr.1: 32669816-32669816 on Build GRCh38
rs750801115 Chr.1: 32669950-32669950 on Build GRCh38
rs879598039 Chr.1: 32669784-32669784 on Build GRCh38
rs921269156 Chr.1: 32669857-32669857 on Build GRCh38
rs750018570 Chr.1: 32669570-32669570 on Build GRCh38
rs891251906 Chr.1: 32669869-32669869 on Build GRCh38
rs6685469 Chr.1: 32669901-32669901 on Build GRCh38
rs911708811 Chr.1: 32669797-32669797 on Build GRCh38
rs1008395660 Chr.1: 32669877-32669877 on Build GRCh38

View Full Product Details