Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 32679921-32680447 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RBBP4
Entrez Gene ID
5928
Gene Name
RB binding protein 4, chromatin remodeling factor
Gene Aliases
NURF55, RBAP48, lin-53
UniGene
Hs.16003
Gene Chromosome Location
Chr.1: 32651148-32686211 on Build GRCh38
Transcripts
NM_001135255.1 NM_001135256.1 NM_005610.2 NM_001161708.1 NM_030786.2

Gene Symbol
SYNC
Entrez Gene ID
81493
Gene Name
syncoilin, intermediate filament protein
Gene Aliases
SYNC1, SYNCOILIN
UniGene
Hs.712631
Gene Chromosome Location
Chr.1: 32679906-32702760 on Build GRCh38
Transcripts
NM_001135255.1 NM_001135256.1 NM_005610.2 NM_001161708.1 NM_030786.2

SNP Detail

SNP ID Chromosome Location
rs199920140 Chr.1: 32680347-32680348 on Build GRCh38
rs630356 Chr.1: 32680323-32680323 on Build GRCh38
rs992180528 Chr.1: 32680207-32680207 on Build GRCh38
rs1045214191 Chr.1: 32680091-32680091 on Build GRCh38
rs1052546915 Chr.1: 32680309-32680311 on Build GRCh38
rs201862108 Chr.1: 32680170-32680170 on Build GRCh38
rs564985598 Chr.1: 32680281-32680281 on Build GRCh38
rs944252470 Chr.1: 32680340-32680340 on Build GRCh38
rs754598275 Chr.1: 32680124-32680124 on Build GRCh38
rs952608546 Chr.1: 32679995-32679995 on Build GRCh38
rs914190847 Chr.1: 32680326-32680326 on Build GRCh38
rs892503736 Chr.1: 32680228-32680228 on Build GRCh38
rs185032778 Chr.1: 32680129-32680129 on Build GRCh38
rs559084429 Chr.1: 32680363-32680363 on Build GRCh38
rs892703560 Chr.1: 32680137-32680137 on Build GRCh38
rs996610832 Chr.1: 32680361-32680361 on Build GRCh38
rs552550152 Chr.1: 32680157-32680158 on Build GRCh38
rs878935421 Chr.1: 32680376-32680376 on Build GRCh38
rs907888388 Chr.1: 32680003-32680004 on Build GRCh38
rs11490888 Chr.1: 32680347-32680347 on Build GRCh38
rs752601182 Chr.1: 32680350-32680365 on Build GRCh38
rs189481690 Chr.1: 32679956-32679956 on Build GRCh38
rs878971886 Chr.1: 32680338-32680339 on Build GRCh38
rs9851 Chr.1: 32680141-32680141 on Build GRCh38
rs948101422 Chr.1: 32680059-32680059 on Build GRCh38
rs1015558567 Chr.1: 32680184-32680184 on Build GRCh38
rs548056152 Chr.1: 32680365-32680365 on Build GRCh38
rs372285843 Chr.1: 32680358-32680358 on Build GRCh38
rs996960229 Chr.1: 32680157-32680157 on Build GRCh38
rs1004036008 Chr.1: 32680181-32680181 on Build GRCh38
rs173397 Chr.1: 32680314-32680314 on Build GRCh38
rs555426509 Chr.1: 32680133-32680133 on Build GRCh38
rs1051870416 Chr.1: 32680353-32680353 on Build GRCh38
rs960222261 Chr.1: 32680199-32680199 on Build GRCh38
rs529957859 Chr.1: 32680364-32680364 on Build GRCh38
rs144062064 Chr.1: 32680009-32680009 on Build GRCh38
rs917873187 Chr.1: 32680224-32680228 on Build GRCh38
rs982671584 Chr.1: 32679999-32679999 on Build GRCh38
rs890520049 Chr.1: 32680379-32680379 on Build GRCh38
rs888179056 Chr.1: 32680366-32680367 on Build GRCh38
rs372304464 Chr.1: 32680165-32680172 on Build GRCh38
rs975305072 Chr.1: 32679966-32679966 on Build GRCh38
rs532255103 Chr.1: 32680282-32680282 on Build GRCh38
rs570996981 Chr.1: 32679991-32679991 on Build GRCh38
rs1040717572 Chr.1: 32680341-32680341 on Build GRCh38
rs1026783657 Chr.1: 32679982-32679982 on Build GRCh38
rs918043061 Chr.1: 32680035-32680035 on Build GRCh38
rs962097776 Chr.1: 32680005-32680005 on Build GRCh38
rs777880789 Chr.1: 32680246-32680246 on Build GRCh38
rs969519199 Chr.1: 32680238-32680238 on Build GRCh38
rs748968329 Chr.1: 32680183-32680183 on Build GRCh38
rs373788617 Chr.1: 32680350-32680357 on Build GRCh38
rs191064854 Chr.1: 32680144-32680144 on Build GRCh38
rs377069414 Chr.1: 32680001-32680001 on Build GRCh38
rs771217211 Chr.1: 32680298-32680298 on Build GRCh38
rs375233693 Chr.1: 32680390-32680390 on Build GRCh38
rs180959850 Chr.1: 32680098-32680098 on Build GRCh38
rs535204011 Chr.1: 32680104-32680104 on Build GRCh38
rs2786169 Chr.1: 32680350-32680350 on Build GRCh38
rs1015402972 Chr.1: 32680398-32680398 on Build GRCh38
rs952494479 Chr.1: 32680168-32680168 on Build GRCh38
rs899472664 Chr.1: 32680344-32680345 on Build GRCh38
rs925359726 Chr.1: 32680261-32680261 on Build GRCh38
rs145459212 Chr.1: 32680163-32680170 on Build GRCh38
rs879220285 Chr.1: 32680357-32680357 on Build GRCh38
rs181724627 Chr.1: 32680245-32680245 on Build GRCh38

View Full Product Details