Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 32779998-32780493 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
YARS
Entrez Gene ID
8565
Gene Name
tyrosyl-tRNA synthetase
Gene Aliases
CMTDIC, TYRRS, YRS, YTS
UniGene
Hs.213264
Gene Chromosome Location
Chr.1: 32775237-32818032 on Build GRCh38
Transcripts
NM_003680.3 XM_011542347.1 XM_011542348.1 XM_017002651.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7001327 c.1217_1218insCGTAC p.Q408fs*15 Chr.1: 32780201-32780202 on Build GRCh38
3789897 c.1293G>T p.M431I Chr.1: 32780126-32780126 on Build GRCh38
7136551 c.1227C>A p.F409L Chr.1: 32780192-32780192 on Build GRCh38
6126082 c.1180G>T p.V394L Chr.1: 32780239-32780239 on Build GRCh38
4842450 c.1334+7G>A p.? Chr.1: 32780078-32780078 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774750132 Chr.1: 32780176-32780176 on Build GRCh38
rs777580175 Chr.1: 32780310-32780310 on Build GRCh38
rs776731822 Chr.1: 32780038-32780038 on Build GRCh38
rs1021595426 Chr.1: 32780336-32780336 on Build GRCh38
rs1025774447 Chr.1: 32780427-32780427 on Build GRCh38
rs745651845 Chr.1: 32780212-32780212 on Build GRCh38
rs1028633163 Chr.1: 32780062-32780062 on Build GRCh38
rs369477473 Chr.1: 32780315-32780315 on Build GRCh38
rs763168140 Chr.1: 32780070-32780070 on Build GRCh38
rs945849781 Chr.1: 32780269-32780269 on Build GRCh38
rs76611863 Chr.1: 32780173-32780173 on Build GRCh38
rs569727346 Chr.1: 32780197-32780197 on Build GRCh38
rs552964149 Chr.1: 32780337-32780337 on Build GRCh38
rs750110125 Chr.1: 32780309-32780309 on Build GRCh38
rs777567617 Chr.1: 32780116-32780116 on Build GRCh38
rs764927716 Chr.1: 32780308-32780308 on Build GRCh38
rs759870198 Chr.1: 32780188-32780188 on Build GRCh38
rs189171835 Chr.1: 32780195-32780195 on Build GRCh38
rs745677887 Chr.1: 32780148-32780148 on Build GRCh38
rs775335977 Chr.1: 32780156-32780156 on Build GRCh38
rs201616999 Chr.1: 32780294-32780294 on Build GRCh38
rs772261295 Chr.1: 32780270-32780270 on Build GRCh38
rs998241043 Chr.1: 32780353-32780353 on Build GRCh38
rs699005 Chr.1: 32780201-32780201 on Build GRCh38
rs146393022 Chr.1: 32780191-32780191 on Build GRCh38
rs937276722 Chr.1: 32780198-32780198 on Build GRCh38
rs558667909 Chr.1: 32780291-32780291 on Build GRCh38
rs377391137 Chr.1: 32780433-32780433 on Build GRCh38
rs756979929 Chr.1: 32780142-32780142 on Build GRCh38
rs939956712 Chr.1: 32780223-32780223 on Build GRCh38
rs764028295 Chr.1: 32780143-32780143 on Build GRCh38
rs766672765 Chr.1: 32780072-32780072 on Build GRCh38
rs765421671 Chr.1: 32780063-32780063 on Build GRCh38
rs763115155 Chr.1: 32780164-32780164 on Build GRCh38
rs997604940 Chr.1: 32780059-32780059 on Build GRCh38
rs994317299 Chr.1: 32780419-32780419 on Build GRCh38
rs368899441 Chr.1: 32780314-32780314 on Build GRCh38
rs962846822 Chr.1: 32780126-32780126 on Build GRCh38
rs76862302 Chr.1: 32780086-32780086 on Build GRCh38
rs770629329 Chr.1: 32780134-32780134 on Build GRCh38
rs754110699 Chr.1: 32780068-32780068 on Build GRCh38
rs995106454 Chr.1: 32780119-32780119 on Build GRCh38
rs914432062 Chr.1: 32780217-32780217 on Build GRCh38
rs761211081 Chr.1: 32780287-32780287 on Build GRCh38
rs375192497 Chr.1: 32780215-32780215 on Build GRCh38
rs772014071 Chr.1: 32780153-32780153 on Build GRCh38
rs749411607 Chr.1: 32780244-32780244 on Build GRCh38
rs563131536 Chr.1: 32780455-32780455 on Build GRCh38
rs754879604 Chr.1: 32780317-32780317 on Build GRCh38
rs971485324 Chr.1: 32780157-32780157 on Build GRCh38
rs1017121501 Chr.1: 32780415-32780415 on Build GRCh38
rs763636325 Chr.1: 32780192-32780192 on Build GRCh38
rs755747938 Chr.1: 32780108-32780108 on Build GRCh38
rs751461018 Chr.1: 32780439-32780439 on Build GRCh38
rs778718842 Chr.1: 32780144-32780144 on Build GRCh38
rs759944936 Chr.1: 32780260-32780260 on Build GRCh38
rs149620809 Chr.1: 32780128-32780128 on Build GRCh38
rs373909518 Chr.1: 32780240-32780240 on Build GRCh38
rs768603403 Chr.1: 32780222-32780222 on Build GRCh38
rs863224717 Chr.1: 32780239-32780239 on Build GRCh38
rs767952376 Chr.1: 32780099-32780099 on Build GRCh38
rs746892773 Chr.1: 32780220-32780220 on Build GRCh38
rs773479387 Chr.1: 32780069-32780069 on Build GRCh38
rs949403106 Chr.1: 32780285-32780285 on Build GRCh38
rs181563125 Chr.1: 32780418-32780418 on Build GRCh38
rs781159171 Chr.1: 32780325-32780325 on Build GRCh38
rs761920995 Chr.1: 32780056-32780056 on Build GRCh38
rs139745541 Chr.1: 32780276-32780276 on Build GRCh38
rs752232252 Chr.1: 32780103-32780103 on Build GRCh38
rs1028111847 Chr.1: 32780411-32780411 on Build GRCh38
rs1016796818 Chr.1: 32780448-32780448 on Build GRCh38
rs576746700 Chr.1: 32780293-32780293 on Build GRCh38
rs758073997 Chr.1: 32780313-32780313 on Build GRCh38
rs778469199 Chr.1: 32780207-32780207 on Build GRCh38
rs574395455 Chr.1: 32780352-32780352 on Build GRCh38
rs774497213 Chr.1: 32780254-32780254 on Build GRCh38
rs1009840070 Chr.1: 32780319-32780319 on Build GRCh38
rs751581247 Chr.1: 32780303-32780304 on Build GRCh38
rs751894456 Chr.1: 32780080-32780080 on Build GRCh38
rs141675786 Chr.1: 32780162-32780162 on Build GRCh38
rs759283424 Chr.1: 32780025-32780025 on Build GRCh38

View Full Product Details