Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 32895123-32895623 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM54
Entrez Gene ID
113452
Gene Name
transmembrane protein 54
Gene Aliases
BCLP, CAC-1, CAC1
UniGene
Hs.534521
Gene Chromosome Location
Chr.1: 32894595-32901435 on Build GRCh38
Transcripts
NM_033504.2 XM_005270408.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6263167 c.468C>T p.S156S Chr.1: 32895431-32895431 on Build GRCh38
4127665 c.501C>T p.C167C Chr.1: 32895398-32895398 on Build GRCh38
6502421 c.408T>G p.S136S Chr.1: 32895606-32895606 on Build GRCh38
1341869 c.452G>A p.R151H Chr.1: 32895562-32895562 on Build GRCh38
3785262 c.526C>T p.R176C Chr.1: 32895373-32895373 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs371265580 Chr.1: 32895405-32895405 on Build GRCh38
rs747413698 Chr.1: 32895258-32895258 on Build GRCh38
rs755559295 Chr.1: 32895300-32895300 on Build GRCh38
rs765563498 Chr.1: 32895330-32895330 on Build GRCh38
rs764908914 Chr.1: 32895517-32895517 on Build GRCh38
rs917751210 Chr.1: 32895461-32895461 on Build GRCh38
rs371955837 Chr.1: 32895444-32895444 on Build GRCh38
rs143677132 Chr.1: 32895398-32895398 on Build GRCh38
rs372718654 Chr.1: 32895506-32895506 on Build GRCh38
rs375870376 Chr.1: 32895526-32895526 on Build GRCh38
rs769942306 Chr.1: 32895453-32895453 on Build GRCh38
rs971885507 Chr.1: 32895237-32895237 on Build GRCh38
rs149167713 Chr.1: 32895286-32895286 on Build GRCh38
rs572588755 Chr.1: 32895434-32895434 on Build GRCh38
rs747905795 Chr.1: 32895276-32895276 on Build GRCh38
rs200142858 Chr.1: 32895397-32895397 on Build GRCh38
rs750406047 Chr.1: 32895522-32895522 on Build GRCh38
rs773746993 Chr.1: 32895333-32895333 on Build GRCh38
rs754546282 Chr.1: 32895219-32895219 on Build GRCh38
rs374131329 Chr.1: 32895358-32895358 on Build GRCh38
rs1041966653 Chr.1: 32895599-32895599 on Build GRCh38
rs200781061 Chr.1: 32895392-32895392 on Build GRCh38
rs1028668292 Chr.1: 32895242-32895242 on Build GRCh38
rs771183649 Chr.1: 32895471-32895471 on Build GRCh38
rs781524571 Chr.1: 32895386-32895386 on Build GRCh38
rs148310921 Chr.1: 32895572-32895572 on Build GRCh38
rs925215364 Chr.1: 32895519-32895519 on Build GRCh38
rs375848678 Chr.1: 32895289-32895289 on Build GRCh38
rs376607828 Chr.1: 32895408-32895408 on Build GRCh38
rs759925306 Chr.1: 32895484-32895484 on Build GRCh38
rs936673721 Chr.1: 32895528-32895528 on Build GRCh38
rs1052878528 Chr.1: 32895549-32895549 on Build GRCh38
rs747846335 Chr.1: 32895372-32895372 on Build GRCh38
rs376198320 Chr.1: 32895305-32895305 on Build GRCh38
rs143264287 Chr.1: 32895562-32895562 on Build GRCh38
rs201182293 Chr.1: 32895404-32895404 on Build GRCh38
rs757675550 Chr.1: 32895567-32895567 on Build GRCh38
rs377069821 Chr.1: 32895259-32895259 on Build GRCh38
rs757951992 Chr.1: 32895256-32895256 on Build GRCh38
rs769092217 Chr.1: 32895413-32895433 on Build GRCh38
rs373374840 Chr.1: 32895264-32895264 on Build GRCh38
rs1019866890 Chr.1: 32895154-32895154 on Build GRCh38
rs752272963 Chr.1: 32895367-32895367 on Build GRCh38
rs991928660 Chr.1: 32895451-32895451 on Build GRCh38
rs144983176 Chr.1: 32895262-32895262 on Build GRCh38
rs539277670 Chr.1: 32895576-32895576 on Build GRCh38
rs993430590 Chr.1: 32895438-32895438 on Build GRCh38
rs780609869 Chr.1: 32895334-32895334 on Build GRCh38
rs190204816 Chr.1: 32895223-32895223 on Build GRCh38
rs892633449 Chr.1: 32895144-32895144 on Build GRCh38
rs368310171 Chr.1: 32895373-32895373 on Build GRCh38
rs760054008 Chr.1: 32895563-32895563 on Build GRCh38
rs761750680 Chr.1: 32895516-32895516 on Build GRCh38
rs763908661 Chr.1: 32895513-32895513 on Build GRCh38
rs570652899 Chr.1: 32895230-32895230 on Build GRCh38
rs753528659 Chr.1: 32895515-32895515 on Build GRCh38
rs747181002 Chr.1: 32895377-32895377 on Build GRCh38
rs768857203 Chr.1: 32895326-32895326 on Build GRCh38
rs1052512256 Chr.1: 32895378-32895378 on Build GRCh38
rs75611037 Chr.1: 32895275-32895275 on Build GRCh38
rs762393156 Chr.1: 32895589-32895589 on Build GRCh38
rs146734508 Chr.1: 32895412-32895412 on Build GRCh38
rs771637960 Chr.1: 32895485-32895485 on Build GRCh38
rs969242656 Chr.1: 32895476-32895476 on Build GRCh38
rs778826833 Chr.1: 32895368-32895368 on Build GRCh38
rs963249771 Chr.1: 32895437-32895437 on Build GRCh38
rs776950119 Chr.1: 32895414-32895415 on Build GRCh38
rs774966593 Chr.1: 32895479-32895479 on Build GRCh38
rs60869063 Chr.1: 32895143-32895143 on Build GRCh38
rs947990884 Chr.1: 32895342-32895342 on Build GRCh38
rs748391059 Chr.1: 32895450-32895450 on Build GRCh38
rs753622774 Chr.1: 32895407-32895407 on Build GRCh38
rs142745217 Chr.1: 32895327-32895327 on Build GRCh38
rs778113030 Chr.1: 32895454-32895454 on Build GRCh38
rs372910271 Chr.1: 32895399-32895399 on Build GRCh38
rs928732336 Chr.1: 32895537-32895537 on Build GRCh38
rs368906706 Chr.1: 32895403-32895403 on Build GRCh38
rs560592488 Chr.1: 32895366-32895366 on Build GRCh38
rs749210213 Chr.1: 32895288-32895288 on Build GRCh38
rs947616637 Chr.1: 32895591-32895591 on Build GRCh38
rs539821033 Chr.1: 32895221-32895221 on Build GRCh38
rs573870806 Chr.1: 32895235-32895235 on Build GRCh38
rs755965238 Chr.1: 32895281-32895281 on Build GRCh38
rs1008836634 Chr.1: 32895147-32895147 on Build GRCh38
rs752704880 Chr.1: 32895543-32895543 on Build GRCh38
rs749716012 Chr.1: 32895463-32895463 on Build GRCh38
rs980653857 Chr.1: 32895512-32895512 on Build GRCh38
rs149039000 Chr.1: 32895424-32895424 on Build GRCh38
rs368627916 Chr.1: 32895490-32895490 on Build GRCh38
rs781344548 Chr.1: 32895540-32895540 on Build GRCh38
rs555338609 Chr.1: 32895561-32895561 on Build GRCh38
rs775066105 Chr.1: 32895432-32895432 on Build GRCh38
rs751021383 Chr.1: 32895359-32895359 on Build GRCh38
rs768606278 Chr.1: 32895385-32895385 on Build GRCh38
rs763460747 Chr.1: 32895393-32895393 on Build GRCh38
rs150379179 Chr.1: 32895413-32895413 on Build GRCh38
rs953087772 Chr.1: 32895188-32895188 on Build GRCh38
rs755125275 Chr.1: 32895443-32895443 on Build GRCh38
rs201782944 Chr.1: 32895376-32895376 on Build GRCh38
rs370419083 Chr.1: 32895350-32895350 on Build GRCh38
rs772408166 Chr.1: 32895590-32895590 on Build GRCh38

View Full Product Details