Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 35560951-35561446 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NCDN
Entrez Gene ID
23154
Gene Name
neurochondrin
Gene Aliases
-
UniGene
Hs.121870
Gene Chromosome Location
Chr.1: 35557792-35566779 on Build GRCh38
Transcripts
NM_001014839.1 NM_001014841.1 NM_014284.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2080560 c.800G>T p.R267L Chr.1: 35560951-35560951 on Build GRCh38
3689637 c.827G>T p.G276V Chr.1: 35561029-35561029 on Build GRCh38
186451 c.895G>A p.V299M Chr.1: 35561097-35561097 on Build GRCh38
1146313 c.1024G>C p.E342Q Chr.1: 35561175-35561175 on Build GRCh38
4007370 c.981G>A p.T327T Chr.1: 35561132-35561132 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs28655939 Chr.1: 35561057-35561057 on Build GRCh38
rs756238661 Chr.1: 35561321-35561321 on Build GRCh38
rs1040985329 Chr.1: 35561141-35561141 on Build GRCh38
rs572728420 Chr.1: 35561125-35561125 on Build GRCh38
rs368464438 Chr.1: 35561032-35561032 on Build GRCh38
rs763194610 Chr.1: 35561078-35561078 on Build GRCh38
rs765991260 Chr.1: 35561099-35561099 on Build GRCh38
rs758521938 Chr.1: 35561045-35561045 on Build GRCh38
rs780475408 Chr.1: 35561047-35561047 on Build GRCh38
rs755247482 Chr.1: 35561269-35561269 on Build GRCh38
rs760286139 Chr.1: 35561223-35561223 on Build GRCh38
rs767016268 Chr.1: 35561111-35561111 on Build GRCh38
rs750301464 Chr.1: 35561265-35561265 on Build GRCh38
rs774014076 Chr.1: 35561107-35561107 on Build GRCh38
rs957700101 Chr.1: 35561389-35561389 on Build GRCh38
rs778101207 Chr.1: 35561331-35561331 on Build GRCh38
rs894968754 Chr.1: 35561419-35561419 on Build GRCh38
rs747039344 Chr.1: 35561048-35561048 on Build GRCh38
rs976517081 Chr.1: 35561406-35561406 on Build GRCh38
rs765139402 Chr.1: 35561261-35561261 on Build GRCh38
rs757143203 Chr.1: 35561131-35561131 on Build GRCh38
rs768217577 Chr.1: 35561345-35561345 on Build GRCh38
rs767390347 Chr.1: 35561027-35561027 on Build GRCh38
rs372758379 Chr.1: 35561033-35561033 on Build GRCh38
rs184232102 Chr.1: 35561226-35561226 on Build GRCh38
rs375174876 Chr.1: 35561003-35561003 on Build GRCh38
rs765525449 Chr.1: 35560973-35560973 on Build GRCh38
rs750035948 Chr.1: 35560983-35560983 on Build GRCh38
rs539762693 Chr.1: 35561424-35561424 on Build GRCh38
rs200105021 Chr.1: 35561271-35561271 on Build GRCh38
rs369068328 Chr.1: 35561156-35561156 on Build GRCh38
rs147290531 Chr.1: 35561178-35561178 on Build GRCh38
rs1047466152 Chr.1: 35561367-35561367 on Build GRCh38
rs965528675 Chr.1: 35561401-35561401 on Build GRCh38
rs774834667 Chr.1: 35561214-35561214 on Build GRCh38
rs11545628 Chr.1: 35561113-35561113 on Build GRCh38
rs943795393 Chr.1: 35561135-35561135 on Build GRCh38
rs191794024 Chr.1: 35561144-35561144 on Build GRCh38
rs781409276 Chr.1: 35561151-35561151 on Build GRCh38
rs753219774 Chr.1: 35561314-35561314 on Build GRCh38
rs536102934 Chr.1: 35561211-35561211 on Build GRCh38
rs779231727 Chr.1: 35561332-35561332 on Build GRCh38
rs1002140888 Chr.1: 35561363-35561363 on Build GRCh38
rs150364361 Chr.1: 35561148-35561148 on Build GRCh38
rs747241342 Chr.1: 35561146-35561146 on Build GRCh38
rs771692573 Chr.1: 35561336-35561336 on Build GRCh38
rs766087108 Chr.1: 35561011-35561011 on Build GRCh38
rs773924666 Chr.1: 35561195-35561195 on Build GRCh38
rs1013869598 Chr.1: 35560984-35560984 on Build GRCh38
rs759004083 Chr.1: 35561109-35561109 on Build GRCh38
rs550971291 Chr.1: 35561375-35561375 on Build GRCh38
rs141623030 Chr.1: 35561093-35561093 on Build GRCh38
rs769855847 Chr.1: 35561028-35561028 on Build GRCh38
rs138111798 Chr.1: 35561397-35561397 on Build GRCh38
rs752697777 Chr.1: 35561301-35561301 on Build GRCh38
rs752618742 Chr.1: 35561114-35561114 on Build GRCh38
rs751036326 Chr.1: 35561021-35561021 on Build GRCh38
rs969318014 Chr.1: 35561022-35561022 on Build GRCh38
rs763414802 Chr.1: 35561096-35561096 on Build GRCh38
rs751953513 Chr.1: 35561338-35561338 on Build GRCh38
rs754530076 Chr.1: 35561025-35561025 on Build GRCh38
rs760066803 Chr.1: 35561342-35561342 on Build GRCh38
rs987912428 Chr.1: 35561112-35561112 on Build GRCh38
rs969180485 Chr.1: 35561359-35561359 on Build GRCh38
rs753848215 Chr.1: 35561249-35561249 on Build GRCh38
rs201718421 Chr.1: 35561164-35561164 on Build GRCh38
rs770391339 Chr.1: 35561072-35561072 on Build GRCh38
rs988246388 Chr.1: 35561395-35561395 on Build GRCh38
rs369452951 Chr.1: 35561034-35561034 on Build GRCh38
rs1016122506 Chr.1: 35561398-35561398 on Build GRCh38
rs373303990 Chr.1: 35561243-35561243 on Build GRCh38
rs140348877 Chr.1: 35561126-35561126 on Build GRCh38
rs773171405 Chr.1: 35561044-35561044 on Build GRCh38
rs777214106 Chr.1: 35561055-35561055 on Build GRCh38
rs773816119 Chr.1: 35561157-35561157 on Build GRCh38
rs759196253 Chr.1: 35561199-35561199 on Build GRCh38
rs761636138 Chr.1: 35561250-35561250 on Build GRCh38

View Full Product Details