Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 59388235-59388737 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
YPEL2
Entrez Gene ID
388403
Gene Name
yippee like 2
Gene Aliases
FKSG4
UniGene
Hs.463613
Gene Chromosome Location
Chr.17: 59331692-59401734 on Build GRCh38
Transcripts
NM_001005404.3 XM_017024621.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6147754 c.153T>G p.F51L Chr.17: 59388362-59388362 on Build GRCh38
4741883 c.151_152delTT p.F51fs*1 Chr.17: 59388360-59388361 on Build GRCh38
5790771 c.161+5G>A p.? Chr.17: 59388375-59388375 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs567910317 Chr.17: 59388298-59388298 on Build GRCh38
rs1037490610 Chr.17: 59388433-59388433 on Build GRCh38
rs747147775 Chr.17: 59388281-59388281 on Build GRCh38
rs1023448486 Chr.17: 59388643-59388643 on Build GRCh38
rs890066699 Chr.17: 59388534-59388534 on Build GRCh38
rs34584449 Chr.17: 59388291-59388292 on Build GRCh38
rs1026153408 Chr.17: 59388555-59388555 on Build GRCh38
rs932977951 Chr.17: 59388715-59388715 on Build GRCh38
rs982017635 Chr.17: 59388657-59388657 on Build GRCh38
rs918766287 Chr.17: 59388483-59388484 on Build GRCh38
rs1021434115 Chr.17: 59388432-59388432 on Build GRCh38
rs901496507 Chr.17: 59388306-59388306 on Build GRCh38
rs763544105 Chr.17: 59388319-59388319 on Build GRCh38
rs568742823 Chr.17: 59388260-59388260 on Build GRCh38
rs557464512 Chr.17: 59388540-59388540 on Build GRCh38
rs1031142315 Chr.17: 59388619-59388619 on Build GRCh38
rs767337667 Chr.17: 59388305-59388305 on Build GRCh38
rs115228294 Chr.17: 59388282-59388282 on Build GRCh38
rs780299243 Chr.17: 59388407-59388407 on Build GRCh38
rs763341903 Chr.17: 59388302-59388302 on Build GRCh38
rs900810021 Chr.17: 59388566-59388566 on Build GRCh38
rs112521166 Chr.17: 59388414-59388414 on Build GRCh38
rs2286485 Chr.17: 59388543-59388543 on Build GRCh38
rs540156297 Chr.17: 59388693-59388693 on Build GRCh38
rs1011183805 Chr.17: 59388641-59388641 on Build GRCh38
rs369760331 Chr.17: 59388356-59388356 on Build GRCh38
rs776367477 Chr.17: 59388688-59388688 on Build GRCh38
rs747197186 Chr.17: 59388415-59388415 on Build GRCh38
rs976957901 Chr.17: 59388677-59388677 on Build GRCh38
rs757158658 Chr.17: 59388353-59388353 on Build GRCh38
rs553571529 Chr.17: 59388475-59388475 on Build GRCh38
rs752376737 Chr.17: 59388307-59388307 on Build GRCh38
rs746782752 Chr.17: 59388651-59388651 on Build GRCh38
rs200331135 Chr.17: 59388385-59388385 on Build GRCh38
rs768882009 Chr.17: 59388420-59388420 on Build GRCh38
rs765725922 Chr.17: 59388292-59388292 on Build GRCh38
rs972883484 Chr.17: 59388554-59388554 on Build GRCh38
rs753342559 Chr.17: 59388349-59388349 on Build GRCh38
rs942976855 Chr.17: 59388649-59388649 on Build GRCh38
rs143628314 Chr.17: 59388605-59388605 on Build GRCh38
rs545153514 Chr.17: 59388526-59388526 on Build GRCh38
rs759901387 Chr.17: 59388278-59388290 on Build GRCh38
rs559352667 Chr.17: 59388679-59388679 on Build GRCh38
rs905797593 Chr.17: 59388508-59388508 on Build GRCh38
rs776595912 Chr.17: 59388422-59388422 on Build GRCh38
rs779863047 Chr.17: 59388638-59388638 on Build GRCh38
rs1053272304 Chr.17: 59388357-59388357 on Build GRCh38
rs578231087 Chr.17: 59388497-59388497 on Build GRCh38
rs762257307 Chr.17: 59388291-59388291 on Build GRCh38
rs536281932 Chr.17: 59388265-59388265 on Build GRCh38
rs375491232 Chr.17: 59388313-59388313 on Build GRCh38
rs541750106 Chr.17: 59388637-59388637 on Build GRCh38
rs745732547 Chr.17: 59388378-59388378 on Build GRCh38

View Full Product Details