Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 59838203-59838703 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
VMP1
Entrez Gene ID
81671
Gene Name
vacuole membrane protein 1
Gene Aliases
EPG3, TANGO5, TMEM49
UniGene
Hs.444569
Gene Chromosome Location
Chr.17: 59707502-59840593 on Build GRCh38
Transcripts

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3927581 c.1009C>T p.P337S Chr.17: 59838329-59838329 on Build GRCh38
1750205 c.1015C>T p.Q339* Chr.17: 59838335-59838335 on Build GRCh38
1235793 c.1045C>A p.L349I Chr.17: 59838365-59838365 on Build GRCh38
1744724 c.1020G>A p.E340E Chr.17: 59838340-59838340 on Build GRCh38
5342983 c.984C>A p.P328P Chr.17: 59838304-59838304 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs376323785 Chr.17: 59838645-59838645 on Build GRCh38
rs764315689 Chr.17: 59838322-59838322 on Build GRCh38
rs917755153 Chr.17: 59838642-59838642 on Build GRCh38
rs969680652 Chr.17: 59838573-59838573 on Build GRCh38
rs763467142 Chr.17: 59838379-59838379 on Build GRCh38
rs914235505 Chr.17: 59838244-59838244 on Build GRCh38
rs973005754 Chr.17: 59838622-59838622 on Build GRCh38
rs768820638 Chr.17: 59838350-59838350 on Build GRCh38
rs185222973 Chr.17: 59838463-59838463 on Build GRCh38
rs371609859 Chr.17: 59838301-59838301 on Build GRCh38
rs60027243 Chr.17: 59838480-59838480 on Build GRCh38
rs374252753 Chr.17: 59838439-59838439 on Build GRCh38
rs970872760 Chr.17: 59838644-59838644 on Build GRCh38
rs768792230 Chr.17: 59838440-59838440 on Build GRCh38
rs959263480 Chr.17: 59838582-59838582 on Build GRCh38
rs75110149 Chr.17: 59838585-59838585 on Build GRCh38
rs199774918 Chr.17: 59838441-59838441 on Build GRCh38
rs750847228 Chr.17: 59838252-59838252 on Build GRCh38
rs956018458 Chr.17: 59838517-59838517 on Build GRCh38
rs10491164 Chr.17: 59838652-59838652 on Build GRCh38
rs181256505 Chr.17: 59838450-59838450 on Build GRCh38
rs189965314 Chr.17: 59838268-59838268 on Build GRCh38
rs548744804 Chr.17: 59838357-59838357 on Build GRCh38
rs773796393 Chr.17: 59838378-59838378 on Build GRCh38
rs771798604 Chr.17: 59838305-59838305 on Build GRCh38
rs765665233 Chr.17: 59838373-59838373 on Build GRCh38
rs918419163 Chr.17: 59838630-59838630 on Build GRCh38
rs555395025 Chr.17: 59838618-59838618 on Build GRCh38
rs756722989 Chr.17: 59838398-59838398 on Build GRCh38
rs528047002 Chr.17: 59838443-59838443 on Build GRCh38
rs758258452 Chr.17: 59838411-59838411 on Build GRCh38
rs777111659 Chr.17: 59838287-59838287 on Build GRCh38
rs902020940 Chr.17: 59838483-59838483 on Build GRCh38
rs773522199 Chr.17: 59838398-59838399 on Build GRCh38
rs979980256 Chr.17: 59838285-59838285 on Build GRCh38
rs540991674 Chr.17: 59838666-59838666 on Build GRCh38
rs778872499 Chr.17: 59838304-59838304 on Build GRCh38
rs113718494 Chr.17: 59838245-59838245 on Build GRCh38
rs774543236 Chr.17: 59838667-59838667 on Build GRCh38
rs539671632 Chr.17: 59838461-59838461 on Build GRCh38
rs752433943 Chr.17: 59838387-59838387 on Build GRCh38
rs752380826 Chr.17: 59838282-59838282 on Build GRCh38
rs766529897 Chr.17: 59838430-59838430 on Build GRCh38
rs770304293 Chr.17: 59838261-59838261 on Build GRCh38
rs749300808 Chr.17: 59838288-59838288 on Build GRCh38
rs570088939 Chr.17: 59838527-59838527 on Build GRCh38
rs745793895 Chr.17: 59838402-59838402 on Build GRCh38
rs766842980 Chr.17: 59838380-59838380 on Build GRCh38
rs200319964 Chr.17: 59838323-59838323 on Build GRCh38
rs778838462 Chr.17: 59838399-59838399 on Build GRCh38
rs755696480 Chr.17: 59838390-59838390 on Build GRCh38
rs771126869 Chr.17: 59838591-59838591 on Build GRCh38
rs371666723 Chr.17: 59838266-59838266 on Build GRCh38
rs998067706 Chr.17: 59838496-59838496 on Build GRCh38
rs545174483 Chr.17: 59838242-59838242 on Build GRCh38
rs552882136 Chr.17: 59838675-59838675 on Build GRCh38
rs761913056 Chr.17: 59838371-59838371 on Build GRCh38
rs779793924 Chr.17: 59838413-59838413 on Build GRCh38
rs767288702 Chr.17: 59838271-59838271 on Build GRCh38
rs755712512 Chr.17: 59838284-59838284 on Build GRCh38
rs1000513812 Chr.17: 59838501-59838501 on Build GRCh38
rs773212213 Chr.17: 59838528-59838528 on Build GRCh38
rs989844752 Chr.17: 59838229-59838229 on Build GRCh38
rs763758018 Chr.17: 59838394-59838394 on Build GRCh38
rs749358810 Chr.17: 59838565-59838565 on Build GRCh38
rs747227852 Chr.17: 59838339-59838339 on Build GRCh38
rs548133375 Chr.17: 59838445-59838445 on Build GRCh38

View Full Product Details