Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 62480809-62481357 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TLK2
Entrez Gene ID
11011
Gene Name
tousled like kinase 2
Gene Aliases
HsHPK, PKU-ALPHA
UniGene
Hs.445078
Gene Chromosome Location
Chr.17: 62477481-62615481 on Build GRCh38
Transcripts
NM_001284333.1 NM_001284363.1 NM_006852.3 XM_011524215.2 XM_011524216.2 XM_011524217.2 XM_011524218.2 XM_011524220.2 XM_011524221.2 XM_011524222.2 XM_011524223.2 XM_011524224.2 XM_011524225.2 XM_011524226.2 XM_011524227.2 XM_011524228.2 XM_011524229.2 XM_011524231.1 XM_017024044.1 XM_017024045.1 XM_017024046.1 XM_017024047.1 XM_017024048.1 XM_017024049.1 XM_017024050.1 XM_017024051.1 XM_017024052.1 XM_017024053.1 XM_017024054.1 XM_017024055.1 XM_017024056.1 XM_017024057.1 XM_017024058.1 XM_017024059.1 XM_017024060.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4442793 c.31C>T p.R11* Chr.17: 62481156-62481156 on Build GRCh38
4068460 c.48G>A p.L16L Chr.17: 62481173-62481173 on Build GRCh38
6802336 c.10G>T p.E4* Chr.17: 62481135-62481135 on Build GRCh38
4068461 c.48G>A p.L16L Chr.17: 62481173-62481173 on Build GRCh38
3717516 c.81G>A p.K27K Chr.17: 62481206-62481206 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780467453 Chr.17: 62481089-62481089 on Build GRCh38
rs371461807 Chr.17: 62481083-62481083 on Build GRCh38
rs572413273 Chr.17: 62480964-62480964 on Build GRCh38
rs752803734 Chr.17: 62481256-62481257 on Build GRCh38
rs543091761 Chr.17: 62480982-62480982 on Build GRCh38
rs760662218 Chr.17: 62481220-62481220 on Build GRCh38
rs1031387285 Chr.17: 62481021-62481021 on Build GRCh38
rs11514500 Chr.17: 62481283-62481283 on Build GRCh38
rs767892159 Chr.17: 62481221-62481221 on Build GRCh38
rs747275126 Chr.17: 62481092-62481092 on Build GRCh38
rs547996339 Chr.17: 62480946-62480946 on Build GRCh38
rs774967353 Chr.17: 62481143-62481143 on Build GRCh38
rs771592366 Chr.17: 62481121-62481121 on Build GRCh38
rs777416664 Chr.17: 62481258-62481258 on Build GRCh38
rs751606010 Chr.17: 62481303-62481310 on Build GRCh38
rs796730233 Chr.17: 62481159-62481159 on Build GRCh38
rs568121416 Chr.17: 62481013-62481013 on Build GRCh38
rs866608902 Chr.17: 62481257-62481257 on Build GRCh38
rs796144410 Chr.17: 62481160-62481160 on Build GRCh38
rs201542232 Chr.17: 62481101-62481101 on Build GRCh38
rs751665045 Chr.17: 62481250-62481250 on Build GRCh38
rs116017183 Chr.17: 62481256-62481256 on Build GRCh38
rs889778045 Chr.17: 62481323-62481323 on Build GRCh38
rs1028298481 Chr.17: 62480863-62480863 on Build GRCh38
rs576378664 Chr.17: 62481318-62481318 on Build GRCh38
rs374327169 Chr.17: 62481245-62481245 on Build GRCh38
rs45550140 Chr.17: 62481142-62481142 on Build GRCh38
rs779126513 Chr.17: 62481078-62481078 on Build GRCh38
rs554401949 Chr.17: 62481114-62481114 on Build GRCh38
rs989790210 Chr.17: 62481048-62481048 on Build GRCh38
rs796353186 Chr.17: 62481157-62481157 on Build GRCh38
rs879037638 Chr.17: 62481307-62481307 on Build GRCh38
rs957086962 Chr.17: 62481029-62481029 on Build GRCh38
rs766594574 Chr.17: 62481243-62481243 on Build GRCh38
rs759210184 Chr.17: 62480838-62480838 on Build GRCh38
rs750505615 Chr.17: 62481237-62481237 on Build GRCh38
rs377519248 Chr.17: 62481221-62481221 on Build GRCh38
rs1007459158 Chr.17: 62480910-62480910 on Build GRCh38
rs889055944 Chr.17: 62480867-62480867 on Build GRCh38
rs750585478 Chr.17: 62481276-62481276 on Build GRCh38
rs556547791 Chr.17: 62481298-62481298 on Build GRCh38
rs769432252 Chr.17: 62481099-62481099 on Build GRCh38
rs915557563 Chr.17: 62481059-62481059 on Build GRCh38
rs752518206 Chr.17: 62481106-62481106 on Build GRCh38
rs764310563 Chr.17: 62481202-62481202 on Build GRCh38
rs537819528 Chr.17: 62480855-62480855 on Build GRCh38
rs760380556 Chr.17: 62481192-62481192 on Build GRCh38
rs762409167 Chr.17: 62481226-62481226 on Build GRCh38
rs762938024 Chr.17: 62481140-62481140 on Build GRCh38
rs375836581 Chr.17: 62480984-62480984 on Build GRCh38
rs758500890 Chr.17: 62481238-62481238 on Build GRCh38
rs770913379 Chr.17: 62481103-62481103 on Build GRCh38
rs554794059 Chr.17: 62481082-62481082 on Build GRCh38
rs773845384 Chr.17: 62481112-62481112 on Build GRCh38
rs760617063 Chr.17: 62481301-62481301 on Build GRCh38
rs370542297 Chr.17: 62481235-62481235 on Build GRCh38
rs546616404 Chr.17: 62480844-62480844 on Build GRCh38
rs755150783 Chr.17: 62481078-62481079 on Build GRCh38

View Full Product Details