Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 44924632-44925136 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIF18B
Entrez Gene ID
146909
Gene Name
kinesin family member 18B
Gene Aliases
-
UniGene
Hs.135094
Gene Chromosome Location
Chr.17: 44924711-44947746 on Build GRCh38
Transcripts
NM_001264573.1 NM_001265577.1 XM_011524385.2 XM_011524386.2 XM_011524387.2 XM_011524388.2 XM_011524389.2 XM_011524390.2

SNP Detail

SNP ID Chromosome Location
rs572463318 Chr.17: 44924723-44924723 on Build GRCh38
rs781218432 Chr.17: 44924892-44924892 on Build GRCh38
rs893652937 Chr.17: 44925071-44925071 on Build GRCh38
rs778151368 Chr.17: 44924920-44924920 on Build GRCh38
rs199739153 Chr.17: 44924857-44924860 on Build GRCh38
rs534905426 Chr.17: 44925075-44925075 on Build GRCh38
rs953376336 Chr.17: 44924857-44924857 on Build GRCh38
rs886802830 Chr.17: 44924870-44924870 on Build GRCh38
rs1010971586 Chr.17: 44925101-44925101 on Build GRCh38
rs112349208 Chr.17: 44924863-44924863 on Build GRCh38
rs1031080353 Chr.17: 44924807-44924807 on Build GRCh38
rs940857315 Chr.17: 44924676-44924676 on Build GRCh38
rs912040401 Chr.17: 44924662-44924662 on Build GRCh38
rs111886688 Chr.17: 44924864-44924864 on Build GRCh38
rs940862089 Chr.17: 44924859-44924859 on Build GRCh38
rs751630399 Chr.17: 44924692-44924692 on Build GRCh38
rs747308869 Chr.17: 44924962-44924962 on Build GRCh38
rs3837838 Chr.17: 44924858-44924861 on Build GRCh38
rs576993701 Chr.17: 44924902-44924902 on Build GRCh38
rs949035309 Chr.17: 44925113-44925113 on Build GRCh38
rs986817403 Chr.17: 44925039-44925039 on Build GRCh38
rs112719720 Chr.17: 44924865-44924865 on Build GRCh38
rs906331079 Chr.17: 44924757-44924758 on Build GRCh38
rs1048146965 Chr.17: 44924914-44924914 on Build GRCh38
rs1024868223 Chr.17: 44924752-44924752 on Build GRCh38
rs895056317 Chr.17: 44924722-44924722 on Build GRCh38
rs942602594 Chr.17: 44925061-44925061 on Build GRCh38
rs971632687 Chr.17: 44924932-44924932 on Build GRCh38
rs1012092290 Chr.17: 44924748-44924748 on Build GRCh38
rs1059248 Chr.17: 44924786-44924786 on Build GRCh38
rs561480547 Chr.17: 44924949-44924949 on Build GRCh38
rs1034495274 Chr.17: 44924979-44924979 on Build GRCh38
rs987682544 Chr.17: 44924858-44924859 on Build GRCh38
rs749397039 Chr.17: 44924857-44924858 on Build GRCh38
rs990217663 Chr.17: 44925028-44925028 on Build GRCh38
rs931120928 Chr.17: 44925089-44925089 on Build GRCh38
rs542744923 Chr.17: 44924768-44924768 on Build GRCh38
rs1028968878 Chr.17: 44924855-44924855 on Build GRCh38
rs967388421 Chr.17: 44925030-44925030 on Build GRCh38
rs375478979 Chr.17: 44925060-44925060 on Build GRCh38
rs1039504464 Chr.17: 44924731-44924731 on Build GRCh38
rs188412605 Chr.17: 44924682-44924682 on Build GRCh38
rs371900576 Chr.17: 44924821-44924821 on Build GRCh38
rs1046880559 Chr.17: 44925093-44925093 on Build GRCh38
rs1013250343 Chr.17: 44924875-44924875 on Build GRCh38
rs559296513 Chr.17: 44925114-44925114 on Build GRCh38
rs958385066 Chr.17: 44924825-44924825 on Build GRCh38
rs547146850 Chr.17: 44925108-44925108 on Build GRCh38
rs757475018 Chr.17: 44924808-44924808 on Build GRCh38
rs919779234 Chr.17: 44925087-44925087 on Build GRCh38
rs1031278193 Chr.17: 44925068-44925068 on Build GRCh38
rs543446550 Chr.17: 44925015-44925015 on Build GRCh38
rs564804919 Chr.17: 44925036-44925036 on Build GRCh38
rs746118181 Chr.17: 44924934-44924934 on Build GRCh38
rs532297723 Chr.17: 44925038-44925038 on Build GRCh38
rs576337706 Chr.17: 44925012-44925012 on Build GRCh38
rs989467767 Chr.17: 44924826-44924826 on Build GRCh38
rs1050994167 Chr.17: 44924683-44924683 on Build GRCh38
rs1059249 Chr.17: 44924772-44924772 on Build GRCh38
rs898250090 Chr.17: 44924760-44924760 on Build GRCh38
rs1036750459 Chr.17: 44924681-44924681 on Build GRCh38
rs931095638 Chr.17: 44924913-44924913 on Build GRCh38
rs913308707 Chr.17: 44925029-44925029 on Build GRCh38
rs1048073009 Chr.17: 44924714-44924714 on Build GRCh38

View Full Product Details