Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 44928326-44928823 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIF18B
Entrez Gene ID
146909
Gene Name
kinesin family member 18B
Gene Aliases
-
UniGene
Hs.135094
Gene Chromosome Location
Chr.17: 44924711-44947746 on Build GRCh38
Transcripts
NM_001264573.1 NM_001265577.1 XM_011524385.2 XM_011524386.2 XM_011524387.2 XM_011524388.2 XM_011524389.2 XM_011524390.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7259911 c.2006G>A p.C669Y Chr.17: 44928332-44928332 on Build GRCh38
5762185 c.1838G>T p.S613I Chr.17: 44928500-44928500 on Build GRCh38
1212387 c.1987C>T p.R663C Chr.17: 44928351-44928351 on Build GRCh38
6523596 c.1769C>T p.P590L Chr.17: 44928569-44928569 on Build GRCh38
6563780 c.1843C>T p.P615S Chr.17: 44928495-44928495 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs918183393 Chr.17: 44928381-44928381 on Build GRCh38
rs60374269 Chr.17: 44928354-44928354 on Build GRCh38
rs945916845 Chr.17: 44928624-44928624 on Build GRCh38
rs1000983612 Chr.17: 44928463-44928463 on Build GRCh38
rs766863396 Chr.17: 44928540-44928540 on Build GRCh38
rs562184263 Chr.17: 44928494-44928494 on Build GRCh38
rs533415241 Chr.17: 44928626-44928626 on Build GRCh38
rs1045326664 Chr.17: 44928760-44928760 on Build GRCh38
rs769683332 Chr.17: 44928502-44928502 on Build GRCh38
rs756421332 Chr.17: 44928414-44928414 on Build GRCh38
rs936046222 Chr.17: 44928351-44928351 on Build GRCh38
rs369863717 Chr.17: 44928535-44928535 on Build GRCh38
rs528668469 Chr.17: 44928458-44928458 on Build GRCh38
rs777380757 Chr.17: 44928522-44928522 on Build GRCh38
rs779155006 Chr.17: 44928438-44928438 on Build GRCh38
rs199951508 Chr.17: 44928506-44928506 on Build GRCh38
rs754205877 Chr.17: 44928564-44928564 on Build GRCh38
rs757119537 Chr.17: 44928578-44928578 on Build GRCh38
rs189290711 Chr.17: 44928663-44928663 on Build GRCh38
rs115310285 Chr.17: 44928748-44928748 on Build GRCh38
rs746876013 Chr.17: 44928481-44928481 on Build GRCh38
rs767059833 Chr.17: 44928418-44928418 on Build GRCh38
rs755571528 Chr.17: 44928573-44928573 on Build GRCh38
rs757426054 Chr.17: 44928796-44928796 on Build GRCh38
rs201643239 Chr.17: 44928501-44928501 on Build GRCh38
rs935827825 Chr.17: 44928733-44928733 on Build GRCh38
rs759406754 Chr.17: 44928380-44928380 on Build GRCh38
rs1052572708 Chr.17: 44928472-44928472 on Build GRCh38
rs755104339 Chr.17: 44928356-44928356 on Build GRCh38
rs770901247 Chr.17: 44928371-44928371 on Build GRCh38
rs892508846 Chr.17: 44928500-44928500 on Build GRCh38
rs546084222 Chr.17: 44928357-44928357 on Build GRCh38
rs758391211 Chr.17: 44928779-44928779 on Build GRCh38
rs762699986 Chr.17: 44928402-44928402 on Build GRCh38
rs776532109 Chr.17: 44928374-44928374 on Build GRCh38
rs992525693 Chr.17: 44928680-44928680 on Build GRCh38
rs1004400586 Chr.17: 44928795-44928795 on Build GRCh38
rs770704894 Chr.17: 44928486-44928486 on Build GRCh38
rs887385334 Chr.17: 44928417-44928419 on Build GRCh38
rs200142587 Chr.17: 44928789-44928789 on Build GRCh38
rs569266892 Chr.17: 44928617-44928617 on Build GRCh38
rs777832643 Chr.17: 44928473-44928473 on Build GRCh38
rs371805837 Chr.17: 44928368-44928368 on Build GRCh38
rs1016181612 Chr.17: 44928568-44928568 on Build GRCh38
rs762349356 Chr.17: 44928511-44928511 on Build GRCh38
rs111810720 Chr.17: 44928455-44928455 on Build GRCh38
rs774935576 Chr.17: 44928398-44928398 on Build GRCh38
rs763624751 Chr.17: 44928403-44928403 on Build GRCh38
rs746703063 Chr.17: 44928726-44928726 on Build GRCh38
rs898657884 Chr.17: 44928797-44928797 on Build GRCh38
rs887123677 Chr.17: 44928790-44928790 on Build GRCh38
rs35432916 Chr.17: 44928397-44928398 on Build GRCh38
rs566690007 Chr.17: 44928741-44928741 on Build GRCh38
rs367973974 Chr.17: 44928432-44928432 on Build GRCh38
rs746748162 Chr.17: 44928369-44928369 on Build GRCh38
rs35121533 Chr.17: 44928427-44928428 on Build GRCh38
rs765806954 Chr.17: 44928616-44928616 on Build GRCh38
rs555272626 Chr.17: 44928762-44928762 on Build GRCh38
rs764743878 Chr.17: 44928388-44928388 on Build GRCh38
rs371129096 Chr.17: 44928350-44928350 on Build GRCh38
rs371973434 Chr.17: 44928512-44928512 on Build GRCh38
rs567311715 Chr.17: 44928768-44928768 on Build GRCh38
rs755484398 Chr.17: 44928434-44928434 on Build GRCh38
rs763790347 Chr.17: 44928404-44928404 on Build GRCh38
rs149288036 Chr.17: 44928528-44928528 on Build GRCh38
rs960030315 Chr.17: 44928675-44928675 on Build GRCh38
rs773872542 Chr.17: 44928523-44928523 on Build GRCh38
rs183876562 Chr.17: 44928464-44928464 on Build GRCh38
rs759820172 Chr.17: 44928364-44928364 on Build GRCh38
rs772134493 Chr.17: 44928367-44928367 on Build GRCh38
rs751410509 Chr.17: 44928442-44928442 on Build GRCh38

View Full Product Details