Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 45135963-45136472 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACBD4
Entrez Gene ID
79777
Gene Name
acyl-CoA binding domain containing 4
Gene Aliases
HMFT0700
UniGene
Hs.110298
Gene Chromosome Location
Chr.17: 45132600-45144181 on Build GRCh38
Transcripts
NM_001135705.2 NM_001135706.2 NM_001135707.2 NM_001321352.1 NM_001321353.1 NM_024722.3 XM_006722085.2 XM_011525259.2 XM_011525261.2 XM_017025084.1 XM_017025085.1 XM_017025086.1 XM_017025087.1 XM_017025088.1 XM_017025089.1 XM_017025090.1 XM_017025091.1 XM_017025092.1 XM_017025093.1 XM_017025094.1 XM_017025095.1 XM_017025096.1 XM_017025097.1 XM_017025098.1 XM_017025099.1 XM_017025100.1 XM_017025101.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5030516 c.69C>G p.I23M Chr.17: 45136213-45136213 on Build GRCh38
5055527 c.67A>G p.I23V Chr.17: 45136211-45136211 on Build GRCh38
3191954 c.4G>T p.G2C Chr.17: 45136148-45136148 on Build GRCh38
3191953 c.4G>T p.G2C Chr.17: 45136148-45136148 on Build GRCh38
4835883 c.60G>A p.V20V Chr.17: 45136204-45136204 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757756306 Chr.17: 45136252-45136252 on Build GRCh38
rs562531927 Chr.17: 45136136-45136136 on Build GRCh38
rs377728794 Chr.17: 45136162-45136162 on Build GRCh38
rs370856181 Chr.17: 45136213-45136213 on Build GRCh38
rs539334500 Chr.17: 45135987-45135987 on Build GRCh38
rs1000165344 Chr.17: 45136307-45136307 on Build GRCh38
rs951646357 Chr.17: 45136372-45136372 on Build GRCh38
rs758757918 Chr.17: 45136156-45136156 on Build GRCh38
rs770050455 Chr.17: 45136199-45136199 on Build GRCh38
rs762194858 Chr.17: 45136401-45136401 on Build GRCh38
rs374150924 Chr.17: 45136276-45136276 on Build GRCh38
rs778134360 Chr.17: 45136245-45136245 on Build GRCh38
rs907557676 Chr.17: 45136116-45136116 on Build GRCh38
rs200581584 Chr.17: 45136165-45136165 on Build GRCh38
rs199574823 Chr.17: 45136148-45136148 on Build GRCh38
rs372421100 Chr.17: 45136375-45136375 on Build GRCh38
rs1018278609 Chr.17: 45136311-45136311 on Build GRCh38
rs376895522 Chr.17: 45136047-45136047 on Build GRCh38
rs541468728 Chr.17: 45136132-45136132 on Build GRCh38
rs201940756 Chr.17: 45136161-45136161 on Build GRCh38
rs574483257 Chr.17: 45136052-45136052 on Build GRCh38
rs937121767 Chr.17: 45136435-45136435 on Build GRCh38
rs773750392 Chr.17: 45136437-45136437 on Build GRCh38
rs547397259 Chr.17: 45136362-45136362 on Build GRCh38
rs762833461 Chr.17: 45136205-45136205 on Build GRCh38
rs200711802 Chr.17: 45136445-45136445 on Build GRCh38
rs773894998 Chr.17: 45136280-45136280 on Build GRCh38
rs998361090 Chr.17: 45136358-45136358 on Build GRCh38
rs964151380 Chr.17: 45136000-45136000 on Build GRCh38
rs1017213313 Chr.17: 45136388-45136388 on Build GRCh38
rs763904286 Chr.17: 45136211-45136211 on Build GRCh38
rs977330271 Chr.17: 45136013-45136013 on Build GRCh38
rs775470080 Chr.17: 45136202-45136202 on Build GRCh38
rs779376946 Chr.17: 45136153-45136153 on Build GRCh38
rs796925582 Chr.17: 45136439-45136439 on Build GRCh38
rs532411432 Chr.17: 45136352-45136352 on Build GRCh38
rs563347192 Chr.17: 45136154-45136154 on Build GRCh38
rs113924400 Chr.17: 45136258-45136258 on Build GRCh38
rs559175008 Chr.17: 45136364-45136364 on Build GRCh38
rs765845136 Chr.17: 45136239-45136239 on Build GRCh38
rs374047564 Chr.17: 45136224-45136224 on Build GRCh38
rs951755486 Chr.17: 45136074-45136074 on Build GRCh38
rs200345749 Chr.17: 45136109-45136109 on Build GRCh38
rs554542844 Chr.17: 45136083-45136083 on Build GRCh38
rs374486547 Chr.17: 45136185-45136185 on Build GRCh38
rs748801834 Chr.17: 45136146-45136146 on Build GRCh38
rs529958234 Chr.17: 45136391-45136391 on Build GRCh38
rs751969715 Chr.17: 45136251-45136251 on Build GRCh38
rs746135410 Chr.17: 45136267-45136267 on Build GRCh38
rs781544591 Chr.17: 45136263-45136263 on Build GRCh38
rs899927982 Chr.17: 45136351-45136351 on Build GRCh38
rs530539203 Chr.17: 45136232-45136232 on Build GRCh38
rs769957662 Chr.17: 45136270-45136270 on Build GRCh38
rs2306828 Chr.17: 45136405-45136405 on Build GRCh38
rs1023106618 Chr.17: 45136334-45136334 on Build GRCh38
rs915163190 Chr.17: 45136393-45136393 on Build GRCh38
rs758850521 Chr.17: 45136243-45136243 on Build GRCh38
rs983417548 Chr.17: 45136387-45136387 on Build GRCh38
rs149561067 Chr.17: 45136242-45136242 on Build GRCh38
rs372278855 Chr.17: 45136285-45136285 on Build GRCh38
rs1041700030 Chr.17: 45136279-45136279 on Build GRCh38
rs768576767 Chr.17: 45136244-45136245 on Build GRCh38
rs946538311 Chr.17: 45135999-45135999 on Build GRCh38
rs371955318 Chr.17: 45136274-45136274 on Build GRCh38

View Full Product Details