Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 45845613-45846118 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAPT-AS1
Entrez Gene ID
100128977
Gene Name
MAPT antisense RNA 1
Gene Aliases
-
UniGene
Hs.666302
Gene Chromosome Location
Chr.17: 45843356-45895513 on Build GRCh38
Transcripts
NR_024559.1 NM_175882.2

Gene Symbol
SPPL2C
Entrez Gene ID
162540
Gene Name
signal peptide peptidase like 2C
Gene Aliases
IMP5
UniGene
Hs.144491
Gene Chromosome Location
Chr.17: 45844890-45847072 on Build GRCh38
Transcripts
NR_024559.1 NM_175882.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3518318 c.825C>T p.D275D Chr.17: 45845731-45845731 on Build GRCh38
6498343 c.727G>C p.D243H Chr.17: 45845633-45845633 on Build GRCh38
5464071 c.760G>A p.A254T Chr.17: 45845666-45845666 on Build GRCh38
4067124 c.707C>A p.A236D Chr.17: 45845613-45845613 on Build GRCh38
5678141 c.991T>A p.C331S Chr.17: 45845897-45845897 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs375579334 Chr.17: 45845749-45845749 on Build GRCh38
rs950020106 Chr.17: 45845908-45845908 on Build GRCh38
rs557873899 Chr.17: 45845944-45845944 on Build GRCh38
rs967653621 Chr.17: 45845742-45845742 on Build GRCh38
rs750941235 Chr.17: 45846083-45846083 on Build GRCh38
rs758276593 Chr.17: 45845641-45845641 on Build GRCh38
rs1033636690 Chr.17: 45845789-45845789 on Build GRCh38
rs139341754 Chr.17: 45845678-45845678 on Build GRCh38
rs953142718 Chr.17: 45845839-45845839 on Build GRCh38
rs150854154 Chr.17: 45845662-45845662 on Build GRCh38
rs1043430015 Chr.17: 45845766-45845766 on Build GRCh38
rs943013046 Chr.17: 45845803-45845806 on Build GRCh38
rs760501071 Chr.17: 45846040-45846040 on Build GRCh38
rs758100393 Chr.17: 45845704-45845704 on Build GRCh38
rs759219260 Chr.17: 45846064-45846064 on Build GRCh38
rs747564396 Chr.17: 45845913-45845913 on Build GRCh38
rs780242914 Chr.17: 45846097-45846097 on Build GRCh38
rs775642446 Chr.17: 45845932-45845932 on Build GRCh38
rs376423300 Chr.17: 45845842-45845842 on Build GRCh38
rs868254309 Chr.17: 45845911-45845911 on Build GRCh38
rs141462911 Chr.17: 45846008-45846008 on Build GRCh38
rs747372075 Chr.17: 45845857-45845857 on Build GRCh38
rs140968506 Chr.17: 45846080-45846080 on Build GRCh38
rs979155945 Chr.17: 45845926-45845926 on Build GRCh38
rs370018511 Chr.17: 45845954-45845954 on Build GRCh38
rs754600209 Chr.17: 45845905-45845905 on Build GRCh38
rs778406852 Chr.17: 45845906-45845906 on Build GRCh38
rs141007000 Chr.17: 45845804-45845804 on Build GRCh38
rs1016290358 Chr.17: 45845652-45845652 on Build GRCh38
rs145566891 Chr.17: 45845899-45845899 on Build GRCh38
rs144966344 Chr.17: 45845813-45845813 on Build GRCh38
rs373967033 Chr.17: 45845999-45845999 on Build GRCh38
rs777587977 Chr.17: 45845706-45845706 on Build GRCh38
rs865792222 Chr.17: 45845700-45845700 on Build GRCh38
rs369312392 Chr.17: 45845796-45845796 on Build GRCh38
rs781651763 Chr.17: 45845929-45845929 on Build GRCh38
rs148824691 Chr.17: 45845825-45845825 on Build GRCh38
rs144752984 Chr.17: 45845873-45845873 on Build GRCh38
rs780809938 Chr.17: 45845819-45845819 on Build GRCh38
rs200647385 Chr.17: 45845650-45845650 on Build GRCh38
rs779097529 Chr.17: 45845943-45845943 on Build GRCh38
rs756848275 Chr.17: 45845724-45845724 on Build GRCh38
rs138704863 Chr.17: 45845889-45845889 on Build GRCh38
rs770604717 Chr.17: 45845654-45845654 on Build GRCh38
rs767879204 Chr.17: 45845664-45845664 on Build GRCh38
rs745379513 Chr.17: 45845738-45845738 on Build GRCh38
rs767339204 Chr.17: 45845896-45845896 on Build GRCh38
rs746708887 Chr.17: 45845653-45845653 on Build GRCh38
rs182389290 Chr.17: 45845740-45845740 on Build GRCh38
rs369336082 Chr.17: 45846002-45846002 on Build GRCh38
rs748240047 Chr.17: 45846007-45846007 on Build GRCh38
rs369440162 Chr.17: 45845647-45845647 on Build GRCh38
rs773661649 Chr.17: 45845759-45845759 on Build GRCh38
rs780828496 Chr.17: 45845731-45845731 on Build GRCh38
rs757783016 Chr.17: 45845983-45845983 on Build GRCh38
rs773087259 Chr.17: 45846025-45846025 on Build GRCh38
rs138116075 Chr.17: 45846056-45846056 on Build GRCh38
rs903539041 Chr.17: 45845769-45845769 on Build GRCh38
rs148108168 Chr.17: 45846015-45846015 on Build GRCh38
rs774989388 Chr.17: 45845665-45845665 on Build GRCh38
rs536730578 Chr.17: 45845689-45845689 on Build GRCh38
rs141910280 Chr.17: 45846030-45846030 on Build GRCh38
rs370058596 Chr.17: 45845656-45845657 on Build GRCh38
rs546882470 Chr.17: 45845638-45845638 on Build GRCh38
rs553257171 Chr.17: 45845843-45845843 on Build GRCh38
rs765077000 Chr.17: 45845697-45845697 on Build GRCh38
rs753908459 Chr.17: 45845687-45845687 on Build GRCh38
rs762071305 Chr.17: 45845674-45845674 on Build GRCh38
rs767972482 Chr.17: 45845677-45845677 on Build GRCh38
rs377723590 Chr.17: 45845950-45845950 on Build GRCh38
rs376789321 Chr.17: 45845977-45845977 on Build GRCh38
rs774363885 Chr.17: 45846003-45846003 on Build GRCh38
rs771209318 Chr.17: 45845778-45845778 on Build GRCh38
rs147727109 Chr.17: 45845757-45845757 on Build GRCh38
rs150041799 Chr.17: 45845722-45845722 on Build GRCh38
rs369966982 Chr.17: 45845894-45845894 on Build GRCh38
rs753319831 Chr.17: 45845904-45845904 on Build GRCh38
rs748729879 Chr.17: 45845750-45845750 on Build GRCh38
rs1045797370 Chr.17: 45845923-45845923 on Build GRCh38
rs776072847 Chr.17: 45845661-45845661 on Build GRCh38
rs750685052 Chr.17: 45845701-45845701 on Build GRCh38
rs761131514 Chr.17: 45845765-45845765 on Build GRCh38
rs373368449 Chr.17: 45845936-45845936 on Build GRCh38
rs778425499 Chr.17: 45845846-45845846 on Build GRCh38
rs769040377 Chr.17: 45845805-45845805 on Build GRCh38
rs62054815 Chr.17: 45845900-45845900 on Build GRCh38
rs758052686 Chr.17: 45846079-45846079 on Build GRCh38
rs1051606044 Chr.17: 45845642-45845642 on Build GRCh38
rs766047756 Chr.17: 45845949-45845949 on Build GRCh38
rs751557549 Chr.17: 45845893-45845893 on Build GRCh38
rs920390012 Chr.17: 45845882-45845882 on Build GRCh38
rs242944 Chr.17: 45845814-45845814 on Build GRCh38
rs373857259 Chr.17: 45845807-45845807 on Build GRCh38
rs368754074 Chr.17: 45845874-45845874 on Build GRCh38
rs756471815 Chr.17: 45846087-45846087 on Build GRCh38
rs769994819 Chr.17: 45845931-45845931 on Build GRCh38
rs757112085 Chr.17: 45845815-45845815 on Build GRCh38
rs149355286 Chr.17: 45845930-45845930 on Build GRCh38
rs752192485 Chr.17: 45846068-45846068 on Build GRCh38
rs750126870 Chr.17: 45845820-45845820 on Build GRCh38
rs761698150 Chr.17: 45845942-45845942 on Build GRCh38
rs140813443 Chr.17: 45845869-45845869 on Build GRCh38
rs752755138 Chr.17: 45845806-45845806 on Build GRCh38
rs148362814 Chr.17: 45845826-45845826 on Build GRCh38
rs200843300 Chr.17: 45845898-45845898 on Build GRCh38
rs764998078 Chr.17: 45846067-45846067 on Build GRCh38
rs150431364 Chr.17: 45846044-45846044 on Build GRCh38
rs768773015 Chr.17: 45845935-45845935 on Build GRCh38
rs747827104 Chr.17: 45845639-45845639 on Build GRCh38
rs777056651 Chr.17: 45845788-45845788 on Build GRCh38
rs913720141 Chr.17: 45845699-45845699 on Build GRCh38
rs754511759 Chr.17: 45845952-45845952 on Build GRCh38
rs771477040 Chr.17: 45845868-45845868 on Build GRCh38
rs781558369 Chr.17: 45845992-45845992 on Build GRCh38
rs749375562 Chr.17: 45845934-45845934 on Build GRCh38
rs141850346 Chr.17: 45845890-45845890 on Build GRCh38
rs376056516 Chr.17: 45846001-45846001 on Build GRCh38
rs200592127 Chr.17: 45846049-45846049 on Build GRCh38
rs756494530 Chr.17: 45845998-45845998 on Build GRCh38
rs150961436 Chr.17: 45845845-45845845 on Build GRCh38
rs757874674 Chr.17: 45845928-45845928 on Build GRCh38
rs568111018 Chr.17: 45845681-45845681 on Build GRCh38
rs1055892455 Chr.17: 45845686-45845686 on Build GRCh38
rs116972939 Chr.17: 45845993-45845993 on Build GRCh38

View Full Product Details