Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 46330966-46331468 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARL17B
Entrez Gene ID
100506084
Gene Name
ADP ribosylation factor like GTPase 17B
Gene Aliases
ARL17, ARL17A
UniGene
Hs.559259
Gene Chromosome Location
Chr.17: 46274184-46361872 on Build GRCh38
Transcripts
NM_001103154.1 XM_005256918.4 XM_011524160.2 XM_011524161.2 XM_011524162.2 XM_011524163.2 XM_011524164.2 XM_011524166.2 XM_011524167.2 NM_014834.4 XM_005257879.4 XM_005257880.4 XM_005257882.4 XM_006722210.3 XM_006722211.3 XM_011525539.2 XM_017025472.1 XM_017025473.1

Gene Symbol
LRRC37A
Entrez Gene ID
9884
Gene Name
leucine rich repeat containing 37A
Gene Aliases
LRRC37
UniGene
Hs.579108
Gene Chromosome Location
Chr.17: 46248044-46339367 on Build GRCh38
Transcripts
NM_001103154.1 XM_005256918.4 XM_011524160.2 XM_011524161.2 XM_011524162.2 XM_011524163.2 XM_011524164.2 XM_011524166.2 XM_011524167.2 NM_014834.4 XM_005257879.4 XM_005257880.4 XM_005257882.4 XM_006722210.3 XM_006722211.3 XM_011525539.2 XM_017025472.1 XM_017025473.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6336402 c.4029A>G p.P1343P Chr.17: 46331306-46331306 on Build GRCh38
5055535 c.3883T>C p.S1295P Chr.17: 46331160-46331160 on Build GRCh38
6081115 c.3737C>A p.S1246Y Chr.17: 46331014-46331014 on Build GRCh38
3958453 c.3863G>C p.R1288T Chr.17: 46331140-46331140 on Build GRCh38
4598670 c.3815C>G p.S1272W Chr.17: 46331092-46331092 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755374634 Chr.17: 46331401-46331401 on Build GRCh38
rs530674879 Chr.17: 46331119-46331119 on Build GRCh38
rs755143842 Chr.17: 46331231-46331231 on Build GRCh38
rs2667848 Chr.17: 46331429-46331429 on Build GRCh38
rs566622963 Chr.17: 46331346-46331346 on Build GRCh38
rs550966947 Chr.17: 46331148-46331148 on Build GRCh38
rs757660633 Chr.17: 46331023-46331023 on Build GRCh38
rs879123478 Chr.17: 46331194-46331194 on Build GRCh38
rs562017927 Chr.17: 46331109-46331109 on Build GRCh38
rs376252120 Chr.17: 46331159-46331159 on Build GRCh38
rs774826403 Chr.17: 46331171-46331171 on Build GRCh38
rs768689256 Chr.17: 46331049-46331049 on Build GRCh38
rs763723453 Chr.17: 46331337-46331337 on Build GRCh38
rs781740217 Chr.17: 46331031-46331031 on Build GRCh38
rs372382482 Chr.17: 46331041-46331041 on Build GRCh38
rs776394391 Chr.17: 46331137-46331137 on Build GRCh38
rs570541063 Chr.17: 46331217-46331217 on Build GRCh38
rs758563409 Chr.17: 46331251-46331251 on Build GRCh38
rs750983877 Chr.17: 46331199-46331199 on Build GRCh38
rs747104743 Chr.17: 46331420-46331420 on Build GRCh38
rs541655426 Chr.17: 46331075-46331075 on Build GRCh38
rs746985821 Chr.17: 46331278-46331278 on Build GRCh38
rs752087224 Chr.17: 46330990-46330990 on Build GRCh38
rs754163264 Chr.17: 46331370-46331370 on Build GRCh38
rs778046922 Chr.17: 46331414-46331414 on Build GRCh38
rs776521338 Chr.17: 46331300-46331300 on Build GRCh38
rs775218060 Chr.17: 46331317-46331317 on Build GRCh38
rs765363221 Chr.17: 46331406-46331406 on Build GRCh38
rs8065693 Chr.17: 46331040-46331040 on Build GRCh38
rs762353221 Chr.17: 46331318-46331318 on Build GRCh38
rs749577078 Chr.17: 46331042-46331042 on Build GRCh38
rs746787835 Chr.17: 46331113-46331113 on Build GRCh38
rs757216962 Chr.17: 46331425-46331425 on Build GRCh38
rs559472762 Chr.17: 46331033-46331033 on Build GRCh38
rs759114190 Chr.17: 46330985-46330985 on Build GRCh38
rs756464366 Chr.17: 46331213-46331213 on Build GRCh38
rs200993737 Chr.17: 46331232-46331232 on Build GRCh38
rs748119666 Chr.17: 46331248-46331248 on Build GRCh38
rs770829393 Chr.17: 46331292-46331292 on Build GRCh38
rs17855643 Chr.17: 46331428-46331428 on Build GRCh38
rs199683082 Chr.17: 46331092-46331092 on Build GRCh38
rs149365331 Chr.17: 46331256-46331256 on Build GRCh38
rs368493396 Chr.17: 46331193-46331193 on Build GRCh38
rs546731558 Chr.17: 46331306-46331306 on Build GRCh38
rs773675556 Chr.17: 46331343-46331343 on Build GRCh38
rs771864444 Chr.17: 46331096-46331096 on Build GRCh38
rs273532 Chr.17: 46331410-46331410 on Build GRCh38
rs147486645 Chr.17: 46331426-46331426 on Build GRCh38
rs533141888 Chr.17: 46331302-46331302 on Build GRCh38
rs753029452 Chr.17: 46331407-46331407 on Build GRCh38
rs778908423 Chr.17: 46331057-46331057 on Build GRCh38
rs762524182 Chr.17: 46331192-46331192 on Build GRCh38
rs754036546 Chr.17: 46331225-46331225 on Build GRCh38
rs766893322 Chr.17: 46331358-46331358 on Build GRCh38

View Full Product Details