Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 46728741-46729239 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NSF
Entrez Gene ID
4905
Gene Name
N-ethylmaleimide sensitive factor, vesicle fusing ATPase
Gene Aliases
SKD2
UniGene
Hs.431279
Gene Chromosome Location
Chr.17: 46590669-46757464 on Build GRCh38
Transcripts
NM_006178.3 NR_040116.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6369486 c.1889T>C p.L630P Chr.17: 46728915-46728915 on Build GRCh38
706057 c.1833C>T p.Y611Y Chr.17: 46728859-46728859 on Build GRCh38
2695989 c.1834G>A p.V612I Chr.17: 46728860-46728860 on Build GRCh38
6398516 c.1897G>A p.A633T Chr.17: 46728923-46728923 on Build GRCh38
980333 c.1861C>A p.L621I Chr.17: 46728887-46728887 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs376204380 Chr.17: 46728933-46728933 on Build GRCh38
rs764779910 Chr.17: 46728819-46728819 on Build GRCh38
rs925068138 Chr.17: 46729045-46729045 on Build GRCh38
rs923201215 Chr.17: 46728774-46728774 on Build GRCh38
rs202122205 Chr.17: 46729157-46729157 on Build GRCh38
rs764702820 Chr.17: 46728769-46728769 on Build GRCh38
rs759440444 Chr.17: 46728815-46728815 on Build GRCh38
rs755313462 Chr.17: 46728857-46728857 on Build GRCh38
rs933258971 Chr.17: 46729159-46729159 on Build GRCh38
rs770765055 Chr.17: 46728928-46728928 on Build GRCh38
rs940050065 Chr.17: 46729217-46729217 on Build GRCh38
rs747124941 Chr.17: 46728924-46728924 on Build GRCh38
rs752025843 Chr.17: 46728969-46728970 on Build GRCh38
rs376923479 Chr.17: 46728773-46728774 on Build GRCh38
rs370293583 Chr.17: 46728949-46728949 on Build GRCh38
rs974575259 Chr.17: 46729075-46729075 on Build GRCh38
rs766818164 Chr.17: 46728847-46728847 on Build GRCh38
rs1013551677 Chr.17: 46728871-46728871 on Build GRCh38
rs755138580 Chr.17: 46728968-46728969 on Build GRCh38
rs1007014791 Chr.17: 46729215-46729215 on Build GRCh38
rs202140145 Chr.17: 46729196-46729196 on Build GRCh38
rs1011970196 Chr.17: 46729082-46729082 on Build GRCh38
rs1002396322 Chr.17: 46728989-46728989 on Build GRCh38
rs752285996 Chr.17: 46728824-46728824 on Build GRCh38
rs377523999 Chr.17: 46728969-46728969 on Build GRCh38
rs778166889 Chr.17: 46729000-46729000 on Build GRCh38
rs750550053 Chr.17: 46729181-46729181 on Build GRCh38
rs62074086 Chr.17: 46728946-46728946 on Build GRCh38
rs756551764 Chr.17: 46728837-46728837 on Build GRCh38
rs758899516 Chr.17: 46728992-46728992 on Build GRCh38
rs1048732213 Chr.17: 46728902-46728902 on Build GRCh38
rs531135277 Chr.17: 46728892-46728892 on Build GRCh38
rs867657686 Chr.17: 46729004-46729004 on Build GRCh38
rs1020618733 Chr.17: 46729020-46729020 on Build GRCh38
rs773404010 Chr.17: 46728908-46728908 on Build GRCh38
rs1017447120 Chr.17: 46729218-46729218 on Build GRCh38
rs758080952 Chr.17: 46728825-46728825 on Build GRCh38
rs923065442 Chr.17: 46729109-46729109 on Build GRCh38
rs775289653 Chr.17: 46728955-46728955 on Build GRCh38
rs772443136 Chr.17: 46728907-46728907 on Build GRCh38
rs779152160 Chr.17: 46728875-46728875 on Build GRCh38
rs749602886 Chr.17: 46728852-46728852 on Build GRCh38
rs566313333 Chr.17: 46729067-46729067 on Build GRCh38
rs370377514 Chr.17: 46728940-46728940 on Build GRCh38
rs369915141 Chr.17: 46728843-46728843 on Build GRCh38
rs966382446 Chr.17: 46729026-46729026 on Build GRCh38
rs763548545 Chr.17: 46728828-46728828 on Build GRCh38
rs751123815 Chr.17: 46728829-46728829 on Build GRCh38
rs75013657 Chr.17: 46729034-46729034 on Build GRCh38
rs776256020 Chr.17: 46728808-46728808 on Build GRCh38
rs752187129 Chr.17: 46728784-46728784 on Build GRCh38
rs1013785641 Chr.17: 46729019-46729019 on Build GRCh38
rs954098745 Chr.17: 46729167-46729167 on Build GRCh38
rs892063404 Chr.17: 46729072-46729072 on Build GRCh38
rs1051103310 Chr.17: 46728789-46728789 on Build GRCh38
rs751068720 Chr.17: 46728977-46728977 on Build GRCh38
rs373778816 Chr.17: 46728899-46728899 on Build GRCh38
rs184650201 Chr.17: 46728943-46728943 on Build GRCh38
rs1053748055 Chr.17: 46729066-46729066 on Build GRCh38
rs551707911 Chr.17: 46729180-46729180 on Build GRCh38
rs781485853 Chr.17: 46728971-46728972 on Build GRCh38
rs762574496 Chr.17: 46728967-46728967 on Build GRCh38

View Full Product Details