Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 46751338-46751871 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NSF
Entrez Gene ID
4905
Gene Name
N-ethylmaleimide sensitive factor, vesicle fusing ATPase
Gene Aliases
SKD2
UniGene
Hs.431279
Gene Chromosome Location
Chr.17: 46590669-46757464 on Build GRCh38
Transcripts
NM_006178.3 NR_040116.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3387975 c.2142C>G p.I714M Chr.17: 46751601-46751601 on Build GRCh38
3387974 c.2124G>C p.K708N Chr.17: 46751583-46751583 on Build GRCh38
7318120 c.2124G>A p.K708K Chr.17: 46751583-46751583 on Build GRCh38
80246 c.2088G>A p.Q696Q Chr.17: 46751547-46751547 on Build GRCh38
5342948 c.2071C>T p.R691C Chr.17: 46751530-46751530 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs540304545 Chr.17: 46751530-46751530 on Build GRCh38
rs1004072173 Chr.17: 46751729-46751729 on Build GRCh38
rs749238369 Chr.17: 46751491-46751491 on Build GRCh38
rs780112172 Chr.17: 46751624-46751624 on Build GRCh38
rs888278412 Chr.17: 46751387-46751387 on Build GRCh38
rs769722145 Chr.17: 46751452-46751452 on Build GRCh38
rs529221896 Chr.17: 46751572-46751572 on Build GRCh38
rs987926347 Chr.17: 46751406-46751406 on Build GRCh38
rs370654068 Chr.17: 46751601-46751601 on Build GRCh38
rs202064467 Chr.17: 46751568-46751568 on Build GRCh38
rs751921136 Chr.17: 46751582-46751582 on Build GRCh38
rs368783747 Chr.17: 46751655-46751655 on Build GRCh38
rs1050878380 Chr.17: 46751652-46751652 on Build GRCh38
rs76289190 Chr.17: 46751479-46751479 on Build GRCh38
rs1038205376 Chr.17: 46751762-46751762 on Build GRCh38
rs750507104 Chr.17: 46751610-46751610 on Build GRCh38
rs112056493 Chr.17: 46751718-46751718 on Build GRCh38
rs374687013 Chr.17: 46751639-46751639 on Build GRCh38
rs1023533480 Chr.17: 46751449-46751449 on Build GRCh38
rs966039413 Chr.17: 46751683-46751683 on Build GRCh38
rs199533 Chr.17: 46751565-46751565 on Build GRCh38
rs371178494 Chr.17: 46751636-46751636 on Build GRCh38
rs756042382 Chr.17: 46751615-46751615 on Build GRCh38
rs754931750 Chr.17: 46751634-46751634 on Build GRCh38
rs369078079 Chr.17: 46751553-46751553 on Build GRCh38
rs562577633 Chr.17: 46751691-46751691 on Build GRCh38
rs910773981 Chr.17: 46751664-46751664 on Build GRCh38
rs376726891 Chr.17: 46751541-46751541 on Build GRCh38
rs1040327122 Chr.17: 46751630-46751630 on Build GRCh38
rs919257279 Chr.17: 46751801-46751801 on Build GRCh38
rs767702555 Chr.17: 46751604-46751604 on Build GRCh38
rs772797553 Chr.17: 46751642-46751642 on Build GRCh38
rs373537714 Chr.17: 46751533-46751533 on Build GRCh38
rs1004018330 Chr.17: 46751404-46751404 on Build GRCh38
rs375174991 Chr.17: 46751519-46751519 on Build GRCh38
rs747785435 Chr.17: 46751520-46751520 on Build GRCh38
rs549068756 Chr.17: 46751623-46751623 on Build GRCh38
rs373507225 Chr.17: 46751629-46751629 on Build GRCh38
rs768375765 Chr.17: 46751496-46751496 on Build GRCh38
rs201877473 Chr.17: 46751635-46751635 on Build GRCh38

View Full Product Details