Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 46978305-46978792 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RPRML
Entrez Gene ID
388394
Gene Name
reprimo like
Gene Aliases
-
UniGene
Hs.367999
Gene Chromosome Location
Chr.17: 46978156-46979248 on Build GRCh38
Transcripts
NM_203400.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3958461 c.342A>G p.A114A Chr.17: 46978666-46978666 on Build GRCh38
3989076 c.357G>A p.L119L Chr.17: 46978651-46978651 on Build GRCh38
6586963 c.313G>C p.E105Q Chr.17: 46978695-46978695 on Build GRCh38
6769381 c.218T>G p.L73R Chr.17: 46978790-46978790 on Build GRCh38
4414037 c.291C>A p.S97R Chr.17: 46978717-46978717 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs962588603 Chr.17: 46978523-46978523 on Build GRCh38
rs998807932 Chr.17: 46978348-46978348 on Build GRCh38
rs879308275 Chr.17: 46978590-46978590 on Build GRCh38
rs552811167 Chr.17: 46978417-46978417 on Build GRCh38
rs375855663 Chr.17: 46978531-46978531 on Build GRCh38
rs780102724 Chr.17: 46978626-46978626 on Build GRCh38
rs1015119300 Chr.17: 46978714-46978714 on Build GRCh38
rs1030419213 Chr.17: 46978462-46978462 on Build GRCh38
rs949233466 Chr.17: 46978563-46978563 on Build GRCh38
rs562652222 Chr.17: 46978722-46978722 on Build GRCh38
rs767948977 Chr.17: 46978713-46978713 on Build GRCh38
rs375223858 Chr.17: 46978396-46978397 on Build GRCh38
rs199695818 Chr.17: 46978381-46978382 on Build GRCh38
rs544387954 Chr.17: 46978352-46978352 on Build GRCh38
rs996403903 Chr.17: 46978458-46978458 on Build GRCh38
rs775015304 Chr.17: 46978608-46978608 on Build GRCh38
rs373305815 Chr.17: 46978674-46978674 on Build GRCh38
rs868025811 Chr.17: 46978725-46978725 on Build GRCh38
rs900181508 Chr.17: 46978585-46978585 on Build GRCh38
rs1010945760 Chr.17: 46978474-46978474 on Build GRCh38
rs770779916 Chr.17: 46978688-46978688 on Build GRCh38
rs539507911 Chr.17: 46978411-46978411 on Build GRCh38
rs746758980 Chr.17: 46978687-46978687 on Build GRCh38
rs561922402 Chr.17: 46978609-46978609 on Build GRCh38
rs953437672 Chr.17: 46978704-46978704 on Build GRCh38
rs1005446196 Chr.17: 46978691-46978691 on Build GRCh38
rs570772172 Chr.17: 46978521-46978522 on Build GRCh38
rs1033248167 Chr.17: 46978382-46978385 on Build GRCh38
rs769249005 Chr.17: 46978546-46978546 on Build GRCh38
rs754135542 Chr.17: 46978629-46978629 on Build GRCh38
rs764853972 Chr.17: 46978600-46978600 on Build GRCh38
rs776126784 Chr.17: 46978689-46978689 on Build GRCh38
rs756478608 Chr.17: 46978624-46978624 on Build GRCh38
rs7501608 Chr.17: 46978773-46978773 on Build GRCh38
rs758922953 Chr.17: 46978598-46978598 on Build GRCh38
rs754979145 Chr.17: 46978639-46978639 on Build GRCh38
rs1015422426 Chr.17: 46978486-46978486 on Build GRCh38
rs750733017 Chr.17: 46978620-46978620 on Build GRCh38
rs572850422 Chr.17: 46978480-46978480 on Build GRCh38
rs766804348 Chr.17: 46978734-46978734 on Build GRCh38
rs890755084 Chr.17: 46978668-46978668 on Build GRCh38
rs753999116 Chr.17: 46978747-46978747 on Build GRCh38
rs927290800 Chr.17: 46978528-46978528 on Build GRCh38
rs750962106 Chr.17: 46978729-46978729 on Build GRCh38
rs920752081 Chr.17: 46978536-46978536 on Build GRCh38
rs911914864 Chr.17: 46978518-46978518 on Build GRCh38
rs955055839 Chr.17: 46978465-46978465 on Build GRCh38
rs1028123827 Chr.17: 46978700-46978705 on Build GRCh38
rs762375220 Chr.17: 46978607-46978607 on Build GRCh38
rs113736567 Chr.17: 46978329-46978329 on Build GRCh38
rs769182844 Chr.17: 46978696-46978696 on Build GRCh38
rs754141248 Chr.17: 46978526-46978526 on Build GRCh38
rs747021696 Chr.17: 46978395-46978396 on Build GRCh38
rs1053414970 Chr.17: 46978326-46978326 on Build GRCh38
rs777857837 Chr.17: 46978677-46978677 on Build GRCh38
rs893059861 Chr.17: 46978678-46978678 on Build GRCh38
rs936448330 Chr.17: 46978580-46978580 on Build GRCh38
rs1053362225 Chr.17: 46978582-46978582 on Build GRCh38
rs959252146 Chr.17: 46978544-46978544 on Build GRCh38
rs767858770 Chr.17: 46978611-46978611 on Build GRCh38
rs150010793 Chr.17: 46978761-46978761 on Build GRCh38
rs745551561 Chr.17: 46978545-46978545 on Build GRCh38
rs917722275 Chr.17: 46978547-46978547 on Build GRCh38
rs3072527 Chr.17: 46978394-46978395 on Build GRCh38
rs779111700 Chr.17: 46978653-46978653 on Build GRCh38
rs572620058 Chr.17: 46978561-46978567 on Build GRCh38
rs902878988 Chr.17: 46978630-46978630 on Build GRCh38
rs1042262604 Chr.17: 46978572-46978572 on Build GRCh38
rs760982616 Chr.17: 46978730-46978730 on Build GRCh38
rs3048484 Chr.17: 46978401-46978402 on Build GRCh38
rs748081575 Chr.17: 46978673-46978673 on Build GRCh38
rs199624011 Chr.17: 46978642-46978642 on Build GRCh38
rs541851849 Chr.17: 46978604-46978604 on Build GRCh38
rs548940213 Chr.17: 46978393-46978394 on Build GRCh38
rs780647419 Chr.17: 46978525-46978525 on Build GRCh38
rs376342637 Chr.17: 46978750-46978750 on Build GRCh38
rs963516409 Chr.17: 46978509-46978509 on Build GRCh38
rs923673042 Chr.17: 46978577-46978577 on Build GRCh38
rs775039874 Chr.17: 46978708-46978708 on Build GRCh38
rs762467207 Chr.17: 46978710-46978710 on Build GRCh38

View Full Product Details