Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 48576601-48577098 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HOXB3
Entrez Gene ID
3213
Gene Name
homeobox B3
Gene Aliases
HOX2, HOX2G, Hox-2.7
UniGene
Hs.654560
Gene Chromosome Location
Chr.17: 48548870-48590369 on Build GRCh38
Transcripts
XM_005257277.3 XM_005257280.3 XM_017024558.1 XM_017024560.1 XM_017024561.1 XM_017024562.1 XM_017024563.1 NM_024015.4

Gene Symbol
HOXB4
Entrez Gene ID
3214
Gene Name
homeobox B4
Gene Aliases
HOX-2.6, HOX2, HOX2F
UniGene
Hs.664706
Gene Chromosome Location
Chr.17: 48575507-48578381 on Build GRCh38
Transcripts
XM_005257277.3 XM_005257280.3 XM_017024558.1 XM_017024560.1 XM_017024561.1 XM_017024562.1 XM_017024563.1 NM_024015.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4747156 c.722delC p.P241fs*>11 Chr.17: 48576756-48576756 on Build GRCh38
4799874 c.499A>T p.T167S Chr.17: 48576979-48576979 on Build GRCh38
472953 c.686G>A p.R229H Chr.17: 48576792-48576792 on Build GRCh38
6480129 c.621G>C p.K207N Chr.17: 48576857-48576857 on Build GRCh38
472954 c.672C>G p.P224P Chr.17: 48576806-48576806 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751397865 Chr.17: 48577049-48577049 on Build GRCh38
rs760942924 Chr.17: 48576756-48576756 on Build GRCh38
rs781490366 Chr.17: 48577009-48577010 on Build GRCh38
rs777502859 Chr.17: 48576791-48576791 on Build GRCh38
rs931949435 Chr.17: 48576814-48576814 on Build GRCh38
rs200388738 Chr.17: 48576779-48576779 on Build GRCh38
rs529290290 Chr.17: 48576679-48576679 on Build GRCh38
rs116058353 Chr.17: 48576930-48576930 on Build GRCh38
rs745422628 Chr.17: 48576958-48576958 on Build GRCh38
rs750126056 Chr.17: 48577066-48577066 on Build GRCh38
rs1034769921 Chr.17: 48576733-48576733 on Build GRCh38
rs762936719 Chr.17: 48577015-48577015 on Build GRCh38
rs776142392 Chr.17: 48576729-48576729 on Build GRCh38
rs752099638 Chr.17: 48576820-48576820 on Build GRCh38
rs767851206 Chr.17: 48576706-48576706 on Build GRCh38
rs749183012 Chr.17: 48576715-48576715 on Build GRCh38
rs201545381 Chr.17: 48576941-48576941 on Build GRCh38
rs975036413 Chr.17: 48576881-48576881 on Build GRCh38
rs769086823 Chr.17: 48576825-48576825 on Build GRCh38
rs754884797 Chr.17: 48576716-48576716 on Build GRCh38
rs752710819 Chr.17: 48576906-48576906 on Build GRCh38
rs888075993 Chr.17: 48576701-48576701 on Build GRCh38
rs764371963 Chr.17: 48576686-48576686 on Build GRCh38
rs753938234 Chr.17: 48576760-48576760 on Build GRCh38
rs757026315 Chr.17: 48577050-48577050 on Build GRCh38
rs2740755 Chr.17: 48576637-48576637 on Build GRCh38
rs772689748 Chr.17: 48576971-48576971 on Build GRCh38
rs766578016 Chr.17: 48576758-48576758 on Build GRCh38
rs772599680 Chr.17: 48576867-48576867 on Build GRCh38
rs777558289 Chr.17: 48576912-48576912 on Build GRCh38
rs372443829 Chr.17: 48577029-48577029 on Build GRCh38
rs138753340 Chr.17: 48576896-48576896 on Build GRCh38
rs750575261 Chr.17: 48576708-48576708 on Build GRCh38
rs764016180 Chr.17: 48577042-48577042 on Build GRCh38
rs940017798 Chr.17: 48576662-48576662 on Build GRCh38
rs956781625 Chr.17: 48576745-48576745 on Build GRCh38
rs774903494 Chr.17: 48576860-48576860 on Build GRCh38
rs779395632 Chr.17: 48576965-48576965 on Build GRCh38
rs549541586 Chr.17: 48576687-48576687 on Build GRCh38
rs765179311 Chr.17: 48576904-48576904 on Build GRCh38
rs773774380 Chr.17: 48576977-48576977 on Build GRCh38
rs1051276456 Chr.17: 48576926-48576926 on Build GRCh38
rs748672425 Chr.17: 48576967-48576967 on Build GRCh38
rs769192702 Chr.17: 48576744-48576744 on Build GRCh38
rs753093681 Chr.17: 48577060-48577064 on Build GRCh38
rs3744776 Chr.17: 48577068-48577068 on Build GRCh38
rs756242347 Chr.17: 48576711-48576711 on Build GRCh38
rs368788191 Chr.17: 48576802-48576802 on Build GRCh38
rs771104213 Chr.17: 48577000-48577000 on Build GRCh38
rs758264665 Chr.17: 48576910-48576910 on Build GRCh38
rs993902571 Chr.17: 48576629-48576629 on Build GRCh38
rs747343741 Chr.17: 48576993-48576993 on Build GRCh38
rs977293314 Chr.17: 48576817-48576817 on Build GRCh38
rs767904414 Chr.17: 48576751-48576751 on Build GRCh38
rs757480194 Chr.17: 48576691-48576691 on Build GRCh38
rs569250918 Chr.17: 48576688-48576688 on Build GRCh38
rs79154089 Chr.17: 48577073-48577073 on Build GRCh38
rs138198807 Chr.17: 48577011-48577011 on Build GRCh38
rs775491961 Chr.17: 48577012-48577012 on Build GRCh38
rs753901753 Chr.17: 48576884-48576884 on Build GRCh38
rs146993812 Chr.17: 48577006-48577006 on Build GRCh38
rs552830566 Chr.17: 48576797-48576797 on Build GRCh38
rs766702664 Chr.17: 48576877-48576877 on Build GRCh38
rs746670209 Chr.17: 48576793-48576793 on Build GRCh38
rs202211147 Chr.17: 48576731-48576731 on Build GRCh38
rs897983702 Chr.17: 48576915-48576915 on Build GRCh38
rs769370544 Chr.17: 48576962-48576962 on Build GRCh38
rs1003454710 Chr.17: 48577055-48577055 on Build GRCh38
rs770457825 Chr.17: 48576725-48576725 on Build GRCh38
rs778874678 Chr.17: 48576774-48576774 on Build GRCh38
rs756972884 Chr.17: 48576920-48576920 on Build GRCh38
rs537257453 Chr.17: 48576719-48576719 on Build GRCh38
rs143090260 Chr.17: 48576795-48576795 on Build GRCh38
rs989281275 Chr.17: 48577075-48577075 on Build GRCh38
rs754933999 Chr.17: 48576767-48576767 on Build GRCh38
rs570743872 Chr.17: 48576736-48576736 on Build GRCh38
rs899388207 Chr.17: 48577030-48577030 on Build GRCh38
rs758124264 Chr.17: 48577004-48577004 on Build GRCh38
rs776932671 Chr.17: 48577002-48577002 on Build GRCh38
rs751342746 Chr.17: 48576913-48576913 on Build GRCh38
rs761031489 Chr.17: 48576872-48576872 on Build GRCh38
rs149049923 Chr.17: 48576750-48576750 on Build GRCh38
rs370288251 Chr.17: 48577067-48577067 on Build GRCh38
rs755477319 Chr.17: 48576833-48576834 on Build GRCh38
rs767170863 Chr.17: 48577060-48577060 on Build GRCh38
rs772827973 Chr.17: 48576721-48576721 on Build GRCh38
rs764708289 Chr.17: 48576713-48576714 on Build GRCh38
rs773651269 Chr.17: 48576869-48576869 on Build GRCh38
rs748798994 Chr.17: 48577072-48577072 on Build GRCh38
rs376222523 Chr.17: 48576724-48576724 on Build GRCh38
rs780236068 Chr.17: 48576714-48576714 on Build GRCh38
rs188952344 Chr.17: 48576950-48576950 on Build GRCh38
rs752483339 Chr.17: 48576776-48576776 on Build GRCh38
rs925852719 Chr.17: 48576841-48576841 on Build GRCh38

View Full Product Details