Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 48607935-48608483 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HOXB7
Entrez Gene ID
3217
Gene Name
homeobox B7
Gene Aliases
HHO.C1, HOX2, HOX2C, Hox-2.3
UniGene
Hs.436181
Gene Chromosome Location
Chr.17: 48607232-48611021 on Build GRCh38
Transcripts
NM_004502.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4930121 c.416G>A p.R139Q Chr.17: 48608080-48608080 on Build GRCh38
707059 c.489G>T p.T163T Chr.17: 48608007-48608007 on Build GRCh38
4881312 c.511G>T p.A171S Chr.17: 48607985-48607985 on Build GRCh38
707058 c.436C>T p.R146C Chr.17: 48608060-48608060 on Build GRCh38
6681717 c.405T>C p.T135T Chr.17: 48608091-48608091 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779768655 Chr.17: 48608116-48608116 on Build GRCh38
rs56158178 Chr.17: 48608285-48608285 on Build GRCh38
rs369320098 Chr.17: 48608379-48608379 on Build GRCh38
rs532956213 Chr.17: 48608388-48608388 on Build GRCh38
rs557880912 Chr.17: 48608190-48608190 on Build GRCh38
rs1037229387 Chr.17: 48608200-48608200 on Build GRCh38
rs948261250 Chr.17: 48608437-48608437 on Build GRCh38
rs201728872 Chr.17: 48608449-48608449 on Build GRCh38
rs775327106 Chr.17: 48608045-48608045 on Build GRCh38
rs767002458 Chr.17: 48607997-48607997 on Build GRCh38
rs770931876 Chr.17: 48608018-48608018 on Build GRCh38
rs201477004 Chr.17: 48608107-48608107 on Build GRCh38
rs35411494 Chr.17: 48608447-48608447 on Build GRCh38
rs756739078 Chr.17: 48607995-48607995 on Build GRCh38
rs754330441 Chr.17: 48607999-48607999 on Build GRCh38
rs753094787 Chr.17: 48608083-48608083 on Build GRCh38
rs768494905 Chr.17: 48608134-48608134 on Build GRCh38
rs774086175 Chr.17: 48608059-48608059 on Build GRCh38
rs1010065591 Chr.17: 48608240-48608240 on Build GRCh38
rs747017326 Chr.17: 48607964-48607964 on Build GRCh38
rs71144507 Chr.17: 48608447-48608448 on Build GRCh38
rs765558409 Chr.17: 48608081-48608081 on Build GRCh38
rs764769851 Chr.17: 48608027-48608027 on Build GRCh38
rs781229849 Chr.17: 48608102-48608102 on Build GRCh38
rs931619852 Chr.17: 48608248-48608248 on Build GRCh38
rs747284350 Chr.17: 48608236-48608237 on Build GRCh38
rs553301781 Chr.17: 48608154-48608154 on Build GRCh38
rs749008302 Chr.17: 48607996-48607998 on Build GRCh38
rs763822704 Chr.17: 48608049-48608049 on Build GRCh38
rs765046532 Chr.17: 48607972-48607972 on Build GRCh38
rs1018382389 Chr.17: 48608344-48608345 on Build GRCh38
rs533071036 Chr.17: 48608005-48608005 on Build GRCh38
rs62637586 Chr.17: 48607986-48607986 on Build GRCh38
rs747192791 Chr.17: 48608090-48608090 on Build GRCh38
rs747106896 Chr.17: 48608016-48608016 on Build GRCh38
rs745772585 Chr.17: 48608023-48608023 on Build GRCh38
rs763554332 Chr.17: 48607985-48607985 on Build GRCh38
rs762648399 Chr.17: 48607984-48607984 on Build GRCh38
rs1012338358 Chr.17: 48608024-48608024 on Build GRCh38
rs532933268 Chr.17: 48608222-48608222 on Build GRCh38
rs779286425 Chr.17: 48608003-48608003 on Build GRCh38
rs868398142 Chr.17: 48608317-48608317 on Build GRCh38
rs906643536 Chr.17: 48608311-48608311 on Build GRCh38
rs751434541 Chr.17: 48608340-48608340 on Build GRCh38
rs765789072 Chr.17: 48608143-48608143 on Build GRCh38
rs908953850 Chr.17: 48608148-48608148 on Build GRCh38
rs777889579 Chr.17: 48608012-48608012 on Build GRCh38
rs1028581898 Chr.17: 48608405-48608405 on Build GRCh38
rs377132219 Chr.17: 48607962-48607962 on Build GRCh38
rs984292311 Chr.17: 48608407-48608407 on Build GRCh38
rs775030826 Chr.17: 48607974-48607974 on Build GRCh38
rs918848740 Chr.17: 48608105-48608105 on Build GRCh38
rs1021908211 Chr.17: 48608247-48608247 on Build GRCh38
rs754323715 Chr.17: 48608074-48608074 on Build GRCh38
rs910063195 Chr.17: 48608436-48608437 on Build GRCh38
rs372811667 Chr.17: 48608144-48608144 on Build GRCh38
rs769768136 Chr.17: 48608115-48608115 on Build GRCh38
rs772745522 Chr.17: 48608139-48608139 on Build GRCh38
rs758643585 Chr.17: 48608010-48608010 on Build GRCh38
rs1021711604 Chr.17: 48607996-48607996 on Build GRCh38
rs897390701 Chr.17: 48608216-48608216 on Build GRCh38
rs374080435 Chr.17: 48608198-48608198 on Build GRCh38
rs200174441 Chr.17: 48608087-48608087 on Build GRCh38
rs541643262 Chr.17: 48608301-48608301 on Build GRCh38
rs761408048 Chr.17: 48608060-48608060 on Build GRCh38
rs770843856 Chr.17: 48607965-48607965 on Build GRCh38
rs369413024 Chr.17: 48608138-48608138 on Build GRCh38
rs776585340 Chr.17: 48608021-48608021 on Build GRCh38
rs370772095 Chr.17: 48608307-48608307 on Build GRCh38
rs541181851 Chr.17: 48608100-48608100 on Build GRCh38
rs369415843 Chr.17: 48608048-48608048 on Build GRCh38
rs1042216 Chr.17: 48607979-48607979 on Build GRCh38
rs755486920 Chr.17: 48608075-48608075 on Build GRCh38
rs1044021114 Chr.17: 48608172-48608172 on Build GRCh38
rs931694212 Chr.17: 48608164-48608164 on Build GRCh38
rs200454962 Chr.17: 48608086-48608086 on Build GRCh38
rs897428031 Chr.17: 48608007-48608007 on Build GRCh38
rs958193486 Chr.17: 48608372-48608372 on Build GRCh38
rs375129723 Chr.17: 48608447-48608447 on Build GRCh38
rs201345179 Chr.17: 48608137-48608137 on Build GRCh38
rs564074643 Chr.17: 48608381-48608381 on Build GRCh38
rs534029306 Chr.17: 48608251-48608251 on Build GRCh38
rs1030870090 Chr.17: 48608349-48608349 on Build GRCh38
rs377413674 Chr.17: 48608199-48608199 on Build GRCh38
rs553845261 Chr.17: 48608273-48608273 on Build GRCh38
rs755328654 Chr.17: 48608002-48608002 on Build GRCh38
rs138865004 Chr.17: 48607966-48607966 on Build GRCh38
rs760021518 Chr.17: 48608141-48608141 on Build GRCh38
rs953067660 Chr.17: 48608436-48608436 on Build GRCh38
rs551277185 Chr.17: 48608337-48608337 on Build GRCh38
rs544454282 Chr.17: 48608380-48608380 on Build GRCh38
rs201959822 Chr.17: 48608135-48608135 on Build GRCh38
rs1048384425 Chr.17: 48608250-48608250 on Build GRCh38
rs749117591 Chr.17: 48608122-48608122 on Build GRCh38
rs912706717 Chr.17: 48608194-48608194 on Build GRCh38
rs575464884 Chr.17: 48608378-48608378 on Build GRCh38
rs745837009 Chr.17: 48608103-48608103 on Build GRCh38
rs183965234 Chr.17: 48608146-48608146 on Build GRCh38
rs769530061 Chr.17: 48608043-48608043 on Build GRCh38
rs1006954471 Chr.17: 48608327-48608327 on Build GRCh38
rs1027019817 Chr.17: 48608017-48608017 on Build GRCh38
rs979788937 Chr.17: 48608446-48608446 on Build GRCh38

View Full Product Details