Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 49411588-49412087 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PHB
Entrez Gene ID
5245
Gene Name
prohibitin
Gene Aliases
HEL-215, HEL-S-54e, PHB1
UniGene
Hs.514303
Gene Chromosome Location
Chr.17: 49404048-49414905 on Build GRCh38
Transcripts
NM_001281496.1 NM_001281497.1 NM_001281715.1 NM_002634.3 XM_017024762.1 XM_017024763.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6338242 c.110T>C p.V37A Chr.17: 49411818-49411818 on Build GRCh38
5817887 c.89-4C>G p.? Chr.17: 49411843-49411843 on Build GRCh38
6389294 c.229C>T p.P77S Chr.17: 49411699-49411699 on Build GRCh38
5852860 c.208C>T p.R70C Chr.17: 49411720-49411720 on Build GRCh38
400184 c.228G>T p.V76V Chr.17: 49411700-49411700 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746550219 Chr.17: 49411868-49411868 on Build GRCh38
rs747919114 Chr.17: 49411748-49411748 on Build GRCh38
rs1002620919 Chr.17: 49411953-49411956 on Build GRCh38
rs938193533 Chr.17: 49411633-49411633 on Build GRCh38
rs778272458 Chr.17: 49411713-49411713 on Build GRCh38
rs559011871 Chr.17: 49411960-49411960 on Build GRCh38
rs536400006 Chr.17: 49411757-49411757 on Build GRCh38
rs1021272457 Chr.17: 49412030-49412030 on Build GRCh38
rs769816421 Chr.17: 49411752-49411752 on Build GRCh38
rs968764517 Chr.17: 49412032-49412032 on Build GRCh38
rs370766193 Chr.17: 49411746-49411746 on Build GRCh38
rs763158814 Chr.17: 49411649-49411649 on Build GRCh38
rs575435645 Chr.17: 49412037-49412037 on Build GRCh38
rs766645565 Chr.17: 49411653-49411653 on Build GRCh38
rs988994207 Chr.17: 49412016-49412016 on Build GRCh38
rs930087263 Chr.17: 49411785-49411785 on Build GRCh38
rs757863990 Chr.17: 49411787-49411787 on Build GRCh38
rs767677971 Chr.17: 49411801-49411801 on Build GRCh38
rs748212379 Chr.17: 49411726-49411726 on Build GRCh38
rs905798657 Chr.17: 49411950-49411950 on Build GRCh38
rs745340447 Chr.17: 49411719-49411719 on Build GRCh38
rs955806195 Chr.17: 49411980-49411980 on Build GRCh38
rs753039512 Chr.17: 49411697-49411697 on Build GRCh38
rs781612334 Chr.17: 49411689-49411689 on Build GRCh38
rs367962991 Chr.17: 49411710-49411710 on Build GRCh38
rs370837005 Chr.17: 49411635-49411635 on Build GRCh38
rs200614404 Chr.17: 49411654-49411654 on Build GRCh38
rs764517263 Chr.17: 49411829-49411829 on Build GRCh38
rs751202413 Chr.17: 49411951-49411951 on Build GRCh38
rs779623438 Chr.17: 49411723-49411723 on Build GRCh38
rs375867420 Chr.17: 49411664-49411664 on Build GRCh38
rs992313146 Chr.17: 49412040-49412040 on Build GRCh38
rs768234153 Chr.17: 49411745-49411745 on Build GRCh38
rs754611263 Chr.17: 49411875-49411875 on Build GRCh38
rs753054107 Chr.17: 49411806-49411806 on Build GRCh38
rs1004147217 Chr.17: 49411895-49411895 on Build GRCh38
rs974111403 Chr.17: 49411947-49411947 on Build GRCh38
rs201359263 Chr.17: 49411776-49411776 on Build GRCh38
rs763072596 Chr.17: 49411767-49411767 on Build GRCh38
rs2233664 Chr.17: 49411881-49411881 on Build GRCh38
rs192900341 Chr.17: 49411857-49411857 on Build GRCh38
rs754355612 Chr.17: 49411835-49411835 on Build GRCh38
rs962686632 Chr.17: 49411919-49411919 on Build GRCh38
rs945635098 Chr.17: 49411714-49411714 on Build GRCh38
rs572371549 Chr.17: 49412044-49412044 on Build GRCh38
rs2233665 Chr.17: 49411800-49411800 on Build GRCh38
rs926719576 Chr.17: 49411610-49411610 on Build GRCh38
rs756499521 Chr.17: 49411826-49411826 on Build GRCh38
rs757906969 Chr.17: 49411720-49411720 on Build GRCh38
rs766557694 Chr.17: 49411789-49411789 on Build GRCh38
rs757819160 Chr.17: 49411844-49411844 on Build GRCh38
rs888470045 Chr.17: 49411840-49411840 on Build GRCh38
rs944311231 Chr.17: 49412060-49412060 on Build GRCh38
rs147481037 Chr.17: 49411799-49411799 on Build GRCh38
rs779585659 Chr.17: 49411855-49411855 on Build GRCh38
rs1035524015 Chr.17: 49411972-49411972 on Build GRCh38
rs746659527 Chr.17: 49411743-49411743 on Build GRCh38
rs544942841 Chr.17: 49411903-49411903 on Build GRCh38
rs1022810829 Chr.17: 49411948-49411948 on Build GRCh38
rs935691813 Chr.17: 49411769-49411769 on Build GRCh38

View Full Product Details