Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 49581044-49581543 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NXPH3
Entrez Gene ID
11248
Gene Name
neurexophilin 3
Gene Aliases
NPH3
UniGene
Hs.55069
Gene Chromosome Location
Chr.17: 49575936-49583810 on Build GRCh38
Transcripts
NM_007225.2

SNP Detail

SNP ID Chromosome Location
rs772700758 Chr.17: 49581140-49581140 on Build GRCh38
rs550013446 Chr.17: 49581212-49581212 on Build GRCh38
rs375788506 Chr.17: 49581453-49581453 on Build GRCh38
rs983008976 Chr.17: 49581303-49581303 on Build GRCh38
rs974260511 Chr.17: 49581498-49581498 on Build GRCh38
rs549887942 Chr.17: 49581514-49581514 on Build GRCh38
rs1026212266 Chr.17: 49581499-49581499 on Build GRCh38
rs569782986 Chr.17: 49581348-49581348 on Build GRCh38
rs1033011269 Chr.17: 49581236-49581236 on Build GRCh38
rs112907538 Chr.17: 49581391-49581391 on Build GRCh38
rs748910533 Chr.17: 49581448-49581448 on Build GRCh38
rs904811987 Chr.17: 49581227-49581227 on Build GRCh38
rs1009876398 Chr.17: 49581181-49581181 on Build GRCh38
rs955471793 Chr.17: 49581468-49581468 on Build GRCh38
rs940332297 Chr.17: 49581101-49581101 on Build GRCh38
rs955705019 Chr.17: 49581301-49581301 on Build GRCh38
rs951476380 Chr.17: 49581350-49581351 on Build GRCh38
rs773559901 Chr.17: 49581220-49581220 on Build GRCh38
rs530464107 Chr.17: 49581246-49581246 on Build GRCh38
rs908827225 Chr.17: 49581329-49581329 on Build GRCh38
rs1000222754 Chr.17: 49581233-49581233 on Build GRCh38
rs759187851 Chr.17: 49581426-49581426 on Build GRCh38
rs962964416 Chr.17: 49581475-49581475 on Build GRCh38
rs980014731 Chr.17: 49581398-49581398 on Build GRCh38
rs1014173173 Chr.17: 49581176-49581176 on Build GRCh38
rs937113795 Chr.17: 49581418-49581418 on Build GRCh38
rs760280402 Chr.17: 49581326-49581326 on Build GRCh38
rs1055911055 Chr.17: 49581425-49581425 on Build GRCh38
rs879187316 Chr.17: 49581334-49581334 on Build GRCh38
rs1025697499 Chr.17: 49581330-49581330 on Build GRCh38
rs1029833562 Chr.17: 49581266-49581266 on Build GRCh38
rs183005842 Chr.17: 49581437-49581437 on Build GRCh38
rs1037315677 Chr.17: 49581104-49581104 on Build GRCh38
rs1018436171 Chr.17: 49581274-49581274 on Build GRCh38
rs906446017 Chr.17: 49581351-49581369 on Build GRCh38
rs375493711 Chr.17: 49581323-49581323 on Build GRCh38
rs561700324 Chr.17: 49581178-49581178 on Build GRCh38
rs1042763086 Chr.17: 49581446-49581446 on Build GRCh38
rs1001688734 Chr.17: 49581123-49581123 on Build GRCh38
rs113854498 Chr.17: 49581392-49581392 on Build GRCh38
rs1015711125 Chr.17: 49581474-49581474 on Build GRCh38
rs920274877 Chr.17: 49581500-49581500 on Build GRCh38
rs975870068 Chr.17: 49581258-49581258 on Build GRCh38
rs529964874 Chr.17: 49581190-49581190 on Build GRCh38
rs920246301 Chr.17: 49581400-49581400 on Build GRCh38
rs927106154 Chr.17: 49581118-49581118 on Build GRCh38
rs541828458 Chr.17: 49581095-49581095 on Build GRCh38
rs903142764 Chr.17: 49581443-49581443 on Build GRCh38
rs909105220 Chr.17: 49581081-49581081 on Build GRCh38
rs982935538 Chr.17: 49581472-49581472 on Build GRCh38
rs762655126 Chr.17: 49581463-49581463 on Build GRCh38
rs532333242 Chr.17: 49581370-49581370 on Build GRCh38
rs528116517 Chr.17: 49581449-49581449 on Build GRCh38
rs568017596 Chr.17: 49581442-49581442 on Build GRCh38
rs895603150 Chr.17: 49581155-49581155 on Build GRCh38
rs936977613 Chr.17: 49581505-49581505 on Build GRCh38
rs537380394 Chr.17: 49581447-49581447 on Build GRCh38
rs191162023 Chr.17: 49581414-49581414 on Build GRCh38
rs12452102 Chr.17: 49581172-49581172 on Build GRCh38
rs964196952 Chr.17: 49581238-49581238 on Build GRCh38
rs898937106 Chr.17: 49581106-49581106 on Build GRCh38
rs548610380 Chr.17: 49581355-49581355 on Build GRCh38

View Full Product Details