Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 50560939-50561477 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CACNA1G
Entrez Gene ID
8913
Gene Name
calcium voltage-gated channel subunit alpha1 G
Gene Aliases
Ca(V)T.1, Cav3.1, NBR13, SCA42
UniGene
Hs.591169
Gene Chromosome Location
Chr.17: 50560929-50629625 on Build GRCh38
Transcripts
NM_001256324.1 NM_001256325.1 NM_001256326.1 NM_001256327.1 NM_001256328.1 NM_001256329.1 NM_001256330.1 NM_001256331.1 NM_001256332.1 NM_001256333.1 NM_001256334.1 NM_001256359.1 NM_001256360.1 NM_001256361.1 NM_018896.4 NM_198376.2 NM_198377.2 NM_198378.2 NM_198379.2 NM_198380.2 NM_198382.2 NM_198383.2 NM_198384.2 NM_198385.2 NM_198386.2 NM_198387.2 NM_198388.2 NM_198396.2 NR_046054.1 NR_046055.1 NR_046056.1 NR_046057.1 NR_046058.1 XM_006722160.3 XM_006722161.3 NR_038439.1

Gene Symbol
CACNA1G-AS1
Entrez Gene ID
253962
Gene Name
CACNA1G antisense RNA 1
Gene Aliases
-
UniGene
Hs.558704
Gene Chromosome Location
Chr.17: 50556207-50562108 on Build GRCh38
Transcripts
NM_001256324.1 NM_001256325.1 NM_001256326.1 NM_001256327.1 NM_001256328.1 NM_001256329.1 NM_001256330.1 NM_001256331.1 NM_001256332.1 NM_001256333.1 NM_001256334.1 NM_001256359.1 NM_001256360.1 NM_001256361.1 NM_018896.4 NM_198376.2 NM_198377.2 NM_198378.2 NM_198379.2 NM_198380.2 NM_198382.2 NM_198383.2 NM_198384.2 NM_198385.2 NM_198386.2 NM_198387.2 NM_198388.2 NM_198396.2 NR_046054.1 NR_046055.1 NR_046056.1 NR_046057.1 NR_046058.1 XM_006722160.3 XM_006722161.3 NR_038439.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4617789 c.9G>C p.E3D Chr.17: 50561468-50561468 on Build GRCh38
4617787 c.9G>C p.E3D Chr.17: 50561468-50561468 on Build GRCh38
4634221 c.12G>T p.E4D Chr.17: 50561471-50561471 on Build GRCh38
4634222 c.12G>T p.E4D Chr.17: 50561471-50561471 on Build GRCh38
4617788 c.9G>C p.E3D Chr.17: 50561468-50561468 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775780381 Chr.17: 50561212-50561212 on Build GRCh38
rs746019689 Chr.17: 50561090-50561090 on Build GRCh38
rs912740921 Chr.17: 50561037-50561037 on Build GRCh38
rs777736883 Chr.17: 50561023-50561023 on Build GRCh38
rs1049960795 Chr.17: 50561011-50561011 on Build GRCh38
rs915797423 Chr.17: 50561084-50561084 on Build GRCh38
rs527555654 Chr.17: 50561041-50561041 on Build GRCh38
rs931307983 Chr.17: 50561065-50561065 on Build GRCh38
rs551065651 Chr.17: 50560967-50560967 on Build GRCh38
rs763864006 Chr.17: 50561317-50561317 on Build GRCh38
rs571252731 Chr.17: 50560988-50560988 on Build GRCh38
rs892929231 Chr.17: 50561143-50561143 on Build GRCh38
rs919894706 Chr.17: 50561055-50561055 on Build GRCh38
rs1045945766 Chr.17: 50561095-50561095 on Build GRCh38
rs1023469747 Chr.17: 50561094-50561094 on Build GRCh38
rs556446615 Chr.17: 50561254-50561254 on Build GRCh38
rs1003352743 Chr.17: 50561448-50561448 on Build GRCh38
rs920011126 Chr.17: 50561328-50561328 on Build GRCh38
rs985994495 Chr.17: 50561226-50561226 on Build GRCh38
rs772452335 Chr.17: 50561109-50561109 on Build GRCh38
rs764578323 Chr.17: 50561319-50561319 on Build GRCh38
rs987359196 Chr.17: 50561007-50561007 on Build GRCh38
rs569354413 Chr.17: 50561270-50561270 on Build GRCh38
rs953196832 Chr.17: 50561220-50561220 on Build GRCh38
rs1008495516 Chr.17: 50561280-50561280 on Build GRCh38
rs777184186 Chr.17: 50561342-50561342 on Build GRCh38
rs906392544 Chr.17: 50561443-50561443 on Build GRCh38
rs888488120 Chr.17: 50561377-50561377 on Build GRCh38
rs1045271329 Chr.17: 50561439-50561439 on Build GRCh38
rs977020611 Chr.17: 50561242-50561242 on Build GRCh38
rs547688355 Chr.17: 50561087-50561087 on Build GRCh38
rs1055090902 Chr.17: 50561079-50561079 on Build GRCh38
rs956353528 Chr.17: 50561158-50561158 on Build GRCh38
rs1030599036 Chr.17: 50561145-50561145 on Build GRCh38
rs944275188 Chr.17: 50561034-50561034 on Build GRCh38
rs948622285 Chr.17: 50561429-50561429 on Build GRCh38
rs929952497 Chr.17: 50561284-50561284 on Build GRCh38
rs989206166 Chr.17: 50560961-50560961 on Build GRCh38
rs948168803 Chr.17: 50561412-50561412 on Build GRCh38
rs972793029 Chr.17: 50561049-50561049 on Build GRCh38
rs761001427 Chr.17: 50561229-50561229 on Build GRCh38
rs907054784 Chr.17: 50561106-50561106 on Build GRCh38
rs916598276 Chr.17: 50561409-50561409 on Build GRCh38
rs981384668 Chr.17: 50561440-50561440 on Build GRCh38
rs1048428374 Chr.17: 50561366-50561366 on Build GRCh38
rs757285743 Chr.17: 50561140-50561140 on Build GRCh38
rs538372568 Chr.17: 50561389-50561389 on Build GRCh38
rs77330952 Chr.17: 50560984-50560984 on Build GRCh38
rs112692320 Chr.17: 50561288-50561288 on Build GRCh38
rs954343331 Chr.17: 50561003-50561003 on Build GRCh38
rs536443718 Chr.17: 50561237-50561237 on Build GRCh38
rs954409365 Chr.17: 50561271-50561271 on Build GRCh38
rs944243903 Chr.17: 50561240-50561240 on Build GRCh38
rs1011228453 Chr.17: 50561186-50561186 on Build GRCh38
rs769671608 Chr.17: 50561022-50561022 on Build GRCh38
rs961734925 Chr.17: 50561039-50561039 on Build GRCh38
rs1052846717 Chr.17: 50561132-50561132 on Build GRCh38
rs952824648 Chr.17: 50561332-50561332 on Build GRCh38
rs972911903 Chr.17: 50561327-50561327 on Build GRCh38
rs749394192 Chr.17: 50561066-50561066 on Build GRCh38
rs567430839 Chr.17: 50561111-50561111 on Build GRCh38
rs1028576630 Chr.17: 50560989-50560989 on Build GRCh38
rs961574055 Chr.17: 50561323-50561323 on Build GRCh38
rs1028020379 Chr.17: 50561223-50561223 on Build GRCh38
rs989137541 Chr.17: 50561185-50561185 on Build GRCh38

View Full Product Details