Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 50744243-50744790 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LUC7L3
Entrez Gene ID
51747
Gene Name
LUC7 like 3 pre-mRNA splicing factor
Gene Aliases
CRA, CREAP-1, CROP, LUC7A, OA48-18, hLuc7A
UniGene
Hs.130293
Gene Chromosome Location
Chr.17: 50719565-50754269 on Build GRCh38
Transcripts
NM_006107.3 NM_016424.4 XM_005257448.1 XM_005257449.2 XM_005257452.1 XM_006721943.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4067778 c.395T>C p.V132A Chr.17: 50744743-50744743 on Build GRCh38
4802997 c.368T>C p.L123S Chr.17: 50744716-50744716 on Build GRCh38
6705162 c.425T>C p.M142T Chr.17: 50744773-50744773 on Build GRCh38
1324967 c.607G>C p.D203H Chr.17: 50744727-50744727 on Build GRCh38
981311 c.544T>A p.F182I Chr.17: 50744664-50744664 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748963617 Chr.17: 50744320-50744320 on Build GRCh38
rs1032531086 Chr.17: 50744341-50744341 on Build GRCh38
rs764129310 Chr.17: 50744609-50744609 on Build GRCh38
rs36105689 Chr.17: 50744739-50744740 on Build GRCh38
rs750774062 Chr.17: 50744768-50744768 on Build GRCh38
rs778401010 Chr.17: 50744423-50744423 on Build GRCh38
rs754429072 Chr.17: 50744409-50744409 on Build GRCh38
rs368517414 Chr.17: 50744545-50744545 on Build GRCh38
rs866193717 Chr.17: 50744731-50744731 on Build GRCh38
rs753675119 Chr.17: 50744613-50744615 on Build GRCh38
rs192593481 Chr.17: 50744410-50744410 on Build GRCh38
rs919760989 Chr.17: 50744550-50744550 on Build GRCh38
rs775753035 Chr.17: 50744619-50744619 on Build GRCh38
rs931113456 Chr.17: 50744552-50744552 on Build GRCh38
rs765657404 Chr.17: 50744756-50744756 on Build GRCh38
rs762029991 Chr.17: 50744715-50744715 on Build GRCh38
rs776732997 Chr.17: 50744670-50744670 on Build GRCh38
rs1046507099 Chr.17: 50744540-50744540 on Build GRCh38
rs764183348 Chr.17: 50744666-50744666 on Build GRCh38
rs202223403 Chr.17: 50744447-50744448 on Build GRCh38
rs949398247 Chr.17: 50744537-50744537 on Build GRCh38
rs958154125 Chr.17: 50744408-50744408 on Build GRCh38
rs999406430 Chr.17: 50744573-50744573 on Build GRCh38
rs757171738 Chr.17: 50744676-50744677 on Build GRCh38
rs8070210 Chr.17: 50744629-50744629 on Build GRCh38
rs902588760 Chr.17: 50744563-50744563 on Build GRCh38
rs1028627216 Chr.17: 50744304-50744307 on Build GRCh38
rs773373597 Chr.17: 50744531-50744531 on Build GRCh38
rs907853444 Chr.17: 50744530-50744530 on Build GRCh38
rs1025045951 Chr.17: 50744305-50744305 on Build GRCh38
rs189246750 Chr.17: 50744532-50744532 on Build GRCh38
rs561040837 Chr.17: 50744270-50744270 on Build GRCh38
rs894920901 Chr.17: 50744631-50744631 on Build GRCh38
rs755690340 Chr.17: 50744283-50744283 on Build GRCh38
rs141810780 Chr.17: 50744672-50744672 on Build GRCh38
rs867407378 Chr.17: 50744516-50744516 on Build GRCh38
rs760699029 Chr.17: 50744647-50744647 on Build GRCh38
rs201002018 Chr.17: 50744744-50744744 on Build GRCh38
rs535442345 Chr.17: 50744462-50744462 on Build GRCh38
rs867005119 Chr.17: 50744361-50744361 on Build GRCh38
rs973303103 Chr.17: 50744431-50744431 on Build GRCh38
rs760469204 Chr.17: 50744602-50744602 on Build GRCh38
rs779640033 Chr.17: 50744290-50744290 on Build GRCh38
rs548037944 Chr.17: 50744557-50744557 on Build GRCh38
rs529853897 Chr.17: 50744326-50744326 on Build GRCh38
rs551786077 Chr.17: 50744518-50744518 on Build GRCh38
rs184778856 Chr.17: 50744460-50744460 on Build GRCh38
rs371577003 Chr.17: 50744572-50744572 on Build GRCh38
rs201795356 Chr.17: 50744620-50744620 on Build GRCh38
rs549710687 Chr.17: 50744353-50744353 on Build GRCh38
rs972598505 Chr.17: 50744546-50744546 on Build GRCh38

View Full Product Details