Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 58270472-58270982 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MPO
Entrez Gene ID
4353
Gene Name
myeloperoxidase
Gene Aliases
-
UniGene
Hs.458272
Gene Chromosome Location
Chr.17: 58269856-58280935 on Build GRCh38
Transcripts
NM_000250.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1710522 c.2041G>C p.E681Q Chr.17: 58270853-58270853 on Build GRCh38
436982 c.2174G>A p.R725Q Chr.17: 58270720-58270720 on Build GRCh38
7377479 c.2142C>A p.N714K Chr.17: 58270848-58270848 on Build GRCh38
706701 c.2127C>T p.I709I Chr.17: 58270767-58270767 on Build GRCh38
4068112 c.2152T>C p.F718L Chr.17: 58270838-58270838 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750182650 Chr.17: 58270768-58270768 on Build GRCh38
rs144557565 Chr.17: 58270792-58270792 on Build GRCh38
rs1011844354 Chr.17: 58270722-58270722 on Build GRCh38
rs772562923 Chr.17: 58270745-58270747 on Build GRCh38
rs752406825 Chr.17: 58270732-58270732 on Build GRCh38
rs35897051 Chr.17: 58270865-58270865 on Build GRCh38
rs554986250 Chr.17: 58270922-58270922 on Build GRCh38
rs906263253 Chr.17: 58270893-58270893 on Build GRCh38
rs780418062 Chr.17: 58270773-58270773 on Build GRCh38
rs1035833511 Chr.17: 58270672-58270672 on Build GRCh38
rs984364524 Chr.17: 58270596-58270596 on Build GRCh38
rs751384055 Chr.17: 58270884-58270884 on Build GRCh38
rs2071409 Chr.17: 58270869-58270869 on Build GRCh38
rs770133415 Chr.17: 58270815-58270815 on Build GRCh38
rs536611098 Chr.17: 58270897-58270897 on Build GRCh38
rs770356554 Chr.17: 58270720-58270720 on Build GRCh38
rs756233510 Chr.17: 58270899-58270899 on Build GRCh38
rs758324501 Chr.17: 58270771-58270771 on Build GRCh38
rs763293793 Chr.17: 58270843-58270843 on Build GRCh38
rs1042457586 Chr.17: 58270872-58270872 on Build GRCh38
rs746364041 Chr.17: 58270639-58270640 on Build GRCh38
rs780053814 Chr.17: 58270876-58270876 on Build GRCh38
rs760299413 Chr.17: 58270848-58270848 on Build GRCh38
rs749254387 Chr.17: 58270623-58270623 on Build GRCh38
rs753796920 Chr.17: 58270760-58270760 on Build GRCh38
rs11555567 Chr.17: 58270503-58270503 on Build GRCh38
rs531709146 Chr.17: 58270599-58270599 on Build GRCh38
rs754301094 Chr.17: 58270708-58270708 on Build GRCh38
rs775170924 Chr.17: 58270529-58270529 on Build GRCh38
rs367673422 Chr.17: 58270908-58270908 on Build GRCh38
rs958924282 Chr.17: 58270701-58270701 on Build GRCh38
rs764394088 Chr.17: 58270653-58270653 on Build GRCh38
rs34738107 Chr.17: 58270654-58270655 on Build GRCh38
rs748290797 Chr.17: 58270811-58270811 on Build GRCh38
rs781507932 Chr.17: 58270894-58270894 on Build GRCh38
rs932535698 Chr.17: 58270822-58270822 on Build GRCh38
rs773724375 Chr.17: 58270823-58270823 on Build GRCh38
rs763313821 Chr.17: 58270721-58270721 on Build GRCh38
rs919909030 Chr.17: 58270734-58270734 on Build GRCh38
rs1003265878 Chr.17: 58270923-58270923 on Build GRCh38
rs753251694 Chr.17: 58270864-58270864 on Build GRCh38
rs1052496038 Chr.17: 58270944-58270944 on Build GRCh38
rs776055976 Chr.17: 58270824-58270825 on Build GRCh38
rs755089250 Chr.17: 58270889-58270889 on Build GRCh38
rs567352560 Chr.17: 58270950-58270950 on Build GRCh38
rs758265207 Chr.17: 58270873-58270873 on Build GRCh38
rs61747608 Chr.17: 58270761-58270761 on Build GRCh38
rs369638284 Chr.17: 58270584-58270584 on Build GRCh38
rs757049213 Chr.17: 58270628-58270628 on Build GRCh38
rs35702888 Chr.17: 58270847-58270847 on Build GRCh38
rs150925221 Chr.17: 58270793-58270793 on Build GRCh38
rs375686331 Chr.17: 58270612-58270612 on Build GRCh38
rs772350102 Chr.17: 58270635-58270635 on Build GRCh38
rs777163659 Chr.17: 58270713-58270713 on Build GRCh38
rs34685535 Chr.17: 58270959-58270959 on Build GRCh38
rs763556195 Chr.17: 58270757-58270757 on Build GRCh38
rs778518485 Chr.17: 58270630-58270630 on Build GRCh38
rs935344002 Chr.17: 58270881-58270881 on Build GRCh38
rs763816239 Chr.17: 58270863-58270863 on Build GRCh38
rs762275674 Chr.17: 58270714-58270714 on Build GRCh38
rs921115203 Chr.17: 58270816-58270816 on Build GRCh38
rs543839552 Chr.17: 58270615-58270615 on Build GRCh38
rs1002964958 Chr.17: 58270668-58270668 on Build GRCh38
rs935394938 Chr.17: 58270640-58270640 on Build GRCh38
rs911474440 Chr.17: 58270597-58270597 on Build GRCh38
rs2759 Chr.17: 58270745-58270745 on Build GRCh38
rs139752741 Chr.17: 58270781-58270781 on Build GRCh38
rs113203253 Chr.17: 58270707-58270707 on Build GRCh38
rs767101775 Chr.17: 58270731-58270731 on Build GRCh38
rs745685034 Chr.17: 58270631-58270631 on Build GRCh38
rs778995484 Chr.17: 58270766-58270766 on Build GRCh38
rs944109881 Chr.17: 58270780-58270780 on Build GRCh38
rs780434488 Chr.17: 58270643-58270643 on Build GRCh38
rs931495759 Chr.17: 58270591-58270591 on Build GRCh38
rs972497970 Chr.17: 58270528-58270528 on Build GRCh38

View Full Product Details