Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 76053362-76053872 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SRP68
Entrez Gene ID
6730
Gene Name
signal recognition particle 68
Gene Aliases
-
UniGene
Hs.514495
Gene Chromosome Location
Chr.17: 76038775-76072526 on Build GRCh38
Transcripts
NM_001260502.1 NM_001260503.1 NM_014230.3 NR_048541.1

SNP Detail

SNP ID Chromosome Location
rs1045617199 Chr.17: 76053612-76053612 on Build GRCh38
rs1002196058 Chr.17: 76053723-76053723 on Build GRCh38
rs955051725 Chr.17: 76053792-76053792 on Build GRCh38
rs946047601 Chr.17: 76053401-76053401 on Build GRCh38
rs563187594 Chr.17: 76053728-76053728 on Build GRCh38
rs573594682 Chr.17: 76053697-76053697 on Build GRCh38
rs908276146 Chr.17: 76053434-76053434 on Build GRCh38
rs371203161 Chr.17: 76053748-76053748 on Build GRCh38
rs181123834 Chr.17: 76053769-76053769 on Build GRCh38
rs144311457 Chr.17: 76053537-76053537 on Build GRCh38
rs753698221 Chr.17: 76053679-76053679 on Build GRCh38
rs941219190 Chr.17: 76053488-76053488 on Build GRCh38
rs907253548 Chr.17: 76053625-76053625 on Build GRCh38
rs113823201 Chr.17: 76053403-76053403 on Build GRCh38
rs554014884 Chr.17: 76053833-76053833 on Build GRCh38
rs573014624 Chr.17: 76053518-76053519 on Build GRCh38
rs890290535 Chr.17: 76053520-76053520 on Build GRCh38
rs866135124 Chr.17: 76053667-76053667 on Build GRCh38
rs147730090 Chr.17: 76053435-76053435 on Build GRCh38
rs960206004 Chr.17: 76053750-76053750 on Build GRCh38
rs766966712 Chr.17: 76053390-76053390 on Build GRCh38
rs997806063 Chr.17: 76053701-76053701 on Build GRCh38
rs753047599 Chr.17: 76053814-76053814 on Build GRCh38
rs552029714 Chr.17: 76053753-76053753 on Build GRCh38
rs948612037 Chr.17: 76053587-76053587 on Build GRCh38
rs1008567862 Chr.17: 76053545-76053545 on Build GRCh38
rs575067315 Chr.17: 76053392-76053392 on Build GRCh38
rs915834632 Chr.17: 76053558-76053558 on Build GRCh38
rs765999087 Chr.17: 76053669-76053669 on Build GRCh38
rs540222516 Chr.17: 76053661-76053661 on Build GRCh38
rs768084277 Chr.17: 76053556-76053556 on Build GRCh38
rs368574234 Chr.17: 76053610-76053610 on Build GRCh38
rs189791007 Chr.17: 76053680-76053680 on Build GRCh38
rs1020403034 Chr.17: 76053838-76053838 on Build GRCh38
rs982413192 Chr.17: 76053806-76053806 on Build GRCh38
rs1043346423 Chr.17: 76053442-76053442 on Build GRCh38
rs998794475 Chr.17: 76053638-76053638 on Build GRCh38
rs992886566 Chr.17: 76053752-76053752 on Build GRCh38
rs982550514 Chr.17: 76053416-76053416 on Build GRCh38
rs900660146 Chr.17: 76053686-76053686 on Build GRCh38
rs955021056 Chr.17: 76053393-76053393 on Build GRCh38
rs577695566 Chr.17: 76053546-76053546 on Build GRCh38
rs543312641 Chr.17: 76053709-76053709 on Build GRCh38
rs760365587 Chr.17: 76053657-76053657 on Build GRCh38
rs925923247 Chr.17: 76053480-76053480 on Build GRCh38
rs931801916 Chr.17: 76053497-76053497 on Build GRCh38
rs772676703 Chr.17: 76053417-76053417 on Build GRCh38
rs374518320 Chr.17: 76053799-76053799 on Build GRCh38

View Full Product Details