Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 76544124-76544628 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYGB
Entrez Gene ID
114757
Gene Name
cytoglobin
Gene Aliases
HGB, STAP
UniGene
Hs.95120
Gene Chromosome Location
Chr.17: 76527348-76557692 on Build GRCh38
Transcripts
XM_017024116.1 NM_001077620.2 NR_033357.1 XM_017025013.1 XM_017025014.1 XM_017025015.1

Gene Symbol
PRCD
Entrez Gene ID
768206
Gene Name
progressive rod-cone degeneration
Gene Aliases
RP36
UniGene
Hs.634380
Gene Chromosome Location
Chr.17: 76527586-76555339 on Build GRCh38
Transcripts
XM_017024116.1 NM_001077620.2 NR_033357.1 XM_017025013.1 XM_017025014.1 XM_017025015.1

SNP Detail

SNP ID Chromosome Location
rs750306157 Chr.17: 76544557-76544557 on Build GRCh38
rs545702552 Chr.17: 76544256-76544256 on Build GRCh38
rs991250391 Chr.17: 76544424-76544424 on Build GRCh38
rs560943649 Chr.17: 76544487-76544487 on Build GRCh38
rs959870562 Chr.17: 76544411-76544411 on Build GRCh38
rs886053478 Chr.17: 76544369-76544370 on Build GRCh38
rs771235070 Chr.17: 76544405-76544405 on Build GRCh38
rs902869199 Chr.17: 76544608-76544608 on Build GRCh38
rs550232084 Chr.17: 76544504-76544504 on Build GRCh38
rs754280590 Chr.17: 76544603-76544603 on Build GRCh38
rs902050373 Chr.17: 76544566-76544566 on Build GRCh38
rs886053479 Chr.17: 76544414-76544414 on Build GRCh38
rs778866143 Chr.17: 76544279-76544279 on Build GRCh38
rs951071405 Chr.17: 76544391-76544391 on Build GRCh38
rs942045656 Chr.17: 76544259-76544259 on Build GRCh38
rs776196684 Chr.17: 76544399-76544399 on Build GRCh38
rs760790445 Chr.17: 76544464-76544464 on Build GRCh38
rs753643696 Chr.17: 76544239-76544239 on Build GRCh38
rs1011287370 Chr.17: 76544340-76544340 on Build GRCh38
rs148861933 Chr.17: 76544367-76544368 on Build GRCh38
rs1036860194 Chr.17: 76544317-76544317 on Build GRCh38
rs117784164 Chr.17: 76544582-76544582 on Build GRCh38
rs903223944 Chr.17: 76544254-76544254 on Build GRCh38
rs780217784 Chr.17: 76544423-76544423 on Build GRCh38
rs746497375 Chr.17: 76544594-76544594 on Build GRCh38
rs760052677 Chr.17: 76544427-76544427 on Build GRCh38
rs764800131 Chr.17: 76544216-76544216 on Build GRCh38
rs185614691 Chr.17: 76544488-76544488 on Build GRCh38
rs889380415 Chr.17: 76544270-76544270 on Build GRCh38
rs1021596798 Chr.17: 76544596-76544596 on Build GRCh38
rs576935421 Chr.17: 76544253-76544253 on Build GRCh38
rs756839135 Chr.17: 76544188-76544188 on Build GRCh38
rs751296907 Chr.17: 76544336-76544336 on Build GRCh38
rs779714454 Chr.17: 76544310-76544310 on Build GRCh38
rs768412990 Chr.17: 76544386-76544386 on Build GRCh38
rs890510364 Chr.17: 76544516-76544516 on Build GRCh38
rs943664138 Chr.17: 76544542-76544542 on Build GRCh38
rs774760932 Chr.17: 76544192-76544192 on Build GRCh38
rs991795551 Chr.17: 76544420-76544420 on Build GRCh38
rs767521000 Chr.17: 76544433-76544433 on Build GRCh38
rs745814897 Chr.17: 76544293-76544293 on Build GRCh38
rs901996843 Chr.17: 76544334-76544334 on Build GRCh38
rs200819376 Chr.17: 76544145-76544145 on Build GRCh38
rs528379750 Chr.17: 76544400-76544400 on Build GRCh38
rs550804575 Chr.17: 76544337-76544337 on Build GRCh38
rs949914629 Chr.17: 76544477-76544477 on Build GRCh38
rs886053477 Chr.17: 76544255-76544255 on Build GRCh38
rs963708528 Chr.17: 76544236-76544236 on Build GRCh38
rs929274951 Chr.17: 76544501-76544501 on Build GRCh38
rs777880280 Chr.17: 76544192-76544193 on Build GRCh38
rs937220356 Chr.17: 76544199-76544199 on Build GRCh38
rs1052250316 Chr.17: 76544325-76544325 on Build GRCh38
rs915865010 Chr.17: 76544438-76544438 on Build GRCh38
rs989779384 Chr.17: 76544238-76544238 on Build GRCh38
rs755211458 Chr.17: 76544560-76544560 on Build GRCh38
rs2278639 Chr.17: 76544410-76544410 on Build GRCh38
rs955156906 Chr.17: 76544215-76544215 on Build GRCh38
rs1037754603 Chr.17: 76544147-76544147 on Build GRCh38
rs766888439 Chr.17: 76544184-76544184 on Build GRCh38
rs775531139 Chr.17: 76544434-76544434 on Build GRCh38
rs756335948 Chr.17: 76544228-76544228 on Build GRCh38
rs886869694 Chr.17: 76544601-76544601 on Build GRCh38
rs1037703544 Chr.17: 76544264-76544264 on Build GRCh38
rs372700920 Chr.17: 76544498-76544498 on Build GRCh38
rs1007692136 Chr.17: 76544332-76544332 on Build GRCh38
rs757028440 Chr.17: 76544531-76544531 on Build GRCh38
rs1011150069 Chr.17: 76544584-76544584 on Build GRCh38
rs748261845 Chr.17: 76544585-76544585 on Build GRCh38
rs1000157197 Chr.17: 76544255-76544255 on Build GRCh38
rs764100191 Chr.17: 76544257-76544257 on Build GRCh38
rs1008203164 Chr.17: 76544280-76544280 on Build GRCh38
rs73998961 Chr.17: 76544583-76544583 on Build GRCh38
rs1025397619 Chr.17: 76544372-76544372 on Build GRCh38
rs144319284 Chr.17: 76544240-76544240 on Build GRCh38
rs1021274145 Chr.17: 76544374-76544374 on Build GRCh38
rs11655083 Chr.17: 76544316-76544316 on Build GRCh38
rs559175222 Chr.17: 76544297-76544297 on Build GRCh38
rs752862392 Chr.17: 76544217-76544217 on Build GRCh38
rs764355852 Chr.17: 76544237-76544237 on Build GRCh38
rs897900280 Chr.17: 76544166-76544166 on Build GRCh38
rs890647172 Chr.17: 76544330-76544330 on Build GRCh38
rs951019656 Chr.17: 76544180-76544180 on Build GRCh38
rs909803223 Chr.17: 76544196-76544196 on Build GRCh38
rs759624725 Chr.17: 76544195-76544195 on Build GRCh38
rs1055644821 Chr.17: 76544220-76544220 on Build GRCh38
rs764953862 Chr.17: 76544550-76544550 on Build GRCh38

View Full Product Details