Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 76737765-76738266 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MFSD11
Entrez Gene ID
79157
Gene Name
major facilitator superfamily domain containing 11
Gene Aliases
ET
UniGene
Hs.732019
Gene Chromosome Location
Chr.17: 76736565-76803805 on Build GRCh38
Transcripts
NM_001242532.1 NM_001242533.1 NM_001242534.1 NM_001242535.1 NM_001242536.1 NM_001242537.1 NM_024311.3 XM_011525236.2 XM_011525237.2 XM_011525238.2 XM_011525239.2 XM_011525240.2 XM_011525241.2 XM_011525242.1 XM_011525244.1 XM_011525247.2 XM_017025065.1 XM_017025066.1 XM_017025067.1 XM_017025068.1 XM_017025069.1 XM_017025070.1

SNP Detail

SNP ID Chromosome Location
rs867813792 Chr.17: 76737961-76737961 on Build GRCh38
rs573137937 Chr.17: 76738153-76738153 on Build GRCh38
rs546168489 Chr.17: 76737998-76737998 on Build GRCh38
rs963859170 Chr.17: 76738079-76738079 on Build GRCh38
rs538001372 Chr.17: 76738141-76738141 on Build GRCh38
rs1041985616 Chr.17: 76737789-76737789 on Build GRCh38
rs969265755 Chr.17: 76737838-76737838 on Build GRCh38
rs898548915 Chr.17: 76737949-76737949 on Build GRCh38
rs763706333 Chr.17: 76738235-76738235 on Build GRCh38
rs967780833 Chr.17: 76738098-76738098 on Build GRCh38
rs373795715 Chr.17: 76737819-76737819 on Build GRCh38
rs946475225 Chr.17: 76737897-76737897 on Build GRCh38
rs1015722277 Chr.17: 76737813-76737813 on Build GRCh38
rs940294405 Chr.17: 76738091-76738091 on Build GRCh38
rs891121891 Chr.17: 76737927-76737927 on Build GRCh38
rs1004288611 Chr.17: 76737809-76737809 on Build GRCh38
rs1038138147 Chr.17: 76738031-76738031 on Build GRCh38
rs1025973014 Chr.17: 76738047-76738047 on Build GRCh38
rs237054 Chr.17: 76738241-76738241 on Build GRCh38
rs367792082 Chr.17: 76738034-76738034 on Build GRCh38
rs978059452 Chr.17: 76737956-76737956 on Build GRCh38
rs979176941 Chr.17: 76738038-76738038 on Build GRCh38
rs771209831 Chr.17: 76738188-76738188 on Build GRCh38
rs987904503 Chr.17: 76737869-76737869 on Build GRCh38
rs960657913 Chr.17: 76738005-76738005 on Build GRCh38
rs977484334 Chr.17: 76738115-76738115 on Build GRCh38
rs1018038737 Chr.17: 76738072-76738072 on Build GRCh38
rs1034677812 Chr.17: 76737850-76737850 on Build GRCh38
rs533494382 Chr.17: 76737943-76737943 on Build GRCh38
rs905053409 Chr.17: 76737962-76737962 on Build GRCh38
rs237055 Chr.17: 76737920-76737920 on Build GRCh38
rs898181442 Chr.17: 76737861-76737862 on Build GRCh38
rs564794103 Chr.17: 76737941-76737941 on Build GRCh38
rs905022409 Chr.17: 76738185-76738185 on Build GRCh38
rs947765335 Chr.17: 76738021-76738021 on Build GRCh38
rs938856591 Chr.17: 76737794-76737794 on Build GRCh38
rs986627951 Chr.17: 76737905-76737905 on Build GRCh38
rs981119223 Chr.17: 76737844-76737844 on Build GRCh38
rs781407820 Chr.17: 76738110-76738110 on Build GRCh38
rs987070338 Chr.17: 76738075-76738075 on Build GRCh38
rs553744442 Chr.17: 76738054-76738054 on Build GRCh38
rs148903124 Chr.17: 76738011-76738012 on Build GRCh38
rs544488545 Chr.17: 76737922-76737922 on Build GRCh38
rs568859548 Chr.17: 76738116-76738116 on Build GRCh38
rs573351710 Chr.17: 76737859-76737859 on Build GRCh38
rs562064298 Chr.17: 76737917-76737917 on Build GRCh38
rs929721255 Chr.17: 76738214-76738214 on Build GRCh38
rs58466357 Chr.17: 76738016-76738017 on Build GRCh38
rs553450716 Chr.17: 76737820-76737820 on Build GRCh38
rs1012011494 Chr.17: 76737824-76737824 on Build GRCh38
rs907350528 Chr.17: 76737793-76737793 on Build GRCh38
rs991760564 Chr.17: 76738121-76738121 on Build GRCh38
rs760346850 Chr.17: 76738232-76738232 on Build GRCh38
rs1020587682 Chr.17: 76738029-76738029 on Build GRCh38
rs921448888 Chr.17: 76737911-76737911 on Build GRCh38
rs559752143 Chr.17: 76738050-76738050 on Build GRCh38
rs373687484 Chr.17: 76737991-76737991 on Build GRCh38
rs748348604 Chr.17: 76738117-76738117 on Build GRCh38
rs993885359 Chr.17: 76738046-76738046 on Build GRCh38
rs1035386603 Chr.17: 76737992-76737992 on Build GRCh38
rs1027497897 Chr.17: 76737827-76737827 on Build GRCh38
rs1044533659 Chr.17: 76738173-76738173 on Build GRCh38
rs919719538 Chr.17: 76738181-76738181 on Build GRCh38
rs1055938787 Chr.17: 76737981-76737981 on Build GRCh38
rs237056 Chr.17: 76737899-76737899 on Build GRCh38
rs747071924 Chr.17: 76738027-76738027 on Build GRCh38
rs896364241 Chr.17: 76738228-76738228 on Build GRCh38
rs780769595 Chr.17: 76738008-76738008 on Build GRCh38
rs557065279 Chr.17: 76737825-76737825 on Build GRCh38
rs995604498 Chr.17: 76737960-76737960 on Build GRCh38
rs1056306979 Chr.17: 76738216-76738216 on Build GRCh38
rs891146767 Chr.17: 76737800-76737800 on Build GRCh38
rs1051081640 Chr.17: 76737799-76737799 on Build GRCh38
rs912767861 Chr.17: 76738076-76738076 on Build GRCh38
rs1002118252 Chr.17: 76737983-76737983 on Build GRCh38
rs773987084 Chr.17: 76738169-76738169 on Build GRCh38
rs745453489 Chr.17: 76737881-76737881 on Build GRCh38
rs987731500 Chr.17: 76738010-76738010 on Build GRCh38
rs774177598 Chr.17: 76738117-76738117 on Build GRCh38
rs73996694 Chr.17: 76738095-76738095 on Build GRCh38
rs750772439 Chr.17: 76738236-76738236 on Build GRCh38

View Full Product Details