Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 78215877-78216387 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BIRC5
Entrez Gene ID
332
Gene Name
baculoviral IAP repeat containing 5
Gene Aliases
API4, EPR-1
UniGene
Hs.744872
Gene Chromosome Location
Chr.17: 78214196-78225635 on Build GRCh38
Transcripts
NM_001012270.1 NM_001012271.1 NM_001168.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1723030 c.228G>A p.P76P Chr.17: 78215972-78215972 on Build GRCh38
3388241 c.280C>T p.R94W Chr.17: 78216024-78216024 on Build GRCh38
1723032 c.253A>C p.T85P Chr.17: 78215997-78215997 on Build GRCh38
1723031 c.241T>C p.Y81H Chr.17: 78215985-78215985 on Build GRCh38
1589264 c.230G>A p.G77D Chr.17: 78215974-78215974 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs897524240 Chr.17: 78216248-78216248 on Build GRCh38
rs568503667 Chr.17: 78216283-78216283 on Build GRCh38
rs766583426 Chr.17: 78216014-78216014 on Build GRCh38
rs373663067 Chr.17: 78216282-78216282 on Build GRCh38
rs371423067 Chr.17: 78216227-78216227 on Build GRCh38
rs909029152 Chr.17: 78216036-78216036 on Build GRCh38
rs1020576991 Chr.17: 78216249-78216249 on Build GRCh38
rs768115737 Chr.17: 78216063-78216063 on Build GRCh38
rs200848570 Chr.17: 78215998-78215998 on Build GRCh38
rs941971004 Chr.17: 78216206-78216206 on Build GRCh38
rs189943941 Chr.17: 78216045-78216045 on Build GRCh38
rs17882187 Chr.17: 78216165-78216165 on Build GRCh38
rs775104711 Chr.17: 78215990-78215990 on Build GRCh38
rs959881687 Chr.17: 78216031-78216031 on Build GRCh38
rs778052929 Chr.17: 78216032-78216032 on Build GRCh38
rs926238121 Chr.17: 78215976-78215976 on Build GRCh38
rs745693033 Chr.17: 78216055-78216055 on Build GRCh38
rs35054363 Chr.17: 78216327-78216328 on Build GRCh38
rs55850455 Chr.17: 78216264-78216264 on Build GRCh38
rs143458850 Chr.17: 78215928-78215928 on Build GRCh38
rs922693698 Chr.17: 78215942-78215944 on Build GRCh38
rs147848202 Chr.17: 78216015-78216015 on Build GRCh38
rs146071276 Chr.17: 78215997-78215997 on Build GRCh38
rs756245585 Chr.17: 78216025-78216025 on Build GRCh38
rs759084233 Chr.17: 78215947-78215947 on Build GRCh38
rs552898084 Chr.17: 78215946-78215946 on Build GRCh38
rs112819650 Chr.17: 78216260-78216261 on Build GRCh38
rs775156039 Chr.17: 78215948-78215948 on Build GRCh38
rs371607756 Chr.17: 78216243-78216243 on Build GRCh38
rs770701265 Chr.17: 78215985-78215985 on Build GRCh38
rs765541204 Chr.17: 78216105-78216105 on Build GRCh38
rs373827152 Chr.17: 78216037-78216037 on Build GRCh38
rs375550186 Chr.17: 78216061-78216061 on Build GRCh38
rs1036272877 Chr.17: 78216214-78216214 on Build GRCh38
rs550748287 Chr.17: 78216321-78216321 on Build GRCh38
rs1055787267 Chr.17: 78216174-78216174 on Build GRCh38
rs772161908 Chr.17: 78215988-78215988 on Build GRCh38
rs761176997 Chr.17: 78216084-78216084 on Build GRCh38
rs763369529 Chr.17: 78216003-78216003 on Build GRCh38
rs780548681 Chr.17: 78215959-78215959 on Build GRCh38
rs769507492 Chr.17: 78216072-78216072 on Build GRCh38
rs944458559 Chr.17: 78216117-78216117 on Build GRCh38
rs764025611 Chr.17: 78216089-78216089 on Build GRCh38
rs757384499 Chr.17: 78216099-78216099 on Build GRCh38
rs895918417 Chr.17: 78216215-78216215 on Build GRCh38
rs374475800 Chr.17: 78215987-78215987 on Build GRCh38
rs967199028 Chr.17: 78216255-78216255 on Build GRCh38
rs748301248 Chr.17: 78215927-78215927 on Build GRCh38
rs755105352 Chr.17: 78215926-78215926 on Build GRCh38
rs115296168 Chr.17: 78215971-78215971 on Build GRCh38
rs971798144 Chr.17: 78216305-78216305 on Build GRCh38
rs761312890 Chr.17: 78216066-78216066 on Build GRCh38
rs985872250 Chr.17: 78216208-78216208 on Build GRCh38
rs563540138 Chr.17: 78216109-78216109 on Build GRCh38
rs759987155 Chr.17: 78216020-78216020 on Build GRCh38
rs1027280127 Chr.17: 78216093-78216093 on Build GRCh38
rs776045831 Chr.17: 78216082-78216082 on Build GRCh38
rs749078071 Chr.17: 78215942-78215942 on Build GRCh38
rs368648511 Chr.17: 78215993-78215993 on Build GRCh38
rs763874122 Chr.17: 78215957-78215957 on Build GRCh38
rs371588539 Chr.17: 78216024-78216024 on Build GRCh38
rs776207933 Chr.17: 78216065-78216065 on Build GRCh38
rs372074460 Chr.17: 78215904-78215904 on Build GRCh38
rs918758142 Chr.17: 78216132-78216132 on Build GRCh38
rs531036288 Chr.17: 78216285-78216285 on Build GRCh38
rs1001514296 Chr.17: 78216168-78216168 on Build GRCh38
rs750217306 Chr.17: 78216106-78216106 on Build GRCh38
rs1001260510 Chr.17: 78216280-78216280 on Build GRCh38
rs966224902 Chr.17: 78216297-78216297 on Build GRCh38
rs763273198 Chr.17: 78215967-78215967 on Build GRCh38
rs930221340 Chr.17: 78216149-78216149 on Build GRCh38
rs531403154 Chr.17: 78216228-78216228 on Build GRCh38
rs752796895 Chr.17: 78215978-78215978 on Build GRCh38
rs753977420 Chr.17: 78216098-78216098 on Build GRCh38
rs768042848 Chr.17: 78216021-78216021 on Build GRCh38
rs867317238 Chr.17: 78216115-78216115 on Build GRCh38
rs1045895790 Chr.17: 78216266-78216266 on Build GRCh38
rs760358572 Chr.17: 78215956-78215956 on Build GRCh38
rs183731583 Chr.17: 78216337-78216337 on Build GRCh38
rs370626221 Chr.17: 78215940-78215940 on Build GRCh38
rs35726863 Chr.17: 78216260-78216260 on Build GRCh38
rs1032867010 Chr.17: 78216266-78216266 on Build GRCh38
rs1004534716 Chr.17: 78216329-78216329 on Build GRCh38
rs936275314 Chr.17: 78216226-78216226 on Build GRCh38
rs532641741 Chr.17: 78216122-78216122 on Build GRCh38
rs752070613 Chr.17: 78215972-78215972 on Build GRCh38
rs771479665 Chr.17: 78215920-78215920 on Build GRCh38
rs750764370 Chr.17: 78215966-78215966 on Build GRCh38
rs767999821 Chr.17: 78216077-78216077 on Build GRCh38
rs559685844 Chr.17: 78216204-78216204 on Build GRCh38
rs772108644 Chr.17: 78216056-78216056 on Build GRCh38
rs376739440 Chr.17: 78215913-78215913 on Build GRCh38
rs780185239 Chr.17: 78216060-78216060 on Build GRCh38
rs953277648 Chr.17: 78216275-78216275 on Build GRCh38
rs1048646183 Chr.17: 78216150-78216150 on Build GRCh38
rs1018555790 Chr.17: 78216296-78216296 on Build GRCh38
rs781190694 Chr.17: 78215977-78215977 on Build GRCh38

View Full Product Details