Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 36085833-36086334 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TEKT2
Entrez Gene ID
27285
Gene Name
tektin 2
Gene Aliases
TEKTB1, TEKTIN-T, h-tektin-t
UniGene
Hs.127111
Gene Chromosome Location
Chr.1: 36084075-36088275 on Build GRCh38
Transcripts
NM_014466.2 XM_005270753.2 XM_011541258.2 XM_017001055.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4960239 c.369G>A p.R123R Chr.1: 36085922-36085922 on Build GRCh38
6283869 c.318G>T p.K106N Chr.1: 36085871-36085871 on Build GRCh38
5607101 c.307C>T p.L103L Chr.1: 36085860-36085860 on Build GRCh38
3419168 c.478G>A p.E160K Chr.1: 36086031-36086031 on Build GRCh38
235476 c.306C>T p.N102N Chr.1: 36085859-36085859 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs566607949 Chr.1: 36085948-36085948 on Build GRCh38
rs1020257492 Chr.1: 36086291-36086291 on Build GRCh38
rs764898070 Chr.1: 36085918-36085918 on Build GRCh38
rs897417238 Chr.1: 36086173-36086173 on Build GRCh38
rs1006473850 Chr.1: 36086284-36086284 on Build GRCh38
rs1054634651 Chr.1: 36085874-36085874 on Build GRCh38
rs1045913288 Chr.1: 36086211-36086211 on Build GRCh38
rs747699239 Chr.1: 36086011-36086013 on Build GRCh38
rs188482509 Chr.1: 36086063-36086063 on Build GRCh38
rs776242019 Chr.1: 36085912-36085912 on Build GRCh38
rs753381474 Chr.1: 36086303-36086303 on Build GRCh38
rs112822096 Chr.1: 36086238-36086238 on Build GRCh38
rs773254140 Chr.1: 36085875-36085875 on Build GRCh38
rs375817757 Chr.1: 36085999-36085999 on Build GRCh38
rs762951584 Chr.1: 36085923-36085923 on Build GRCh38
rs962706839 Chr.1: 36085994-36085994 on Build GRCh38
rs941705643 Chr.1: 36086149-36086149 on Build GRCh38
rs767361605 Chr.1: 36086091-36086091 on Build GRCh38
rs922967664 Chr.1: 36086201-36086201 on Build GRCh38
rs408986 Chr.1: 36086294-36086294 on Build GRCh38
rs769361007 Chr.1: 36086022-36086022 on Build GRCh38
rs548139441 Chr.1: 36086060-36086060 on Build GRCh38
rs146498328 Chr.1: 36085893-36085893 on Build GRCh38
rs368324904 Chr.1: 36085859-36085859 on Build GRCh38
rs759300659 Chr.1: 36086072-36086072 on Build GRCh38
rs772790502 Chr.1: 36086059-36086059 on Build GRCh38
rs1012009591 Chr.1: 36086270-36086270 on Build GRCh38
rs779374541 Chr.1: 36085870-36085870 on Build GRCh38
rs1013324757 Chr.1: 36086008-36086008 on Build GRCh38
rs372793918 Chr.1: 36086285-36086285 on Build GRCh38
rs893190060 Chr.1: 36086258-36086258 on Build GRCh38
rs766027019 Chr.1: 36085924-36085924 on Build GRCh38
rs886283606 Chr.1: 36085967-36085967 on Build GRCh38
rs577598580 Chr.1: 36086178-36086178 on Build GRCh38
rs758959693 Chr.1: 36085984-36085984 on Build GRCh38
rs914300595 Chr.1: 36086160-36086160 on Build GRCh38
rs147164539 Chr.1: 36085933-36085933 on Build GRCh38
rs419653 Chr.1: 36085894-36085894 on Build GRCh38
rs370668257 Chr.1: 36086305-36086305 on Build GRCh38
rs369442918 Chr.1: 36085911-36085911 on Build GRCh38
rs551496416 Chr.1: 36085938-36085938 on Build GRCh38
rs780234653 Chr.1: 36085985-36085985 on Build GRCh38
rs757319942 Chr.1: 36085946-36085946 on Build GRCh38
rs370996150 Chr.1: 36085866-36085866 on Build GRCh38
rs769272787 Chr.1: 36086038-36086038 on Build GRCh38
rs746296836 Chr.1: 36085981-36085981 on Build GRCh38
rs146319916 Chr.1: 36085930-36085930 on Build GRCh38
rs1022996309 Chr.1: 36086056-36086056 on Build GRCh38
rs113338638 Chr.1: 36085986-36085986 on Build GRCh38
rs930196390 Chr.1: 36086198-36086198 on Build GRCh38
rs1034832139 Chr.1: 36086245-36086245 on Build GRCh38
rs956409294 Chr.1: 36086083-36086083 on Build GRCh38
rs542702527 Chr.1: 36086261-36086261 on Build GRCh38
rs538550158 Chr.1: 36086104-36086104 on Build GRCh38
rs1036007553 Chr.1: 36086155-36086155 on Build GRCh38
rs375676685 Chr.1: 36085921-36085921 on Build GRCh38
rs933207960 Chr.1: 36086207-36086207 on Build GRCh38
rs764266107 Chr.1: 36085937-36085937 on Build GRCh38
rs544655982 Chr.1: 36086199-36086199 on Build GRCh38
rs560335590 Chr.1: 36086221-36086221 on Build GRCh38
rs751390286 Chr.1: 36085929-36085929 on Build GRCh38
rs372535680 Chr.1: 36085998-36085998 on Build GRCh38
rs746586703 Chr.1: 36085901-36085901 on Build GRCh38
rs192926017 Chr.1: 36086136-36086136 on Build GRCh38
rs886233305 Chr.1: 36086279-36086279 on Build GRCh38
rs200649133 Chr.1: 36086064-36086064 on Build GRCh38
rs780642414 Chr.1: 36085917-36085918 on Build GRCh38
rs758565290 Chr.1: 36086141-36086141 on Build GRCh38
rs943171596 Chr.1: 36086182-36086182 on Build GRCh38
rs908906537 Chr.1: 36086139-36086139 on Build GRCh38
rs748191599 Chr.1: 36086115-36086115 on Build GRCh38

View Full Product Details