Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 36799338-36799841 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GRIK3
Entrez Gene ID
2899
Gene Name
glutamate ionotropic receptor kainate type subunit 3
Gene Aliases
EAA5, GLR7, GLUR7, GluK3, GluR7a
UniGene
Hs.128848
Gene Chromosome Location
Chr.1: 36795527-37034243 on Build GRCh38
Transcripts
NM_000831.3

SNP Detail

SNP ID Chromosome Location
rs12748869 Chr.1: 36799531-36799531 on Build GRCh38
rs757585924 Chr.1: 36799749-36799749 on Build GRCh38
rs1046530255 Chr.1: 36799391-36799391 on Build GRCh38
rs185577526 Chr.1: 36799507-36799507 on Build GRCh38
rs896404618 Chr.1: 36799711-36799711 on Build GRCh38
rs1030172449 Chr.1: 36799720-36799720 on Build GRCh38
rs115760608 Chr.1: 36799534-36799534 on Build GRCh38
rs568972648 Chr.1: 36799577-36799577 on Build GRCh38
rs572846832 Chr.1: 36799423-36799423 on Build GRCh38
rs986803634 Chr.1: 36799547-36799547 on Build GRCh38
rs1011511995 Chr.1: 36799461-36799461 on Build GRCh38
rs533813026 Chr.1: 36799458-36799458 on Build GRCh38
rs573513347 Chr.1: 36799482-36799482 on Build GRCh38
rs955861527 Chr.1: 36799434-36799434 on Build GRCh38
rs976070831 Chr.1: 36799567-36799567 on Build GRCh38
rs539428195 Chr.1: 36799415-36799415 on Build GRCh38
rs954675686 Chr.1: 36799735-36799735 on Build GRCh38
rs1020700338 Chr.1: 36799488-36799488 on Build GRCh38
rs551382868 Chr.1: 36799748-36799748 on Build GRCh38
rs996087009 Chr.1: 36799647-36799647 on Build GRCh38
rs754348661 Chr.1: 36799646-36799646 on Build GRCh38
rs575643245 Chr.1: 36799496-36799496 on Build GRCh38
rs142466693 Chr.1: 36799599-36799599 on Build GRCh38
rs916852794 Chr.1: 36799557-36799557 on Build GRCh38
rs1021283281 Chr.1: 36799644-36799644 on Build GRCh38
rs922591361 Chr.1: 36799555-36799555 on Build GRCh38
rs148786565 Chr.1: 36799546-36799546 on Build GRCh38
rs948302683 Chr.1: 36799571-36799571 on Build GRCh38
rs181788656 Chr.1: 36799395-36799395 on Build GRCh38
rs935472110 Chr.1: 36799790-36799790 on Build GRCh38
rs550854733 Chr.1: 36799564-36799564 on Build GRCh38
rs902149398 Chr.1: 36799741-36799741 on Build GRCh38
rs375548536 Chr.1: 36799419-36799419 on Build GRCh38
rs573561305 Chr.1: 36799816-36799816 on Build GRCh38
rs954852164 Chr.1: 36799514-36799514 on Build GRCh38
rs147719957 Chr.1: 36799420-36799420 on Build GRCh38
rs533508439 Chr.1: 36799778-36799778 on Build GRCh38
rs895330797 Chr.1: 36799377-36799377 on Build GRCh38
rs544045011 Chr.1: 36799486-36799486 on Build GRCh38
rs948313817 Chr.1: 36799757-36799757 on Build GRCh38
rs868130461 Chr.1: 36799753-36799753 on Build GRCh38
rs114666596 Chr.1: 36799500-36799500 on Build GRCh38
rs189581210 Chr.1: 36799510-36799510 on Build GRCh38
rs116545485 Chr.1: 36799378-36799378 on Build GRCh38
rs114606283 Chr.1: 36799779-36799779 on Build GRCh38
rs79199587 Chr.1: 36799487-36799487 on Build GRCh38
rs1018305583 Chr.1: 36799776-36799776 on Build GRCh38
rs937846322 Chr.1: 36799628-36799628 on Build GRCh38
rs61769766 Chr.1: 36799723-36799723 on Build GRCh38
rs566113069 Chr.1: 36799751-36799751 on Build GRCh38
rs1002631858 Chr.1: 36799597-36799597 on Build GRCh38
rs75487093 Chr.1: 36799462-36799462 on Build GRCh38
rs1042045377 Chr.1: 36799734-36799734 on Build GRCh38
rs1046594655 Chr.1: 36799775-36799775 on Build GRCh38
rs916571623 Chr.1: 36799756-36799756 on Build GRCh38
rs963474523 Chr.1: 36799752-36799752 on Build GRCh38
rs141923059 Chr.1: 36799610-36799611 on Build GRCh38
rs976290432 Chr.1: 36799805-36799805 on Build GRCh38
rs1011687480 Chr.1: 36799634-36799634 on Build GRCh38
rs529493821 Chr.1: 36799548-36799548 on Build GRCh38
rs1052603840 Chr.1: 36799803-36799803 on Build GRCh38
rs528830618 Chr.1: 36799518-36799518 on Build GRCh38
rs1046579960 Chr.1: 36799590-36799590 on Build GRCh38
rs764680945 Chr.1: 36799592-36799592 on Build GRCh38
rs907888105 Chr.1: 36799530-36799530 on Build GRCh38
rs891475249 Chr.1: 36799601-36799601 on Build GRCh38
rs896168874 Chr.1: 36799465-36799465 on Build GRCh38

View Full Product Details