Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 37997444-37997938 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FHL3
Entrez Gene ID
2275
Gene Name
four and a half LIM domains 3
Gene Aliases
SLIM2
UniGene
Hs.57687
Gene Chromosome Location
Chr.1: 37996765-38005515 on Build GRCh38
Transcripts
NM_001243878.1 NM_004468.4 XM_017000675.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4007648 c.790G>A p.V264I Chr.1: 37997458-37997458 on Build GRCh38
6663940 c.685G>A p.V229I Chr.1: 37997687-37997687 on Build GRCh38
4143898 c.651T>G p.F217L Chr.1: 37997721-37997721 on Build GRCh38
6663941 c.789C>T p.F263F Chr.1: 37997459-37997459 on Build GRCh38
6507213 c.752G>T p.C251F Chr.1: 37997496-37997496 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771646402 Chr.1: 37997784-37997784 on Build GRCh38
rs1019890870 Chr.1: 37997864-37997864 on Build GRCh38
rs765462219 Chr.1: 37997559-37997559 on Build GRCh38
rs965704041 Chr.1: 37997638-37997638 on Build GRCh38
rs774164149 Chr.1: 37997779-37997779 on Build GRCh38
rs868832632 Chr.1: 37997602-37997602 on Build GRCh38
rs201910384 Chr.1: 37997491-37997491 on Build GRCh38
rs755193773 Chr.1: 37997807-37997807 on Build GRCh38
rs1000649837 Chr.1: 37997564-37997564 on Build GRCh38
rs751166216 Chr.1: 37997585-37997585 on Build GRCh38
rs200099240 Chr.1: 37997793-37997793 on Build GRCh38
rs368545534 Chr.1: 37997868-37997868 on Build GRCh38
rs777385451 Chr.1: 37997609-37997609 on Build GRCh38
rs772164466 Chr.1: 37997530-37997530 on Build GRCh38
rs902114413 Chr.1: 37997650-37997650 on Build GRCh38
rs750692565 Chr.1: 37997635-37997635 on Build GRCh38
rs953862083 Chr.1: 37997589-37997589 on Build GRCh38
rs767700327 Chr.1: 37997878-37997878 on Build GRCh38
rs1024201474 Chr.1: 37997718-37997718 on Build GRCh38
rs747155888 Chr.1: 37997662-37997662 on Build GRCh38
rs747066275 Chr.1: 37997537-37997537 on Build GRCh38
rs143886790 Chr.1: 37997842-37997842 on Build GRCh38
rs771354760 Chr.1: 37997778-37997778 on Build GRCh38
rs752706606 Chr.1: 37997717-37997717 on Build GRCh38
rs993470135 Chr.1: 37997653-37997653 on Build GRCh38
rs768652261 Chr.1: 37997663-37997663 on Build GRCh38
rs762600357 Chr.1: 37997584-37997585 on Build GRCh38
rs758666706 Chr.1: 37997645-37997645 on Build GRCh38
rs139509332 Chr.1: 37997869-37997869 on Build GRCh38
rs200788145 Chr.1: 37997671-37997671 on Build GRCh38
rs755857737 Chr.1: 37997523-37997523 on Build GRCh38
rs772938745 Chr.1: 37997571-37997571 on Build GRCh38
rs200944160 Chr.1: 37997587-37997587 on Build GRCh38
rs141005636 Chr.1: 37997804-37997804 on Build GRCh38
rs7366048 Chr.1: 37997832-37997832 on Build GRCh38
rs371636908 Chr.1: 37997900-37997900 on Build GRCh38
rs770251666 Chr.1: 37997770-37997770 on Build GRCh38
rs560025614 Chr.1: 37997613-37997613 on Build GRCh38
rs896708881 Chr.1: 37997484-37997484 on Build GRCh38
rs572633614 Chr.1: 37997727-37997727 on Build GRCh38
rs141131193 Chr.1: 37997865-37997865 on Build GRCh38
rs746022826 Chr.1: 37997525-37997525 on Build GRCh38
rs557418594 Chr.1: 37997726-37997726 on Build GRCh38
rs775200045 Chr.1: 37997859-37997859 on Build GRCh38
rs762237978 Chr.1: 37997555-37997555 on Build GRCh38
rs768150107 Chr.1: 37997867-37997867 on Build GRCh38
rs143651640 Chr.1: 37997914-37997914 on Build GRCh38
rs374208054 Chr.1: 37997687-37997687 on Build GRCh38
rs950723866 Chr.1: 37997890-37997890 on Build GRCh38
rs887824163 Chr.1: 37997664-37997664 on Build GRCh38
rs112651655 Chr.1: 37997562-37997562 on Build GRCh38
rs753704692 Chr.1: 37997796-37997796 on Build GRCh38
rs756216175 Chr.1: 37997824-37997824 on Build GRCh38
rs187833193 Chr.1: 37997765-37997765 on Build GRCh38
rs749977641 Chr.1: 37997477-37997477 on Build GRCh38
rs375411104 Chr.1: 37997853-37997853 on Build GRCh38
rs755072543 Chr.1: 37997764-37997764 on Build GRCh38
rs778429342 Chr.1: 37997825-37997825 on Build GRCh38
rs369660742 Chr.1: 37997490-37997490 on Build GRCh38
rs754432586 Chr.1: 37997916-37997916 on Build GRCh38
rs138169868 Chr.1: 37997835-37997835 on Build GRCh38
rs368278227 Chr.1: 37997738-37997738 on Build GRCh38
rs746206991 Chr.1: 37997843-37997843 on Build GRCh38
rs924280934 Chr.1: 37997808-37997808 on Build GRCh38
rs755954119 Chr.1: 37997604-37997604 on Build GRCh38
rs750513419 Chr.1: 37997495-37997495 on Build GRCh38
rs375939327 Chr.1: 37997681-37997681 on Build GRCh38
rs112195274 Chr.1: 37997873-37997873 on Build GRCh38
rs766007184 Chr.1: 37997889-37997896 on Build GRCh38
rs774143566 Chr.1: 37997688-37997688 on Build GRCh38
rs373813086 Chr.1: 37997494-37997494 on Build GRCh38
rs199860121 Chr.1: 37997596-37997596 on Build GRCh38
rs765127621 Chr.1: 37997870-37997870 on Build GRCh38
rs1013823347 Chr.1: 37997504-37997504 on Build GRCh38
rs767755136 Chr.1: 37997809-37997809 on Build GRCh38
rs774905879 Chr.1: 37997786-37997786 on Build GRCh38
rs778330924 Chr.1: 37997771-37997771 on Build GRCh38
rs762653600 Chr.1: 37997574-37997574 on Build GRCh38
rs751719296 Chr.1: 37997763-37997763 on Build GRCh38
rs141853329 Chr.1: 37997695-37997695 on Build GRCh38
rs769206310 Chr.1: 37997544-37997544 on Build GRCh38
rs150225142 Chr.1: 37997805-37997805 on Build GRCh38
rs370858698 Chr.1: 37997783-37997783 on Build GRCh38
rs752779539 Chr.1: 37997820-37997820 on Build GRCh38
rs977074348 Chr.1: 37997478-37997478 on Build GRCh38
rs767558662 Chr.1: 37997594-37997594 on Build GRCh38
rs751809514 Chr.1: 37997806-37997806 on Build GRCh38
rs535348544 Chr.1: 37997521-37997521 on Build GRCh38
rs752898655 Chr.1: 37997881-37997881 on Build GRCh38
rs775510521 Chr.1: 37997532-37997532 on Build GRCh38
rs200107739 Chr.1: 37997794-37997794 on Build GRCh38
rs1016318708 Chr.1: 37997915-37997915 on Build GRCh38
rs780237956 Chr.1: 37997652-37997652 on Build GRCh38
rs1053551861 Chr.1: 37997840-37997840 on Build GRCh38
rs77418439 Chr.1: 37997554-37997554 on Build GRCh38
rs757939175 Chr.1: 37997485-37997485 on Build GRCh38
rs369998925 Chr.1: 37997674-37997674 on Build GRCh38
rs757882807 Chr.1: 37997918-37997918 on Build GRCh38
rs1047769854 Chr.1: 37997656-37997656 on Build GRCh38
rs34220308 Chr.1: 37997907-37997908 on Build GRCh38
rs779204510 Chr.1: 37997524-37997524 on Build GRCh38
rs528940688 Chr.1: 37997734-37997734 on Build GRCh38
rs938202497 Chr.1: 37997744-37997744 on Build GRCh38
rs766884699 Chr.1: 37997696-37997696 on Build GRCh38
rs770223817 Chr.1: 37997673-37997673 on Build GRCh38
rs145496383 Chr.1: 37997476-37997476 on Build GRCh38
rs766100116 Chr.1: 37997583-37997583 on Build GRCh38
rs780256136 Chr.1: 37997745-37997745 on Build GRCh38

View Full Product Details