Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 38867132-38867650 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GJA9-MYCBP
Entrez Gene ID
100527950
Gene Name
GJA9-MYCBP readthrough
Gene Aliases
-
UniGene
Hs.591506
Gene Chromosome Location
Chr.1: 38862490-38881626 on Build GRCh38
Transcripts
NR_037633.1 NR_037634.1 NR_037635.1 NR_037636.1 NR_037637.1 NM_012333.4 NR_037632.1

Gene Symbol
MYCBP
Entrez Gene ID
26292
Gene Name
MYC binding protein
Gene Aliases
AMY-1
UniGene
Hs.591506
Gene Chromosome Location
Chr.1: 38862490-38873378 on Build GRCh38
Transcripts
NR_037633.1 NR_037634.1 NR_037635.1 NR_037636.1 NR_037637.1 NM_012333.4 NR_037632.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1342441 c.91T>C p.L31L Chr.1: 38867608-38867608 on Build GRCh38
4841637 c.106G>A p.E36K Chr.1: 38867593-38867593 on Build GRCh38
6412786 c.112C>T p.P38S Chr.1: 38867587-38867587 on Build GRCh38
909108 c.137+1G>A p.? Chr.1: 38867561-38867561 on Build GRCh38
136122 c.122C>T p.P41L Chr.1: 38867577-38867577 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs554420692 Chr.1: 38867362-38867362 on Build GRCh38
rs750959449 Chr.1: 38867422-38867422 on Build GRCh38
rs748050553 Chr.1: 38867525-38867525 on Build GRCh38
rs549971312 Chr.1: 38867378-38867378 on Build GRCh38
rs754479133 Chr.1: 38867520-38867520 on Build GRCh38
rs1018848474 Chr.1: 38867464-38867464 on Build GRCh38
rs993091987 Chr.1: 38867356-38867356 on Build GRCh38
rs775540156 Chr.1: 38867625-38867625 on Build GRCh38
rs958842312 Chr.1: 38867537-38867537 on Build GRCh38
rs772333895 Chr.1: 38867620-38867620 on Build GRCh38
rs1037639542 Chr.1: 38867177-38867177 on Build GRCh38
rs565744624 Chr.1: 38867289-38867289 on Build GRCh38
rs968972887 Chr.1: 38867400-38867400 on Build GRCh38
rs754215464 Chr.1: 38867511-38867512 on Build GRCh38
rs984291171 Chr.1: 38867346-38867346 on Build GRCh38
rs961884848 Chr.1: 38867524-38867524 on Build GRCh38
rs917528066 Chr.1: 38867610-38867610 on Build GRCh38
rs574581666 Chr.1: 38867370-38867370 on Build GRCh38
rs1027282848 Chr.1: 38867482-38867482 on Build GRCh38
rs200691739 Chr.1: 38867522-38867522 on Build GRCh38
rs867871471 Chr.1: 38867518-38867518 on Build GRCh38
rs879016174 Chr.1: 38867469-38867469 on Build GRCh38
rs946083225 Chr.1: 38867364-38867364 on Build GRCh38
rs942704893 Chr.1: 38867466-38867466 on Build GRCh38
rs1801753 Chr.1: 38867582-38867582 on Build GRCh38
rs1009255245 Chr.1: 38867463-38867463 on Build GRCh38
rs745682117 Chr.1: 38867164-38867164 on Build GRCh38
rs749073844 Chr.1: 38867609-38867609 on Build GRCh38
rs557266616 Chr.1: 38867569-38867569 on Build GRCh38
rs993194612 Chr.1: 38867468-38867468 on Build GRCh38
rs905944536 Chr.1: 38867328-38867328 on Build GRCh38
rs768574925 Chr.1: 38867300-38867300 on Build GRCh38
rs980509474 Chr.1: 38867502-38867502 on Build GRCh38
rs937610327 Chr.1: 38867350-38867350 on Build GRCh38
rs888948048 Chr.1: 38867429-38867429 on Build GRCh38
rs1003403133 Chr.1: 38867339-38867339 on Build GRCh38
rs1029117622 Chr.1: 38867434-38867434 on Build GRCh38
rs11338710 Chr.1: 38867447-38867447 on Build GRCh38
rs1027416819 Chr.1: 38867297-38867297 on Build GRCh38
rs192444125 Chr.1: 38867229-38867229 on Build GRCh38
rs562957210 Chr.1: 38867174-38867174 on Build GRCh38
rs961716350 Chr.1: 38867274-38867274 on Build GRCh38
rs367679547 Chr.1: 38867175-38867175 on Build GRCh38
rs1050258087 Chr.1: 38867426-38867426 on Build GRCh38
rs72925134 Chr.1: 38867345-38867345 on Build GRCh38
rs138907666 Chr.1: 38867465-38867465 on Build GRCh38
rs546987404 Chr.1: 38867428-38867428 on Build GRCh38
rs537611625 Chr.1: 38867273-38867273 on Build GRCh38
rs773059968 Chr.1: 38867600-38867600 on Build GRCh38
rs927411911 Chr.1: 38867348-38867348 on Build GRCh38
rs904619923 Chr.1: 38867404-38867404 on Build GRCh38
rs1044813555 Chr.1: 38867288-38867289 on Build GRCh38
rs1008811043 Chr.1: 38867234-38867234 on Build GRCh38
rs951783757 Chr.1: 38867499-38867499 on Build GRCh38
rs1000003932 Chr.1: 38867170-38867170 on Build GRCh38
rs936055162 Chr.1: 38867423-38867423 on Build GRCh38

View Full Product Details