Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 78806878-78807376 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
USP36
Entrez Gene ID
57602
Gene Name
ubiquitin specific peptidase 36
Gene Aliases
DUB1
UniGene
Hs.464243
Gene Chromosome Location
Chr.17: 78787381-78840898 on Build GRCh38
Transcripts
NM_001321291.1 XM_005257542.2 XM_005257543.2 XM_011525065.1 XM_011525066.2 XM_011525067.2 XM_011525068.1 XM_011525069.2 XM_011525070.1 XM_011525071.2 XM_017024900.1 XM_017024901.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4975029 c.1702A>C p.R568R Chr.17: 78807342-78807342 on Build GRCh38
6034376 c.1691C>A p.S564Y Chr.17: 78807353-78807353 on Build GRCh38
7204548 c.2052A>C p.P684P Chr.17: 78806992-78806992 on Build GRCh38
5607666 c.1829C>T p.S610F Chr.17: 78807215-78807215 on Build GRCh38
6818652 c.1782T>C p.H594H Chr.17: 78807262-78807262 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs370137888 Chr.17: 78807282-78807282 on Build GRCh38
rs774157603 Chr.17: 78806943-78806944 on Build GRCh38
rs776815586 Chr.17: 78806992-78806992 on Build GRCh38
rs374320288 Chr.17: 78807199-78807199 on Build GRCh38
rs746035501 Chr.17: 78807118-78807118 on Build GRCh38
rs550097094 Chr.17: 78807110-78807110 on Build GRCh38
rs2054623 Chr.17: 78807148-78807148 on Build GRCh38
rs369353557 Chr.17: 78807163-78807163 on Build GRCh38
rs543707885 Chr.17: 78807191-78807191 on Build GRCh38
rs753610612 Chr.17: 78807041-78807041 on Build GRCh38
rs376772768 Chr.17: 78807102-78807102 on Build GRCh38
rs759557030 Chr.17: 78807015-78807015 on Build GRCh38
rs201618229 Chr.17: 78806940-78806940 on Build GRCh38
rs996284931 Chr.17: 78807301-78807301 on Build GRCh38
rs947073273 Chr.17: 78807033-78807033 on Build GRCh38
rs529939411 Chr.17: 78807096-78807096 on Build GRCh38
rs543337655 Chr.17: 78807350-78807350 on Build GRCh38
rs949473129 Chr.17: 78807284-78807284 on Build GRCh38
rs372873518 Chr.17: 78807164-78807164 on Build GRCh38
rs780519181 Chr.17: 78807047-78807047 on Build GRCh38
rs777191172 Chr.17: 78807193-78807193 on Build GRCh38
rs139133507 Chr.17: 78807136-78807136 on Build GRCh38
rs776240175 Chr.17: 78807346-78807360 on Build GRCh38
rs769820188 Chr.17: 78806995-78806995 on Build GRCh38
rs770367607 Chr.17: 78807202-78807202 on Build GRCh38
rs1022825491 Chr.17: 78806901-78806901 on Build GRCh38
rs759180087 Chr.17: 78807207-78807207 on Build GRCh38
rs199764785 Chr.17: 78806930-78806930 on Build GRCh38
rs746990181 Chr.17: 78807128-78807128 on Build GRCh38
rs377312015 Chr.17: 78807311-78807311 on Build GRCh38
rs753373680 Chr.17: 78807089-78807089 on Build GRCh38
rs57732935 Chr.17: 78806926-78806926 on Build GRCh38
rs200774801 Chr.17: 78807083-78807083 on Build GRCh38
rs750482825 Chr.17: 78807171-78807171 on Build GRCh38
rs149420360 Chr.17: 78807005-78807005 on Build GRCh38
rs780446860 Chr.17: 78807180-78807180 on Build GRCh38
rs752631032 Chr.17: 78807031-78807031 on Build GRCh38
rs541169522 Chr.17: 78807263-78807263 on Build GRCh38
rs143666818 Chr.17: 78807085-78807085 on Build GRCh38
rs377551507 Chr.17: 78807238-78807238 on Build GRCh38
rs143765903 Chr.17: 78807086-78807086 on Build GRCh38
rs764090613 Chr.17: 78807143-78807143 on Build GRCh38
rs963361107 Chr.17: 78806908-78806908 on Build GRCh38
rs751671826 Chr.17: 78807273-78807273 on Build GRCh38
rs779770423 Chr.17: 78807332-78807332 on Build GRCh38
rs771893500 Chr.17: 78807019-78807020 on Build GRCh38
rs768686464 Chr.17: 78807129-78807129 on Build GRCh38
rs572232460 Chr.17: 78807071-78807071 on Build GRCh38
rs777775978 Chr.17: 78807240-78807240 on Build GRCh38
rs748888707 Chr.17: 78806949-78806949 on Build GRCh38
rs558732144 Chr.17: 78807243-78807243 on Build GRCh38
rs371093651 Chr.17: 78807345-78807345 on Build GRCh38
rs751840907 Chr.17: 78807049-78807049 on Build GRCh38
rs748850563 Chr.17: 78807196-78807196 on Build GRCh38
rs369603546 Chr.17: 78806978-78806978 on Build GRCh38
rs771680038 Chr.17: 78807254-78807254 on Build GRCh38
rs758623047 Chr.17: 78807151-78807151 on Build GRCh38
rs139827515 Chr.17: 78807223-78807223 on Build GRCh38
rs749397640 Chr.17: 78807113-78807113 on Build GRCh38
rs554608025 Chr.17: 78806937-78806937 on Build GRCh38
rs548197375 Chr.17: 78807167-78807167 on Build GRCh38
rs763096581 Chr.17: 78807002-78807002 on Build GRCh38
rs775098038 Chr.17: 78807137-78807137 on Build GRCh38
rs769023609 Chr.17: 78806957-78806957 on Build GRCh38
rs145100246 Chr.17: 78807053-78807053 on Build GRCh38
rs749294584 Chr.17: 78807103-78807103 on Build GRCh38
rs781572520 Chr.17: 78807058-78807058 on Build GRCh38
rs766105638 Chr.17: 78806931-78806931 on Build GRCh38
rs761599624 Chr.17: 78807091-78807091 on Build GRCh38
rs143223744 Chr.17: 78807155-78807155 on Build GRCh38
rs780275432 Chr.17: 78806951-78806951 on Build GRCh38
rs375010260 Chr.17: 78807297-78807297 on Build GRCh38
rs200900561 Chr.17: 78806948-78806948 on Build GRCh38
rs755813219 Chr.17: 78807239-78807239 on Build GRCh38
rs201302268 Chr.17: 78807184-78807184 on Build GRCh38
rs941372823 Chr.17: 78807131-78807131 on Build GRCh38
rs777030818 Chr.17: 78807218-78807218 on Build GRCh38
rs771024669 Chr.17: 78807081-78807081 on Build GRCh38
rs755939658 Chr.17: 78807289-78807289 on Build GRCh38
rs752888548 Chr.17: 78807028-78807028 on Build GRCh38
rs776672459 Chr.17: 78807130-78807130 on Build GRCh38
rs769067138 Chr.17: 78807217-78807217 on Build GRCh38
rs747063989 Chr.17: 78807032-78807032 on Build GRCh38
rs35820493 Chr.17: 78807181-78807181 on Build GRCh38
rs762666876 Chr.17: 78807138-78807138 on Build GRCh38
rs139422837 Chr.17: 78807165-78807165 on Build GRCh38
rs746133689 Chr.17: 78807150-78807150 on Build GRCh38
rs186001931 Chr.17: 78807158-78807158 on Build GRCh38
rs764603428 Chr.17: 78807142-78807142 on Build GRCh38
rs763626403 Chr.17: 78807039-78807039 on Build GRCh38
rs771827935 Chr.17: 78807124-78807124 on Build GRCh38
rs773993759 Chr.17: 78807204-78807204 on Build GRCh38
rs760270597 Chr.17: 78807034-78807034 on Build GRCh38
rs767385597 Chr.17: 78807020-78807020 on Build GRCh38
rs772769833 Chr.17: 78807161-78807161 on Build GRCh38
rs773436494 Chr.17: 78806998-78806998 on Build GRCh38
rs759880106 Chr.17: 78807276-78807276 on Build GRCh38
rs758371951 Chr.17: 78807172-78807172 on Build GRCh38
rs574805840 Chr.17: 78807325-78807325 on Build GRCh38
rs773387266 Chr.17: 78806928-78806928 on Build GRCh38
rs751898095 Chr.17: 78807229-78807229 on Build GRCh38
rs143161068 Chr.17: 78807246-78807246 on Build GRCh38
rs372219044 Chr.17: 78807082-78807082 on Build GRCh38
rs766627365 Chr.17: 78807268-78807268 on Build GRCh38
rs763515271 Chr.17: 78807193-78807199 on Build GRCh38
rs753827258 Chr.17: 78807305-78807305 on Build GRCh38
rs777422484 Chr.17: 78807303-78807303 on Build GRCh38
rs368958421 Chr.17: 78807116-78807116 on Build GRCh38
rs770828514 Chr.17: 78807003-78807003 on Build GRCh38
rs755243023 Chr.17: 78807231-78807231 on Build GRCh38
rs748601527 Chr.17: 78807059-78807059 on Build GRCh38
rs760314946 Chr.17: 78807190-78807216 on Build GRCh38
rs372368137 Chr.17: 78806944-78806944 on Build GRCh38
rs572169318 Chr.17: 78807244-78807244 on Build GRCh38
rs747607736 Chr.17: 78806952-78806952 on Build GRCh38
rs775059509 Chr.17: 78807102-78807103 on Build GRCh38
rs139155069 Chr.17: 78807267-78807267 on Build GRCh38
rs200626859 Chr.17: 78807237-78807237 on Build GRCh38
rs745699572 Chr.17: 78806959-78806960 on Build GRCh38
rs757285017 Chr.17: 78807144-78807144 on Build GRCh38
rs781557175 Chr.17: 78807101-78807101 on Build GRCh38
rs576677638 Chr.17: 78807228-78807228 on Build GRCh38
rs150865699 Chr.17: 78807156-78807156 on Build GRCh38
rs973787935 Chr.17: 78806920-78806920 on Build GRCh38
rs763015564 Chr.17: 78807141-78807141 on Build GRCh38
rs146706684 Chr.17: 78807247-78807247 on Build GRCh38
rs767930328 Chr.17: 78807139-78807139 on Build GRCh38
rs200029537 Chr.17: 78806936-78806936 on Build GRCh38
rs144381842 Chr.17: 78807186-78807186 on Build GRCh38
rs368071453 Chr.17: 78806939-78806939 on Build GRCh38
rs757163190 Chr.17: 78807309-78807309 on Build GRCh38
rs762262292 Chr.17: 78807262-78807262 on Build GRCh38
rs149099006 Chr.17: 78807063-78807063 on Build GRCh38
rs371268256 Chr.17: 78807190-78807190 on Build GRCh38
rs143816156 Chr.17: 78807097-78807097 on Build GRCh38
rs3744796 Chr.17: 78807115-78807115 on Build GRCh38
rs1043080166 Chr.17: 78807035-78807035 on Build GRCh38
rs377142544 Chr.17: 78806950-78806950 on Build GRCh38
rs767814555 Chr.17: 78807281-78807281 on Build GRCh38
rs866304291 Chr.17: 78807222-78807222 on Build GRCh38
rs757795848 Chr.17: 78807329-78807329 on Build GRCh38
rs775411653 Chr.17: 78807125-78807125 on Build GRCh38
rs754369153 Chr.17: 78807166-78807166 on Build GRCh38
rs762265490 Chr.17: 78807219-78807219 on Build GRCh38
rs765281175 Chr.17: 78807168-78807168 on Build GRCh38
rs774965047 Chr.17: 78807258-78807258 on Build GRCh38
rs376513018 Chr.17: 78807140-78807140 on Build GRCh38
rs750848663 Chr.17: 78807221-78807221 on Build GRCh38
rs556848402 Chr.17: 78807220-78807220 on Build GRCh38
rs112843316 Chr.17: 78807153-78807153 on Build GRCh38
rs1052400393 Chr.17: 78806958-78806958 on Build GRCh38
rs756955437 Chr.17: 78807043-78807043 on Build GRCh38
rs1022923099 Chr.17: 78807188-78807188 on Build GRCh38
rs773774964 Chr.17: 78807265-78807265 on Build GRCh38
rs755645646 Chr.17: 78806947-78806947 on Build GRCh38

View Full Product Details