Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 81912244-81912736 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SIRT7
Entrez Gene ID
51547
Gene Name
sirtuin 7
Gene Aliases
SIR2L7
UniGene
Hs.514636
Gene Chromosome Location
Chr.17: 81911939-81918182 on Build GRCh38
Transcripts
NM_016538.2 XM_006722284.2 XM_011523580.1 XM_011523581.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4819721 c.1191G>C p.K397N Chr.17: 81912428-81912428 on Build GRCh38
3989430 c.1170C>T p.C390C Chr.17: 81912449-81912449 on Build GRCh38
4070754 c.1135G>A p.A379T Chr.17: 81912484-81912484 on Build GRCh38
6882047 c.1032G>A p.A344A Chr.17: 81912587-81912587 on Build GRCh38
1387361 c.1185A>C p.K395N Chr.17: 81912434-81912434 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766936470 Chr.17: 81912390-81912390 on Build GRCh38
rs750240504 Chr.17: 81912406-81912406 on Build GRCh38
rs758534175 Chr.17: 81912494-81912494 on Build GRCh38
rs748325161 Chr.17: 81912491-81912491 on Build GRCh38
rs145465834 Chr.17: 81912441-81912441 on Build GRCh38
rs377589552 Chr.17: 81912642-81912642 on Build GRCh38
rs1030629974 Chr.17: 81912359-81912359 on Build GRCh38
rs572701775 Chr.17: 81912476-81912476 on Build GRCh38
rs137989863 Chr.17: 81912442-81912442 on Build GRCh38
rs374568755 Chr.17: 81912419-81912419 on Build GRCh38
rs142687743 Chr.17: 81912458-81912458 on Build GRCh38
rs756959045 Chr.17: 81912404-81912404 on Build GRCh38
rs755947334 Chr.17: 81912328-81912328 on Build GRCh38
rs200734338 Chr.17: 81912549-81912549 on Build GRCh38
rs199923657 Chr.17: 81912611-81912611 on Build GRCh38
rs771558317 Chr.17: 81912596-81912596 on Build GRCh38
rs546577390 Chr.17: 81912301-81912301 on Build GRCh38
rs144915617 Chr.17: 81912501-81912501 on Build GRCh38
rs749929943 Chr.17: 81912397-81912399 on Build GRCh38
rs761159003 Chr.17: 81912424-81912424 on Build GRCh38
rs943504855 Chr.17: 81912311-81912311 on Build GRCh38
rs753208398 Chr.17: 81912570-81912571 on Build GRCh38
rs993349799 Chr.17: 81912672-81912672 on Build GRCh38
rs772273434 Chr.17: 81912380-81912380 on Build GRCh38
rs370966987 Chr.17: 81912643-81912643 on Build GRCh38
rs141859850 Chr.17: 81912516-81912516 on Build GRCh38
rs753687472 Chr.17: 81912543-81912543 on Build GRCh38
rs758544996 Chr.17: 81912562-81912562 on Build GRCh38
rs929927275 Chr.17: 81912408-81912408 on Build GRCh38
rs770771970 Chr.17: 81912614-81912614 on Build GRCh38
rs1026617544 Chr.17: 81912696-81912696 on Build GRCh38
rs746318408 Chr.17: 81912507-81912507 on Build GRCh38
rs893512619 Chr.17: 81912579-81912579 on Build GRCh38
rs766215616 Chr.17: 81912542-81912542 on Build GRCh38
rs761018740 Chr.17: 81912388-81912388 on Build GRCh38
rs999073966 Chr.17: 81912525-81912525 on Build GRCh38
rs756003672 Chr.17: 81912407-81912407 on Build GRCh38
rs752523205 Chr.17: 81912327-81912327 on Build GRCh38
rs370769969 Chr.17: 81912373-81912373 on Build GRCh38
rs1030349590 Chr.17: 81912267-81912267 on Build GRCh38
rs772653671 Chr.17: 81912697-81912697 on Build GRCh38
rs200932386 Chr.17: 81912505-81912505 on Build GRCh38
rs536317348 Chr.17: 81912420-81912420 on Build GRCh38
rs756185592 Chr.17: 81912421-81912422 on Build GRCh38
rs1010216455 Chr.17: 81912344-81912344 on Build GRCh38
rs768102958 Chr.17: 81912567-81912568 on Build GRCh38
rs763548264 Chr.17: 81912452-81912452 on Build GRCh38
rs200017321 Chr.17: 81912497-81912497 on Build GRCh38
rs781471662 Chr.17: 81912577-81912577 on Build GRCh38
rs199664629 Chr.17: 81912526-81912526 on Build GRCh38
rs373659511 Chr.17: 81912633-81912633 on Build GRCh38
rs898967783 Chr.17: 81912663-81912663 on Build GRCh38
rs759452277 Chr.17: 81912626-81912626 on Build GRCh38
rs201850684 Chr.17: 81912550-81912550 on Build GRCh38
rs571463876 Chr.17: 81912342-81912342 on Build GRCh38
rs752640962 Chr.17: 81912630-81912630 on Build GRCh38
rs1042673606 Chr.17: 81912348-81912348 on Build GRCh38
rs752696094 Chr.17: 81912553-81912553 on Build GRCh38
rs1039847528 Chr.17: 81912468-81912468 on Build GRCh38
rs773443530 Chr.17: 81912423-81912423 on Build GRCh38
rs749450016 Chr.17: 81912365-81912365 on Build GRCh38
rs755990757 Chr.17: 81912479-81912479 on Build GRCh38
rs764714847 Chr.17: 81912370-81912371 on Build GRCh38
rs113876228 Chr.17: 81912360-81912360 on Build GRCh38
rs772607546 Chr.17: 81912606-81912606 on Build GRCh38
rs776552077 Chr.17: 81912620-81912620 on Build GRCh38
rs370911311 Chr.17: 81912411-81912411 on Build GRCh38
rs768811478 Chr.17: 81912418-81912418 on Build GRCh38
rs757485586 Chr.17: 81912576-81912576 on Build GRCh38
rs761308620 Chr.17: 81912460-81912460 on Build GRCh38
rs760003418 Chr.17: 81912492-81912493 on Build GRCh38
rs747584480 Chr.17: 81912588-81912588 on Build GRCh38
rs1053873050 Chr.17: 81912319-81912319 on Build GRCh38
rs910302887 Chr.17: 81912305-81912305 on Build GRCh38
rs201056584 Chr.17: 81912584-81912584 on Build GRCh38
rs989214357 Chr.17: 81912548-81912548 on Build GRCh38
rs762497990 Chr.17: 81912391-81912391 on Build GRCh38
rs74006134 Chr.17: 81912396-81912396 on Build GRCh38
rs201329944 Chr.17: 81912587-81912587 on Build GRCh38
rs770275784 Chr.17: 81912583-81912583 on Build GRCh38
rs200045972 Chr.17: 81912552-81912552 on Build GRCh38
rs773731851 Chr.17: 81912515-81912515 on Build GRCh38
rs868565477 Chr.17: 81912604-81912604 on Build GRCh38
rs375370955 Chr.17: 81912394-81912394 on Build GRCh38
rs982244545 Chr.17: 81912625-81912625 on Build GRCh38
rs544376973 Chr.17: 81912585-81912585 on Build GRCh38
rs1005624018 Chr.17: 81912654-81912654 on Build GRCh38
rs766305128 Chr.17: 81912485-81912485 on Build GRCh38
rs773636284 Chr.17: 81912385-81912385 on Build GRCh38
rs756561304 Chr.17: 81912602-81912603 on Build GRCh38
rs774751105 Chr.17: 81912371-81912371 on Build GRCh38
rs148268197 Chr.17: 81912504-81912504 on Build GRCh38
rs746304894 Chr.17: 81912517-81912517 on Build GRCh38
rs754975676 Chr.17: 81912489-81912489 on Build GRCh38
rs200648674 Chr.17: 81912366-81912366 on Build GRCh38
rs367621203 Chr.17: 81912495-81912495 on Build GRCh38
rs969179861 Chr.17: 81912624-81912624 on Build GRCh38
rs199736516 Chr.17: 81912280-81912280 on Build GRCh38
rs747205915 Chr.17: 81912573-81912573 on Build GRCh38
rs201807645 Chr.17: 81912524-81912524 on Build GRCh38
rs200202099 Chr.17: 81912632-81912632 on Build GRCh38
rs770292119 Chr.17: 81912512-81912512 on Build GRCh38
rs560581700 Chr.17: 81912673-81912673 on Build GRCh38
rs772341390 Chr.17: 81912422-81912422 on Build GRCh38
rs767270496 Chr.17: 81912462-81912462 on Build GRCh38
rs374253740 Chr.17: 81912660-81912660 on Build GRCh38
rs566714354 Chr.17: 81912278-81912278 on Build GRCh38
rs762123132 Chr.17: 81912376-81912376 on Build GRCh38
rs756545371 Chr.17: 81912644-81912644 on Build GRCh38
rs534922646 Chr.17: 81912287-81912287 on Build GRCh38
rs753775333 Chr.17: 81912486-81912486 on Build GRCh38
rs777746632 Chr.17: 81912566-81912566 on Build GRCh38
rs780794595 Chr.17: 81912405-81912405 on Build GRCh38
rs780318421 Chr.17: 81912659-81912659 on Build GRCh38
rs200718529 Chr.17: 81912412-81912412 on Build GRCh38
rs760421757 Chr.17: 81912536-81912536 on Build GRCh38
rs369349540 Chr.17: 81912627-81912627 on Build GRCh38
rs751643965 Chr.17: 81912634-81912634 on Build GRCh38
rs1051435780 Chr.17: 81912298-81912298 on Build GRCh38

View Full Product Details