Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 81936628-81937123 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PYCR1
Entrez Gene ID
5831
Gene Name
pyrroline-5-carboxylate reductase 1
Gene Aliases
ARCL2B, ARCL3B, P5C, P5CR, PIG45, PP222, PRO3, PYCR
UniGene
Hs.163451
Gene Chromosome Location
Chr.17: 81932384-81937328 on Build GRCh38
Transcripts
NM_001282279.1 NM_001282280.1 NM_001282281.1 NM_006907.3 NM_153824.2 XM_005256381.2 XM_011523583.2 XM_011523584.2 XM_011523585.2 XM_017024909.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4657176 c.27C>T p.G9G Chr.17: 81936788-81936788 on Build GRCh38
4273330 c.61G>A p.A21T Chr.17: 81936754-81936754 on Build GRCh38
4273331 c.61G>A p.A21T Chr.17: 81936754-81936754 on Build GRCh38
473549 c.51G>C p.K17N Chr.17: 81936764-81936764 on Build GRCh38
4070761 c.37T>G p.F13V Chr.17: 81936778-81936778 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs546125979 Chr.17: 81936776-81936776 on Build GRCh38
rs886053572 Chr.17: 81937064-81937064 on Build GRCh38
rs774943551 Chr.17: 81936843-81936843 on Build GRCh38
rs776837667 Chr.17: 81936741-81936741 on Build GRCh38
rs966497358 Chr.17: 81936920-81936920 on Build GRCh38
rs746066425 Chr.17: 81936818-81936818 on Build GRCh38
rs181969864 Chr.17: 81936720-81936720 on Build GRCh38
rs916215795 Chr.17: 81936982-81936982 on Build GRCh38
rs769150356 Chr.17: 81936842-81936842 on Build GRCh38
rs992837529 Chr.17: 81936907-81936907 on Build GRCh38
rs908763906 Chr.17: 81936942-81936942 on Build GRCh38
rs369725098 Chr.17: 81936744-81936744 on Build GRCh38
rs573221309 Chr.17: 81936876-81936876 on Build GRCh38
rs764755605 Chr.17: 81936866-81936866 on Build GRCh38
rs769954957 Chr.17: 81936825-81936825 on Build GRCh38
rs990230369 Chr.17: 81936902-81936902 on Build GRCh38
rs916901720 Chr.17: 81936692-81936692 on Build GRCh38
rs977920507 Chr.17: 81936921-81936921 on Build GRCh38
rs944515240 Chr.17: 81937044-81937044 on Build GRCh38
rs757343922 Chr.17: 81936805-81936805 on Build GRCh38
rs566655599 Chr.17: 81937060-81937060 on Build GRCh38
rs1005701798 Chr.17: 81936768-81936768 on Build GRCh38
rs765824328 Chr.17: 81936763-81936763 on Build GRCh38
rs760742876 Chr.17: 81936928-81936928 on Build GRCh38
rs780349588 Chr.17: 81936828-81936828 on Build GRCh38
rs962042256 Chr.17: 81936950-81936950 on Build GRCh38
rs770472952 Chr.17: 81936852-81936852 on Build GRCh38
rs200631132 Chr.17: 81936653-81936653 on Build GRCh38
rs529286442 Chr.17: 81937003-81937003 on Build GRCh38
rs779525010 Chr.17: 81936738-81936738 on Build GRCh38
rs771001446 Chr.17: 81936698-81936698 on Build GRCh38
rs994257139 Chr.17: 81937091-81937091 on Build GRCh38
rs1055081206 Chr.17: 81936979-81936979 on Build GRCh38
rs939276832 Chr.17: 81936953-81936953 on Build GRCh38
rs752929017 Chr.17: 81936660-81936660 on Build GRCh38
rs531874252 Chr.17: 81936863-81936863 on Build GRCh38
rs373386140 Chr.17: 81936750-81936750 on Build GRCh38
rs920343814 Chr.17: 81936716-81936716 on Build GRCh38
rs1008772309 Chr.17: 81936771-81936771 on Build GRCh38
rs763934259 Chr.17: 81936894-81936894 on Build GRCh38
rs1036533568 Chr.17: 81936695-81936695 on Build GRCh38
rs746131639 Chr.17: 81936725-81936725 on Build GRCh38
rs781193717 Chr.17: 81936810-81936810 on Build GRCh38
rs11555729 Chr.17: 81936829-81936829 on Build GRCh38
rs886053571 Chr.17: 81936891-81936891 on Build GRCh38
rs564658518 Chr.17: 81936793-81936793 on Build GRCh38
rs759116585 Chr.17: 81936767-81936767 on Build GRCh38
rs376495113 Chr.17: 81936862-81936862 on Build GRCh38
rs913353419 Chr.17: 81936915-81936915 on Build GRCh38
rs770188778 Chr.17: 81936732-81936732 on Build GRCh38
rs992374892 Chr.17: 81936674-81936674 on Build GRCh38
rs994330378 Chr.17: 81936835-81936835 on Build GRCh38
rs762470089 Chr.17: 81936851-81936851 on Build GRCh38
rs751510308 Chr.17: 81936798-81936798 on Build GRCh38
rs752349361 Chr.17: 81936872-81936872 on Build GRCh38
rs939573540 Chr.17: 81936662-81936662 on Build GRCh38
rs547728412 Chr.17: 81937028-81937028 on Build GRCh38
rs897329315 Chr.17: 81936808-81936808 on Build GRCh38
rs1050097226 Chr.17: 81937009-81937009 on Build GRCh38
rs927872423 Chr.17: 81936680-81936680 on Build GRCh38
rs771647093 Chr.17: 81936729-81936729 on Build GRCh38
rs930404113 Chr.17: 81936988-81936988 on Build GRCh38
rs940343566 Chr.17: 81936694-81936694 on Build GRCh38
rs759180800 Chr.17: 81936775-81936775 on Build GRCh38
rs960823609 Chr.17: 81936886-81936886 on Build GRCh38
rs775754588 Chr.17: 81936735-81936735 on Build GRCh38
rs764095488 Chr.17: 81937010-81937010 on Build GRCh38
rs749634868 Chr.17: 81936838-81936838 on Build GRCh38
rs1038077634 Chr.17: 81937048-81937048 on Build GRCh38
rs762218403 Chr.17: 81936755-81936756 on Build GRCh38
rs760198939 Chr.17: 81936755-81936755 on Build GRCh38
rs927877868 Chr.17: 81936935-81936935 on Build GRCh38
rs568538158 Chr.17: 81936973-81936973 on Build GRCh38
rs560639658 Chr.17: 81936672-81936672 on Build GRCh38

View Full Product Details