Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 81956805-81957298 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NOTUM
Entrez Gene ID
147111
Gene Name
NOTUM, palmitoleoyl-protein carboxylesterase
Gene Aliases
hNOTUM
UniGene
Hs.106137
Gene Chromosome Location
Chr.17: 81952507-81961181 on Build GRCh38
Transcripts
NM_178493.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7173798 c.836G>A p.C279Y Chr.17: 81956934-81956934 on Build GRCh38
7288686 c.874C>T p.R292C Chr.17: 81956896-81956896 on Build GRCh38
1589298 c.846G>A p.T282T Chr.17: 81956924-81956924 on Build GRCh38
5741195 c.856G>A p.A286T Chr.17: 81956914-81956914 on Build GRCh38
3680617 c.725T>G p.V242G Chr.17: 81957045-81957045 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757067481 Chr.17: 81956955-81956957 on Build GRCh38
rs61734818 Chr.17: 81956861-81956861 on Build GRCh38
rs753265012 Chr.17: 81956945-81956945 on Build GRCh38
rs755305853 Chr.17: 81956938-81956938 on Build GRCh38
rs372143531 Chr.17: 81957205-81957205 on Build GRCh38
rs138384810 Chr.17: 81956919-81956919 on Build GRCh38
rs754192539 Chr.17: 81956934-81956934 on Build GRCh38
rs773379510 Chr.17: 81957015-81957015 on Build GRCh38
rs1037598998 Chr.17: 81957164-81957164 on Build GRCh38
rs915735201 Chr.17: 81957162-81957162 on Build GRCh38
rs954853719 Chr.17: 81957195-81957195 on Build GRCh38
rs529502823 Chr.17: 81957182-81957182 on Build GRCh38
rs773537242 Chr.17: 81956930-81956930 on Build GRCh38
rs754280791 Chr.17: 81956893-81956893 on Build GRCh38
rs948426890 Chr.17: 81957131-81957131 on Build GRCh38
rs766559254 Chr.17: 81957029-81957029 on Build GRCh38
rs557716494 Chr.17: 81956932-81956932 on Build GRCh38
rs780726135 Chr.17: 81956954-81956954 on Build GRCh38
rs928062399 Chr.17: 81957145-81957145 on Build GRCh38
rs978115395 Chr.17: 81957122-81957122 on Build GRCh38
rs568695867 Chr.17: 81957163-81957163 on Build GRCh38
rs766534260 Chr.17: 81956933-81956933 on Build GRCh38
rs556378468 Chr.17: 81956992-81956992 on Build GRCh38
rs61734814 Chr.17: 81956853-81956853 on Build GRCh38
rs532025424 Chr.17: 81956872-81956872 on Build GRCh38
rs557802527 Chr.17: 81957225-81957225 on Build GRCh38
rs376673453 Chr.17: 81957056-81957056 on Build GRCh38
rs531304548 Chr.17: 81957153-81957153 on Build GRCh38
rs1039168984 Chr.17: 81957208-81957208 on Build GRCh38
rs201191460 Chr.17: 81956874-81956874 on Build GRCh38
rs1017844595 Chr.17: 81956868-81956868 on Build GRCh38
rs772096685 Chr.17: 81957098-81957098 on Build GRCh38
rs566077432 Chr.17: 81957197-81957197 on Build GRCh38
rs200463470 Chr.17: 81957093-81957093 on Build GRCh38
rs928962668 Chr.17: 81957170-81957170 on Build GRCh38
rs199691879 Chr.17: 81956925-81956925 on Build GRCh38
rs748974956 Chr.17: 81957274-81957274 on Build GRCh38
rs547949533 Chr.17: 81957194-81957194 on Build GRCh38
rs373426796 Chr.17: 81956879-81956879 on Build GRCh38
rs966984207 Chr.17: 81957232-81957232 on Build GRCh38
rs772257747 Chr.17: 81956997-81956997 on Build GRCh38
rs1011140184 Chr.17: 81957211-81957211 on Build GRCh38
rs974797078 Chr.17: 81957171-81957171 on Build GRCh38
rs556227446 Chr.17: 81957241-81957241 on Build GRCh38
rs975118874 Chr.17: 81956965-81956965 on Build GRCh38
rs755749056 Chr.17: 81956966-81956966 on Build GRCh38
rs1042928953 Chr.17: 81956838-81956838 on Build GRCh38
rs764613659 Chr.17: 81957081-81957081 on Build GRCh38
rs753177230 Chr.17: 81957061-81957061 on Build GRCh38
rs771453758 Chr.17: 81956832-81956832 on Build GRCh38
rs746527391 Chr.17: 81956850-81956850 on Build GRCh38
rs776734815 Chr.17: 81957043-81957043 on Build GRCh38
rs923395838 Chr.17: 81957039-81957039 on Build GRCh38
rs770445485 Chr.17: 81956902-81956902 on Build GRCh38
rs371282489 Chr.17: 81957071-81957071 on Build GRCh38
rs1009065647 Chr.17: 81957186-81957186 on Build GRCh38
rs747056445 Chr.17: 81957107-81957107 on Build GRCh38
rs746451620 Chr.17: 81956896-81956896 on Build GRCh38
rs144388935 Chr.17: 81957238-81957238 on Build GRCh38
rs542000601 Chr.17: 81957089-81957089 on Build GRCh38
rs768910732 Chr.17: 81957160-81957160 on Build GRCh38
rs745444193 Chr.17: 81957127-81957127 on Build GRCh38
rs763958171 Chr.17: 81956873-81956873 on Build GRCh38
rs377172933 Chr.17: 81956906-81956906 on Build GRCh38
rs778994214 Chr.17: 81956895-81956895 on Build GRCh38
rs920537483 Chr.17: 81957165-81957165 on Build GRCh38
rs370368298 Chr.17: 81956914-81956914 on Build GRCh38
rs777392133 Chr.17: 81956836-81956836 on Build GRCh38
rs749866770 Chr.17: 81957087-81957087 on Build GRCh38
rs539561720 Chr.17: 81957209-81957209 on Build GRCh38
rs754019140 Chr.17: 81956862-81956862 on Build GRCh38
rs923313913 Chr.17: 81956830-81956830 on Build GRCh38
rs745402057 Chr.17: 81956957-81956957 on Build GRCh38
rs780812322 Chr.17: 81956903-81956903 on Build GRCh38
rs569720179 Chr.17: 81957230-81957230 on Build GRCh38
rs538054735 Chr.17: 81957095-81957095 on Build GRCh38
rs1044435347 Chr.17: 81957152-81957152 on Build GRCh38
rs34600945 Chr.17: 81957159-81957159 on Build GRCh38
rs749052839 Chr.17: 81956974-81956974 on Build GRCh38
rs779741644 Chr.17: 81957092-81957092 on Build GRCh38
rs374056429 Chr.17: 81956852-81956852 on Build GRCh38
rs753717722 Chr.17: 81956838-81956838 on Build GRCh38
rs774426016 Chr.17: 81956991-81956991 on Build GRCh38
rs769640604 Chr.17: 81956856-81956856 on Build GRCh38
rs199890100 Chr.17: 81956912-81956912 on Build GRCh38
rs776911948 Chr.17: 81957114-81957114 on Build GRCh38
rs139006803 Chr.17: 81957044-81957044 on Build GRCh38
rs780508873 Chr.17: 81956892-81956892 on Build GRCh38
rs201514031 Chr.17: 81957104-81957104 on Build GRCh38
rs993181736 Chr.17: 81956956-81956956 on Build GRCh38
rs564084591 Chr.17: 81957111-81957111 on Build GRCh38
rs201703462 Chr.17: 81956855-81956855 on Build GRCh38
rs144787172 Chr.17: 81957094-81957094 on Build GRCh38
rs756544524 Chr.17: 81956884-81956884 on Build GRCh38
rs150457980 Chr.17: 81956915-81956915 on Build GRCh38
rs544121637 Chr.17: 81956972-81956972 on Build GRCh38
rs768393795 Chr.17: 81956975-81956975 on Build GRCh38
rs890211185 Chr.17: 81957183-81957183 on Build GRCh38
rs757277733 Chr.17: 81956878-81956878 on Build GRCh38
rs555429654 Chr.17: 81956877-81956877 on Build GRCh38
rs778719752 Chr.17: 81957065-81957066 on Build GRCh38
rs145177151 Chr.17: 81957007-81957007 on Build GRCh38
rs1026002898 Chr.17: 81956987-81956987 on Build GRCh38
rs143082560 Chr.17: 81956946-81956946 on Build GRCh38
rs774167126 Chr.17: 81956918-81956918 on Build GRCh38
rs539503596 Chr.17: 81956865-81956865 on Build GRCh38
rs760869515 Chr.17: 81957022-81957022 on Build GRCh38
rs373383949 Chr.17: 81956940-81956940 on Build GRCh38

View Full Product Details