Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 722767-723315 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
YES1
Entrez Gene ID
7525
Gene Name
YES proto-oncogene 1, Src family tyrosine kinase
Gene Aliases
HsT441, P61-YES, Yes, c-yes
UniGene
Hs.194148
Gene Chromosome Location
Chr.18: 721592-812326 on Build GRCh38
Transcripts
NM_005433.3 XM_005258139.3 XM_017025960.1

SNP Detail

SNP ID Chromosome Location
rs764109167 Chr.18: 722869-722869 on Build GRCh38
rs1033690800 Chr.18: 723091-723091 on Build GRCh38
rs540771883 Chr.18: 723005-723005 on Build GRCh38
rs1014076074 Chr.18: 722984-722984 on Build GRCh38
rs919319285 Chr.18: 723248-723248 on Build GRCh38
rs398120089 Chr.18: 723111-723112 on Build GRCh38
rs528698337 Chr.18: 722997-722997 on Build GRCh38
rs548132166 Chr.18: 723264-723264 on Build GRCh38
rs1041275294 Chr.18: 722904-722904 on Build GRCh38
rs17551234 Chr.18: 723043-723043 on Build GRCh38
rs901514670 Chr.18: 723075-723075 on Build GRCh38
rs577967374 Chr.18: 722815-722815 on Build GRCh38
rs574463045 Chr.18: 722985-722985 on Build GRCh38
rs571835660 Chr.18: 722937-722937 on Build GRCh38
rs561493837 Chr.18: 722908-722908 on Build GRCh38
rs17516244 Chr.18: 723104-723104 on Build GRCh38
rs554432782 Chr.18: 722960-722960 on Build GRCh38
rs925979917 Chr.18: 723037-723037 on Build GRCh38
rs527885552 Chr.18: 722863-722863 on Build GRCh38
rs868013768 Chr.18: 723053-723053 on Build GRCh38
rs1007205322 Chr.18: 723050-723050 on Build GRCh38
rs17551227 Chr.18: 722831-722831 on Build GRCh38
rs957617586 Chr.18: 723059-723059 on Build GRCh38
rs903384145 Chr.18: 723086-723086 on Build GRCh38
rs1024016518 Chr.18: 722898-722898 on Build GRCh38
rs370963250 Chr.18: 722943-722943 on Build GRCh38
rs187036941 Chr.18: 723201-723201 on Build GRCh38
rs564942 Chr.18: 722868-722868 on Build GRCh38
rs181660148 Chr.18: 723054-723054 on Build GRCh38
rs386800142 Chr.18: 722917-722918 on Build GRCh38
rs143342112 Chr.18: 722820-722820 on Build GRCh38
rs181229276 Chr.18: 722903-722903 on Build GRCh38
rs565522812 Chr.18: 723079-723079 on Build GRCh38
rs931458413 Chr.18: 722883-722883 on Build GRCh38
rs936990898 Chr.18: 722825-722825 on Build GRCh38
rs538987264 Chr.18: 722872-722872 on Build GRCh38
rs17516223 Chr.18: 723024-723024 on Build GRCh38
rs529827145 Chr.18: 722917-722917 on Build GRCh38
rs377417798 Chr.18: 722909-722909 on Build GRCh38
rs142007417 Chr.18: 723103-723104 on Build GRCh38
rs192320794 Chr.18: 723234-723234 on Build GRCh38
rs755156149 Chr.18: 722916-722916 on Build GRCh38
rs984607196 Chr.18: 723258-723258 on Build GRCh38
rs964200027 Chr.18: 723020-723020 on Build GRCh38
rs189358268 Chr.18: 722888-722888 on Build GRCh38
rs376355783 Chr.18: 722889-722889 on Build GRCh38
rs550861448 Chr.18: 723028-723028 on Build GRCh38
rs890009068 Chr.18: 722887-722887 on Build GRCh38
rs541978776 Chr.18: 722988-722988 on Build GRCh38
rs492056 Chr.18: 722894-722894 on Build GRCh38
rs545468322 Chr.18: 723047-723047 on Build GRCh38
rs965402064 Chr.18: 722845-722845 on Build GRCh38
rs779256213 Chr.18: 723144-723145 on Build GRCh38
rs146312589 Chr.18: 722900-722900 on Build GRCh38
rs17551241 Chr.18: 723048-723048 on Build GRCh38
rs566585837 Chr.18: 722952-722952 on Build GRCh38
rs184292401 Chr.18: 722906-722906 on Build GRCh38
rs976861205 Chr.18: 723022-723022 on Build GRCh38
rs12961275 Chr.18: 723072-723072 on Build GRCh38
rs1049470949 Chr.18: 723071-723071 on Build GRCh38
rs560253740 Chr.18: 722995-722995 on Build GRCh38
rs927014109 Chr.18: 722824-722824 on Build GRCh38
rs568904944 Chr.18: 722929-722929 on Build GRCh38
rs572763063 Chr.18: 722829-722829 on Build GRCh38
rs566929055 Chr.18: 723040-723040 on Build GRCh38
rs762569020 Chr.18: 722934-722934 on Build GRCh38
rs562572795 Chr.18: 723032-723032 on Build GRCh38
rs991898831 Chr.18: 723174-723174 on Build GRCh38
rs547886723 Chr.18: 723058-723058 on Build GRCh38
rs571808218 Chr.18: 722858-722858 on Build GRCh38
rs58178670 Chr.18: 722914-722914 on Build GRCh38
rs961729018 Chr.18: 722930-722930 on Build GRCh38
rs368125075 Chr.18: 723016-723016 on Build GRCh38
rs1010029365 Chr.18: 722850-722850 on Build GRCh38
rs931515152 Chr.18: 722902-722902 on Build GRCh38
rs1007282671 Chr.18: 722837-722837 on Build GRCh38
rs554623590 Chr.18: 722949-722949 on Build GRCh38
rs1060910 Chr.18: 722830-722830 on Build GRCh38
rs373122756 Chr.18: 722897-722897 on Build GRCh38
rs981549713 Chr.18: 723242-723242 on Build GRCh38
rs17551213 Chr.18: 722819-722819 on Build GRCh38
rs569851260 Chr.18: 723052-723052 on Build GRCh38
rs386800144 Chr.18: 723069-723070 on Build GRCh38
rs146477038 Chr.18: 723038-723038 on Build GRCh38
rs188975592 Chr.18: 722965-722965 on Build GRCh38
rs950073028 Chr.18: 723064-723064 on Build GRCh38
rs115311992 Chr.18: 723263-723263 on Build GRCh38
rs376960673 Chr.18: 722864-722864 on Build GRCh38
rs958417425 Chr.18: 722865-722865 on Build GRCh38
rs949875986 Chr.18: 723041-723041 on Build GRCh38
rs147567387 Chr.18: 723130-723130 on Build GRCh38
rs929145912 Chr.18: 723255-723255 on Build GRCh38
rs535025235 Chr.18: 722809-722809 on Build GRCh38
rs540188109 Chr.18: 722891-722891 on Build GRCh38
rs562808703 Chr.18: 723244-723244 on Build GRCh38
rs985898591 Chr.18: 722945-722945 on Build GRCh38
rs547247955 Chr.18: 722867-722867 on Build GRCh38
rs999009000 Chr.18: 723087-723087 on Build GRCh38
rs568965933 Chr.18: 722878-722878 on Build GRCh38
rs951740442 Chr.18: 723010-723010 on Build GRCh38
rs922834227 Chr.18: 722953-722953 on Build GRCh38
rs759058483 Chr.18: 722810-722810 on Build GRCh38
rs967511295 Chr.18: 723057-723057 on Build GRCh38
rs981518662 Chr.18: 723068-723068 on Build GRCh38
rs750535040 Chr.18: 722886-722886 on Build GRCh38
rs1027775316 Chr.18: 723009-723009 on Build GRCh38
rs549696578 Chr.18: 722956-722956 on Build GRCh38
rs557409541 Chr.18: 722818-722818 on Build GRCh38
rs904389175 Chr.18: 722892-722892 on Build GRCh38
rs889821218 Chr.18: 722835-722835 on Build GRCh38
rs386800143 Chr.18: 722917-722919 on Build GRCh38
rs1023009231 Chr.18: 723214-723214 on Build GRCh38
rs17516230 Chr.18: 723036-723036 on Build GRCh38
rs532113838 Chr.18: 722833-722833 on Build GRCh38
rs748458497 Chr.18: 723021-723021 on Build GRCh38
rs762268461 Chr.18: 722832-722832 on Build GRCh38
rs1039355348 Chr.18: 723272-723272 on Build GRCh38
rs549558075 Chr.18: 722859-722859 on Build GRCh38
rs569264257 Chr.18: 722899-722899 on Build GRCh38
rs576779903 Chr.18: 722895-722895 on Build GRCh38
rs541203119 Chr.18: 722954-722954 on Build GRCh38
rs991475104 Chr.18: 723061-723061 on Build GRCh38
rs574499187 Chr.18: 723055-723055 on Build GRCh38
rs17551255 Chr.18: 723070-723070 on Build GRCh38
rs17551248 Chr.18: 723069-723069 on Build GRCh38
rs11659287 Chr.18: 722821-722821 on Build GRCh38
rs558600964 Chr.18: 722958-722958 on Build GRCh38
rs544166989 Chr.18: 722896-722896 on Build GRCh38
rs1000041115 Chr.18: 722911-722911 on Build GRCh38
rs146943900 Chr.18: 722816-722816 on Build GRCh38
rs550438943 Chr.18: 722838-722838 on Build GRCh38
rs17551206 Chr.18: 722814-722814 on Build GRCh38
rs367596548 Chr.18: 723008-723008 on Build GRCh38
rs1017337296 Chr.18: 723017-723017 on Build GRCh38
rs560393090 Chr.18: 723269-723269 on Build GRCh38
rs1000413690 Chr.18: 722893-722893 on Build GRCh38
rs1024214167 Chr.18: 722928-722928 on Build GRCh38
rs920321396 Chr.18: 722901-722901 on Build GRCh38
rs915986930 Chr.18: 723062-723062 on Build GRCh38
rs17516216 Chr.18: 722875-722875 on Build GRCh38
rs971698858 Chr.18: 722932-722932 on Build GRCh38
rs940381969 Chr.18: 722874-722874 on Build GRCh38
rs550522453 Chr.18: 722919-722919 on Build GRCh38
rs552389307 Chr.18: 723046-723046 on Build GRCh38
rs781545266 Chr.18: 722920-722920 on Build GRCh38
rs562536640 Chr.18: 722855-722855 on Build GRCh38
rs879601196 Chr.18: 723069-723070 on Build GRCh38
rs56793935 Chr.18: 722918-722918 on Build GRCh38
rs957608283 Chr.18: 723108-723108 on Build GRCh38
rs1038658758 Chr.18: 723074-723075 on Build GRCh38
rs965205393 Chr.18: 722817-722817 on Build GRCh38

View Full Product Details