Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 736649-737126 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
YES1
Entrez Gene ID
7525
Gene Name
YES proto-oncogene 1, Src family tyrosine kinase
Gene Aliases
HsT441, P61-YES, Yes, c-yes
UniGene
Hs.194148
Gene Chromosome Location
Chr.18: 721592-812326 on Build GRCh38
Transcripts
NM_005433.3 XM_005258139.3 XM_017025960.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6919051 c.1184G>T p.R395L Chr.18: 736915-736915 on Build GRCh38
6976925 c.1180delC p.H394fs*11 Chr.18: 736919-736919 on Build GRCh38
6976115 c.1288C>T p.Q430* Chr.18: 736811-736811 on Build GRCh38
6028729 c.1176T>A p.Y392* Chr.18: 736923-736923 on Build GRCh38
6917082 c.1210G>T p.V404L Chr.18: 736889-736889 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759573463 Chr.18: 736977-736977 on Build GRCh38
rs761617938 Chr.18: 736784-736784 on Build GRCh38
rs771685902 Chr.18: 736794-736794 on Build GRCh38
rs928662953 Chr.18: 737081-737081 on Build GRCh38
rs9646493 Chr.18: 737083-737083 on Build GRCh38
rs760267714 Chr.18: 736828-736828 on Build GRCh38
rs750644702 Chr.18: 736738-736738 on Build GRCh38
rs781492606 Chr.18: 736939-736939 on Build GRCh38
rs746389614 Chr.18: 736943-736943 on Build GRCh38
rs200762886 Chr.18: 736815-736815 on Build GRCh38
rs766182225 Chr.18: 736829-736829 on Build GRCh38
rs530509859 Chr.18: 736707-736707 on Build GRCh38
rs747772462 Chr.18: 736945-736945 on Build GRCh38
rs1007155838 Chr.18: 736937-736937 on Build GRCh38
rs764251649 Chr.18: 736999-736999 on Build GRCh38
rs1055833656 Chr.18: 737087-737087 on Build GRCh38
rs972286959 Chr.18: 736788-736788 on Build GRCh38
rs28722141 Chr.18: 737036-737036 on Build GRCh38
rs776424908 Chr.18: 736966-736966 on Build GRCh38
rs770207233 Chr.18: 736771-736771 on Build GRCh38
rs550394545 Chr.18: 736757-736757 on Build GRCh38
rs774143672 Chr.18: 736783-736783 on Build GRCh38
rs981940089 Chr.18: 737052-737052 on Build GRCh38
rs746168302 Chr.18: 736769-736769 on Build GRCh38
rs751552293 Chr.18: 736904-736904 on Build GRCh38
rs370920392 Chr.18: 737028-737028 on Build GRCh38
rs763795278 Chr.18: 737043-737044 on Build GRCh38
rs529647749 Chr.18: 736949-736949 on Build GRCh38
rs758664744 Chr.18: 736883-736883 on Build GRCh38
rs910562293 Chr.18: 736915-736915 on Build GRCh38
rs768048101 Chr.18: 736873-736873 on Build GRCh38
rs201413906 Chr.18: 736899-736899 on Build GRCh38
rs770042876 Chr.18: 736944-736944 on Build GRCh38
rs188078358 Chr.18: 737005-737005 on Build GRCh38
rs754841809 Chr.18: 736876-736913 on Build GRCh38
rs960335761 Chr.18: 736742-736742 on Build GRCh38
rs759189339 Chr.18: 736872-736872 on Build GRCh38
rs140526381 Chr.18: 736931-736931 on Build GRCh38
rs373478419 Chr.18: 736964-736964 on Build GRCh38
rs757215837 Chr.18: 737001-737001 on Build GRCh38
rs370834164 Chr.18: 736996-736996 on Build GRCh38
rs760048255 Chr.18: 736960-736960 on Build GRCh38
rs762766824 Chr.18: 736997-736997 on Build GRCh38
rs780808894 Chr.18: 736984-736985 on Build GRCh38
rs1013286020 Chr.18: 737094-737094 on Build GRCh38
rs920742161 Chr.18: 736802-736802 on Build GRCh38
rs182611238 Chr.18: 736989-736989 on Build GRCh38
rs944649564 Chr.18: 736951-736951 on Build GRCh38
rs573487997 Chr.18: 737084-737084 on Build GRCh38
rs773025552 Chr.18: 736955-736955 on Build GRCh38
rs368550202 Chr.18: 736778-736778 on Build GRCh38
rs1002058308 Chr.18: 736720-736720 on Build GRCh38
rs753829901 Chr.18: 736833-736833 on Build GRCh38
rs772890030 Chr.18: 736812-736812 on Build GRCh38
rs765080545 Chr.18: 736874-736874 on Build GRCh38
rs747838528 Chr.18: 736997-736998 on Build GRCh38
rs997710873 Chr.18: 736817-736817 on Build GRCh38
rs199707539 Chr.18: 736830-736830 on Build GRCh38
rs1006129528 Chr.18: 736726-736726 on Build GRCh38
rs759877624 Chr.18: 736798-736798 on Build GRCh38
rs371854059 Chr.18: 736810-736810 on Build GRCh38
rs992326915 Chr.18: 736761-736761 on Build GRCh38
rs752901062 Chr.18: 736882-736882 on Build GRCh38
rs757380529 Chr.18: 736934-736934 on Build GRCh38
rs537147744 Chr.18: 737062-737062 on Build GRCh38
rs986350587 Chr.18: 736907-736907 on Build GRCh38

View Full Product Details