Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 5407618-5408123 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPB41L3
Entrez Gene ID
23136
Gene Name
erythrocyte membrane protein band 4.1 like 3
Gene Aliases
4.1B, DAL-1, DAL1
UniGene
Hs.213394
Gene Chromosome Location
Chr.18: 5392381-5630666 on Build GRCh38
Transcripts
NM_001281533.1 NM_001281534.1 NM_001281535.1 NM_012307.3 XM_011525623.2 XM_011525626.2 XM_011525628.2 XM_011525632.2 XM_011525634.2 XM_011525637.2 XM_017025618.1 XM_017025619.1 XM_017025620.1 XM_017025621.1 XM_017025622.1 XM_017025623.1 XM_017025624.1 XM_017025625.1 XM_017025626.1 XM_017025627.1 XM_017025628.1 XM_017025629.1 XM_017025630.1 XM_017025631.1 XM_017025632.1 XM_017025633.1 XM_017025634.1 XM_017025635.1 XM_017025636.1 XM_017025637.1 XM_017025638.1 XM_017025639.1 XM_017025640.1 XM_017025641.1 XM_017025642.1 XM_017025643.1 XM_017025644.1 XM_017025645.1 XM_017025646.1 XM_017025647.1 XM_017025648.1 XM_017025649.1 XM_017025650.1 XM_017025651.1 XM_017025652.1 XM_017025653.1 XM_017025654.1 XM_017025655.1 XM_017025656.1 XM_017025657.1 XM_017025658.1 XM_017025659.1 XM_017025660.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7238114 c.1620C>A p.D540E Chr.18: 5407731-5407731 on Build GRCh38
4072652 c.2157G>T p.Q719H Chr.18: 5407701-5407701 on Build GRCh38
5897974 c.1650+4G>A p.? Chr.18: 5407697-5407697 on Build GRCh38
4859935 c.1643A>T p.K548M Chr.18: 5407708-5407708 on Build GRCh38
7238113 c.2127C>A p.D709E Chr.18: 5407731-5407731 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781082218 Chr.18: 5407677-5407677 on Build GRCh38
rs189807727 Chr.18: 5407899-5407899 on Build GRCh38
rs988196614 Chr.18: 5408035-5408035 on Build GRCh38
rs763053679 Chr.18: 5407662-5407662 on Build GRCh38
rs901967072 Chr.18: 5407845-5407845 on Build GRCh38
rs375658482 Chr.18: 5407900-5407900 on Build GRCh38
rs116202296 Chr.18: 5408089-5408089 on Build GRCh38
rs1013738588 Chr.18: 5407990-5407990 on Build GRCh38
rs961162085 Chr.18: 5407677-5407681 on Build GRCh38
rs990469177 Chr.18: 5408038-5408038 on Build GRCh38
rs746117256 Chr.18: 5407659-5407659 on Build GRCh38
rs1023905195 Chr.18: 5408062-5408062 on Build GRCh38
rs969502415 Chr.18: 5408010-5408010 on Build GRCh38
rs977698614 Chr.18: 5408021-5408021 on Build GRCh38
rs979665772 Chr.18: 5408020-5408020 on Build GRCh38
rs751052050 Chr.18: 5407852-5407852 on Build GRCh38
rs548833731 Chr.18: 5408098-5408098 on Build GRCh38
rs777486254 Chr.18: 5407781-5407781 on Build GRCh38
rs747847955 Chr.18: 5407750-5407750 on Build GRCh38
rs375320631 Chr.18: 5407836-5407836 on Build GRCh38
rs751614866 Chr.18: 5407727-5407727 on Build GRCh38
rs200388785 Chr.18: 5407668-5407668 on Build GRCh38
rs900389240 Chr.18: 5407958-5407958 on Build GRCh38
rs752152351 Chr.18: 5407692-5407692 on Build GRCh38
rs757921907 Chr.18: 5407704-5407704 on Build GRCh38
rs957717963 Chr.18: 5408024-5408024 on Build GRCh38
rs933395408 Chr.18: 5407877-5407877 on Build GRCh38
rs2322093 Chr.18: 5407912-5407912 on Build GRCh38
rs529098192 Chr.18: 5407892-5407892 on Build GRCh38
rs2874686 Chr.18: 5407840-5407840 on Build GRCh38
rs762301420 Chr.18: 5407673-5407673 on Build GRCh38
rs780259904 Chr.18: 5407742-5407742 on Build GRCh38
rs942476442 Chr.18: 5408085-5408085 on Build GRCh38
rs144865558 Chr.18: 5407812-5407812 on Build GRCh38
rs770035891 Chr.18: 5407660-5407660 on Build GRCh38
rs752653285 Chr.18: 5408019-5408019 on Build GRCh38
rs1050917477 Chr.18: 5408060-5408060 on Build GRCh38
rs376928619 Chr.18: 5407739-5407739 on Build GRCh38
rs767907645 Chr.18: 5407679-5407679 on Build GRCh38
rs957686879 Chr.18: 5407716-5407716 on Build GRCh38
rs769398306 Chr.18: 5407661-5407662 on Build GRCh38
rs898171151 Chr.18: 5407975-5407975 on Build GRCh38
rs764903104 Chr.18: 5407691-5407691 on Build GRCh38
rs375614587 Chr.18: 5407663-5407663 on Build GRCh38
rs563043731 Chr.18: 5408080-5408080 on Build GRCh38
rs923253331 Chr.18: 5408036-5408036 on Build GRCh38
rs186968016 Chr.18: 5407882-5407882 on Build GRCh38
rs545982654 Chr.18: 5407983-5407983 on Build GRCh38
rs564439938 Chr.18: 5407987-5407987 on Build GRCh38
rs754757302 Chr.18: 5407662-5407664 on Build GRCh38
rs367590513 Chr.18: 5407794-5407794 on Build GRCh38
rs749343291 Chr.18: 5407787-5407787 on Build GRCh38
rs781131712 Chr.18: 5407658-5407658 on Build GRCh38
rs777406986 Chr.18: 5407706-5407706 on Build GRCh38
rs144889883 Chr.18: 5407835-5407835 on Build GRCh38
rs918696184 Chr.18: 5407817-5407817 on Build GRCh38
rs1006831550 Chr.18: 5407927-5407927 on Build GRCh38
rs113846843 Chr.18: 5407769-5407769 on Build GRCh38
rs1023828042 Chr.18: 5407995-5407995 on Build GRCh38
rs8086377 Chr.18: 5407967-5407967 on Build GRCh38
rs755827862 Chr.18: 5407753-5407754 on Build GRCh38
rs902269707 Chr.18: 5407984-5407984 on Build GRCh38
rs933444930 Chr.18: 5408058-5408058 on Build GRCh38
rs916814371 Chr.18: 5407746-5407746 on Build GRCh38
rs757231249 Chr.18: 5407734-5407734 on Build GRCh38
rs750774440 Chr.18: 5407685-5407685 on Build GRCh38
rs570754096 Chr.18: 5407733-5407733 on Build GRCh38
rs775458413 Chr.18: 5407661-5407661 on Build GRCh38
rs759129889 Chr.18: 5407689-5407689 on Build GRCh38
rs183962367 Chr.18: 5407797-5407797 on Build GRCh38
rs147538188 Chr.18: 5407922-5407922 on Build GRCh38
rs149578114 Chr.18: 5407735-5407735 on Build GRCh38
rs956952351 Chr.18: 5408028-5408028 on Build GRCh38

View Full Product Details