Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 8609843-8610366 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RAB12
Entrez Gene ID
201475
Gene Name
RAB12, member RAS oncogene family
Gene Aliases
-
UniGene
Hs.270074
Gene Chromosome Location
Chr.18: 8609428-8639383 on Build GRCh38
Transcripts
NM_001025300.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6761257 c.226+3A>G p.? Chr.18: 8609956-8609956 on Build GRCh38
708785 c.182G>A p.R61H Chr.18: 8609909-8609909 on Build GRCh38
4073603 c.194A>T p.D65V Chr.18: 8609921-8609921 on Build GRCh38
708786 c.174G>C p.L58L Chr.18: 8609901-8609901 on Build GRCh38
6274707 c.177G>T p.M59I Chr.18: 8609904-8609904 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs923935127 Chr.18: 8610248-8610248 on Build GRCh38
rs532225832 Chr.18: 8610016-8610016 on Build GRCh38
rs751605134 Chr.18: 8609877-8609877 on Build GRCh38
rs573683177 Chr.18: 8610098-8610098 on Build GRCh38
rs559516979 Chr.18: 8609899-8609899 on Build GRCh38
rs928522226 Chr.18: 8610207-8610207 on Build GRCh38
rs775217738 Chr.18: 8609996-8609996 on Build GRCh38
rs902134520 Chr.18: 8610068-8610068 on Build GRCh38
rs1049569499 Chr.18: 8610327-8610327 on Build GRCh38
rs976583195 Chr.18: 8610257-8610257 on Build GRCh38
rs756435385 Chr.18: 8609915-8609915 on Build GRCh38
rs948126684 Chr.18: 8610272-8610272 on Build GRCh38
rs1034736431 Chr.18: 8610186-8610186 on Build GRCh38
rs371278032 Chr.18: 8610236-8610236 on Build GRCh38
rs1010578281 Chr.18: 8610088-8610088 on Build GRCh38
rs747821699 Chr.18: 8609920-8609920 on Build GRCh38
rs932872844 Chr.18: 8610186-8610226 on Build GRCh38
rs770332482 Chr.18: 8610275-8610275 on Build GRCh38
rs568173526 Chr.18: 8609924-8609924 on Build GRCh38
rs542114342 Chr.18: 8609919-8609919 on Build GRCh38
rs549862802 Chr.18: 8609972-8609972 on Build GRCh38
rs968258982 Chr.18: 8610203-8610203 on Build GRCh38
rs1005244718 Chr.18: 8610150-8610150 on Build GRCh38
rs938603201 Chr.18: 8610212-8610212 on Build GRCh38
rs745752011 Chr.18: 8609992-8609992 on Build GRCh38
rs774469830 Chr.18: 8610009-8610009 on Build GRCh38
rs776451195 Chr.18: 8609977-8609977 on Build GRCh38
rs201144141 Chr.18: 8609901-8609901 on Build GRCh38
rs1023866473 Chr.18: 8610201-8610201 on Build GRCh38
rs757847504 Chr.18: 8609880-8609880 on Build GRCh38
rs534005643 Chr.18: 8609874-8609874 on Build GRCh38
rs531178191 Chr.18: 8609969-8609969 on Build GRCh38
rs766896123 Chr.18: 8610030-8610030 on Build GRCh38
rs911168344 Chr.18: 8610333-8610333 on Build GRCh38
rs1038076695 Chr.18: 8610347-8610347 on Build GRCh38
rs1052577427 Chr.18: 8610328-8610328 on Build GRCh38
rs565008699 Chr.18: 8609980-8609980 on Build GRCh38
rs762654283 Chr.18: 8610182-8610182 on Build GRCh38
rs770963507 Chr.18: 8609973-8609973 on Build GRCh38
rs935486838 Chr.18: 8610321-8610321 on Build GRCh38
rs766019492 Chr.18: 8610191-8610191 on Build GRCh38
rs1044200725 Chr.18: 8610289-8610289 on Build GRCh38
rs1049688717 Chr.18: 8610132-8610133 on Build GRCh38
rs764294417 Chr.18: 8610003-8610003 on Build GRCh38
rs956695676 Chr.18: 8610256-8610256 on Build GRCh38
rs547662432 Chr.18: 8610017-8610017 on Build GRCh38
rs530367140 Chr.18: 8609905-8609905 on Build GRCh38
rs780713558 Chr.18: 8610062-8610062 on Build GRCh38
rs1016060387 Chr.18: 8610176-8610176 on Build GRCh38
rs1038429077 Chr.18: 8610158-8610158 on Build GRCh38
rs761811568 Chr.18: 8610014-8610014 on Build GRCh38
rs924078882 Chr.18: 8609979-8609979 on Build GRCh38
rs941249145 Chr.18: 8610344-8610344 on Build GRCh38
rs745816788 Chr.18: 8610076-8610076 on Build GRCh38
rs916595274 Chr.18: 8610258-8610258 on Build GRCh38
rs896934679 Chr.18: 8610167-8610167 on Build GRCh38
rs948154326 Chr.18: 8609991-8609991 on Build GRCh38
rs570055981 Chr.18: 8610237-8610237 on Build GRCh38
rs984955088 Chr.18: 8610136-8610136 on Build GRCh38
rs1056506131 Chr.18: 8609968-8609968 on Build GRCh38
rs775248927 Chr.18: 8609978-8609992 on Build GRCh38
rs772848937 Chr.18: 8610127-8610127 on Build GRCh38
rs960867898 Chr.18: 8610198-8610198 on Build GRCh38
rs762398519 Chr.18: 8610227-8610227 on Build GRCh38
rs933832738 Chr.18: 8610326-8610326 on Build GRCh38
rs376808169 Chr.18: 8609925-8609925 on Build GRCh38
rs758193835 Chr.18: 8610044-8610044 on Build GRCh38
rs889684857 Chr.18: 8610144-8610144 on Build GRCh38
rs953106867 Chr.18: 8610101-8610101 on Build GRCh38
rs756727585 Chr.18: 8610060-8610060 on Build GRCh38
rs923354709 Chr.18: 8610324-8610324 on Build GRCh38
rs751169578 Chr.18: 8610056-8610056 on Build GRCh38
rs999741632 Chr.18: 8610026-8610026 on Build GRCh38
rs368352729 Chr.18: 8609993-8609993 on Build GRCh38
rs771648737 Chr.18: 8610197-8610197 on Build GRCh38
rs76182132 Chr.18: 8610036-8610036 on Build GRCh38
rs574080387 Chr.18: 8610299-8610299 on Build GRCh38
rs769658729 Chr.18: 8609944-8609944 on Build GRCh38
rs775694623 Chr.18: 8609871-8609871 on Build GRCh38
rs777557654 Chr.18: 8610046-8610046 on Build GRCh38
rs916795786 Chr.18: 8609950-8609950 on Build GRCh38
rs769699727 Chr.18: 8610091-8610091 on Build GRCh38
rs763213141 Chr.18: 8609998-8609998 on Build GRCh38
rs554225643 Chr.18: 8610223-8610223 on Build GRCh38

View Full Product Details