Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 9547459-9547971 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PPP4R1
Entrez Gene ID
9989
Gene Name
protein phosphatase 4 regulatory subunit 1
Gene Aliases
MEG1, PP4(Rmeg), PP4R1
UniGene
Hs.743236
Gene Chromosome Location
Chr.18: 9546791-9615274 on Build GRCh38
Transcripts
NM_001042388.2 NM_005134.3 NR_052003.1 XM_011525775.2 XM_011525776.1 XM_011525779.2 XM_017026105.1 XM_017026106.1 XM_017026107.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7278966 c.2698T>C p.L900L Chr.18: 9547944-9547944 on Build GRCh38
6754503 c.2838G>A p.A946A Chr.18: 9547804-9547804 on Build GRCh38
7385325 c.2736G>T p.Q912H Chr.18: 9547906-9547906 on Build GRCh38
5567042 c.2762G>A p.R921H Chr.18: 9547880-9547880 on Build GRCh38
7208964 c.2775C>G p.V925V Chr.18: 9547867-9547867 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754533832 Chr.18: 9547528-9547528 on Build GRCh38
rs528693789 Chr.18: 9547678-9547678 on Build GRCh38
rs552053501 Chr.18: 9547825-9547825 on Build GRCh38
rs558688008 Chr.18: 9547941-9547941 on Build GRCh38
rs965979365 Chr.18: 9547527-9547527 on Build GRCh38
rs769453585 Chr.18: 9547835-9547835 on Build GRCh38
rs914480059 Chr.18: 9547607-9547607 on Build GRCh38
rs757982905 Chr.18: 9547674-9547674 on Build GRCh38
rs547555358 Chr.18: 9547729-9547729 on Build GRCh38
rs999652290 Chr.18: 9547836-9547836 on Build GRCh38
rs780626264 Chr.18: 9547932-9547932 on Build GRCh38
rs368660563 Chr.18: 9547910-9547910 on Build GRCh38
rs533172260 Chr.18: 9547899-9547899 on Build GRCh38
rs765551012 Chr.18: 9547744-9547744 on Build GRCh38
rs747017763 Chr.18: 9547805-9547805 on Build GRCh38
rs1031221158 Chr.18: 9547522-9547522 on Build GRCh38
rs894196815 Chr.18: 9547702-9547702 on Build GRCh38
rs564697580 Chr.18: 9547675-9547675 on Build GRCh38
rs1002597596 Chr.18: 9547581-9547581 on Build GRCh38
rs201176037 Chr.18: 9547900-9547900 on Build GRCh38
rs1003018456 Chr.18: 9547824-9547824 on Build GRCh38
rs777542389 Chr.18: 9547804-9547804 on Build GRCh38
rs75728131 Chr.18: 9547541-9547541 on Build GRCh38
rs754302922 Chr.18: 9547701-9547701 on Build GRCh38
rs775273881 Chr.18: 9547837-9547837 on Build GRCh38
rs771958891 Chr.18: 9547793-9547793 on Build GRCh38
rs924442032 Chr.18: 9547656-9547656 on Build GRCh38
rs373632806 Chr.18: 9547873-9547873 on Build GRCh38
rs770380040 Chr.18: 9547809-9547809 on Build GRCh38
rs940597894 Chr.18: 9547658-9547658 on Build GRCh38
rs73939879 Chr.18: 9547787-9547787 on Build GRCh38
rs966280586 Chr.18: 9547849-9547849 on Build GRCh38
rs564089896 Chr.18: 9547610-9547610 on Build GRCh38
rs372199594 Chr.18: 9547885-9547885 on Build GRCh38
rs763225839 Chr.18: 9547847-9547847 on Build GRCh38
rs977752184 Chr.18: 9547633-9547633 on Build GRCh38
rs767660565 Chr.18: 9547880-9547880 on Build GRCh38
rs764557185 Chr.18: 9547854-9547854 on Build GRCh38
rs1031190064 Chr.18: 9547842-9547842 on Build GRCh38
rs376332808 Chr.18: 9547774-9547774 on Build GRCh38
rs766683536 Chr.18: 9547585-9547589 on Build GRCh38
rs932115480 Chr.18: 9547556-9547556 on Build GRCh38
rs1021204348 Chr.18: 9547936-9547936 on Build GRCh38
rs759126665 Chr.18: 9547826-9547826 on Build GRCh38
rs909088003 Chr.18: 9547651-9547651 on Build GRCh38
rs753012553 Chr.18: 9547747-9547747 on Build GRCh38
rs763463482 Chr.18: 9547748-9547748 on Build GRCh38
rs752173222 Chr.18: 9547761-9547761 on Build GRCh38
rs185380978 Chr.18: 9547896-9547896 on Build GRCh38
rs529707681 Chr.18: 9547756-9547756 on Build GRCh38
rs757195609 Chr.18: 9547920-9547920 on Build GRCh38
rs186768580 Chr.18: 9547730-9547730 on Build GRCh38
rs752380273 Chr.18: 9547627-9547627 on Build GRCh38
rs958345218 Chr.18: 9547598-9547598 on Build GRCh38
rs199567079 Chr.18: 9547818-9547818 on Build GRCh38
rs757250422 Chr.18: 9547762-9547762 on Build GRCh38
rs750534530 Chr.18: 9547881-9547881 on Build GRCh38
rs1050456827 Chr.18: 9547765-9547765 on Build GRCh38
rs545103595 Chr.18: 9547660-9547660 on Build GRCh38
rs968701038 Chr.18: 9547618-9547619 on Build GRCh38
rs938297241 Chr.18: 9547676-9547676 on Build GRCh38
rs746834228 Chr.18: 9547773-9547773 on Build GRCh38
rs192699370 Chr.18: 9547775-9547775 on Build GRCh38
rs778348288 Chr.18: 9547540-9547540 on Build GRCh38
rs367655172 Chr.18: 9547882-9547882 on Build GRCh38
rs774695430 Chr.18: 9547872-9547872 on Build GRCh38
rs781095018 Chr.18: 9547661-9547661 on Build GRCh38
rs991142493 Chr.18: 9547600-9547600 on Build GRCh38
rs985013559 Chr.18: 9547582-9547582 on Build GRCh38
rs1055495814 Chr.18: 9547696-9547696 on Build GRCh38
rs747908606 Chr.18: 9547791-9547791 on Build GRCh38
rs764498432 Chr.18: 9547755-9547755 on Build GRCh38

View Full Product Details